Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 118838088-118838638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTWD2
Entrez Gene ID
285605
Gene Name
DTW domain containing 2
Gene Aliases
-
UniGene
Hs.655891
Gene Chromosome Location
Chr.5: 118836074-118989938 on Build GRCh38
Transcripts
NM_001308081.1 NM_173666.3 XM_011543338.2 XM_011543340.2

SNP Detail

SNP ID Chromosome Location
rs541266562 Chr.5: 118838467-118838467 on Build GRCh38
rs951601986 Chr.5: 118838189-118838190 on Build GRCh38
rs565591338 Chr.5: 118838378-118838378 on Build GRCh38
rs556852779 Chr.5: 118838201-118838201 on Build GRCh38
rs908723318 Chr.5: 118838383-118838393 on Build GRCh38
rs12654866 Chr.5: 118838375-118838375 on Build GRCh38
rs555217488 Chr.5: 118838471-118838471 on Build GRCh38
rs116984050 Chr.5: 118838165-118838165 on Build GRCh38
rs371004417 Chr.5: 118838280-118838280 on Build GRCh38
rs17144715 Chr.5: 118838308-118838308 on Build GRCh38
rs945938425 Chr.5: 118838353-118838353 on Build GRCh38
rs1055796973 Chr.5: 118838605-118838605 on Build GRCh38
rs76166124 Chr.5: 118838574-118838574 on Build GRCh38
rs111515980 Chr.5: 118838417-118838417 on Build GRCh38
rs913056872 Chr.5: 118838550-118838550 on Build GRCh38
rs186042316 Chr.5: 118838604-118838604 on Build GRCh38
rs557307636 Chr.5: 118838569-118838569 on Build GRCh38
rs937461978 Chr.5: 118838381-118838381 on Build GRCh38
rs1019008383 Chr.5: 118838235-118838235 on Build GRCh38
rs957108261 Chr.5: 118838535-118838535 on Build GRCh38
rs770352031 Chr.5: 118838438-118838438 on Build GRCh38
rs55821851 Chr.5: 118838480-118838480 on Build GRCh38
rs995709958 Chr.5: 118838399-118838399 on Build GRCh38
rs556628253 Chr.5: 118838189-118838189 on Build GRCh38
rs937306885 Chr.5: 118838592-118838592 on Build GRCh38
rs1047258560 Chr.5: 118838409-118838409 on Build GRCh38
rs576832718 Chr.5: 118838429-118838429 on Build GRCh38
rs187000149 Chr.5: 118838171-118838171 on Build GRCh38
rs796091600 Chr.5: 118838397-118838397 on Build GRCh38
rs1045762354 Chr.5: 118838373-118838373 on Build GRCh38
rs369463489 Chr.5: 118838551-118838554 on Build GRCh38
rs17144714 Chr.5: 118838286-118838286 on Build GRCh38
rs543838417 Chr.5: 118838568-118838568 on Build GRCh38
rs140479907 Chr.5: 118838545-118838548 on Build GRCh38
rs770102181 Chr.5: 118838321-118838321 on Build GRCh38
rs537076071 Chr.5: 118838416-118838416 on Build GRCh38
rs887390793 Chr.5: 118838411-118838411 on Build GRCh38
rs866871593 Chr.5: 118838184-118838184 on Build GRCh38
rs921784640 Chr.5: 118838293-118838293 on Build GRCh38
rs61189236 Chr.5: 118838393-118838393 on Build GRCh38
rs1034532268 Chr.5: 118838158-118838158 on Build GRCh38
rs553554023 Chr.5: 118838327-118838327 on Build GRCh38
rs182862169 Chr.5: 118838246-118838246 on Build GRCh38
rs907310268 Chr.5: 118838116-118838116 on Build GRCh38
rs781712771 Chr.5: 118838243-118838243 on Build GRCh38
rs748615129 Chr.5: 118838325-118838325 on Build GRCh38
rs759453324 Chr.5: 118838534-118838534 on Build GRCh38
rs1008882417 Chr.5: 118838415-118838415 on Build GRCh38
rs192341858 Chr.5: 118838195-118838195 on Build GRCh38
rs190012122 Chr.5: 118838609-118838609 on Build GRCh38
rs865865947 Chr.5: 118838514-118838514 on Build GRCh38
rs148781979 Chr.5: 118838142-118838142 on Build GRCh38
rs777428138 Chr.5: 118838357-118838357 on Build GRCh38
rs371604490 Chr.5: 118838328-118838328 on Build GRCh38
rs913183059 Chr.5: 118838334-118838334 on Build GRCh38

View Full Product Details