Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72132700-72133244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCC1
Entrez Gene ID
51008
Gene Name
activating signal cointegrator 1 complex subunit 1
Gene Aliases
ASC1p50, CGI-18, SMABF2, p50
UniGene
Hs.500007
Gene Chromosome Location
Chr.10: 72096032-72217134 on Build GRCh38
Transcripts
NM_001198798.2 NM_001198799.2 NM_001198800.2 NR_045564.1 XM_006717873.3 XM_011539841.2 XM_011539842.2 XM_017016291.1 XM_017016292.1 XM_017016293.1 XM_017016294.1 XM_017016295.1 XM_017016296.1 XM_017016297.1 XM_017016298.1 XM_017016299.1 XM_017016300.1 XM_017016301.1 XM_017016302.1 XM_017016303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6997029 c.846T>C p.V282V Chr.10: 72133166-72133166 on Build GRCh38
4924993 c.891A>G p.K297K Chr.10: 72133121-72133121 on Build GRCh38
3440323 c.766C>T p.R256* Chr.10: 72133162-72133162 on Build GRCh38
4602588 c.875A>G p.E292G Chr.10: 72133119-72133119 on Build GRCh38
3440322 c.850C>T p.R284* Chr.10: 72133162-72133162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919130833 Chr.10: 72132932-72132932 on Build GRCh38
rs558707663 Chr.10: 72132851-72132851 on Build GRCh38
rs748354891 Chr.10: 72133133-72133133 on Build GRCh38
rs113569227 Chr.10: 72133181-72133181 on Build GRCh38
rs773916542 Chr.10: 72133047-72133047 on Build GRCh38
rs752947666 Chr.10: 72133217-72133217 on Build GRCh38
rs959999296 Chr.10: 72132822-72132822 on Build GRCh38
rs926721329 Chr.10: 72132900-72132900 on Build GRCh38
rs1017322728 Chr.10: 72132758-72132758 on Build GRCh38
rs980905937 Chr.10: 72132865-72132865 on Build GRCh38
rs1022514452 Chr.10: 72132831-72132831 on Build GRCh38
rs1003697823 Chr.10: 72133159-72133159 on Build GRCh38
rs138593434 Chr.10: 72132761-72132761 on Build GRCh38
rs997616393 Chr.10: 72132764-72132764 on Build GRCh38
rs973086802 Chr.10: 72132884-72132884 on Build GRCh38
rs377549482 Chr.10: 72133017-72133017 on Build GRCh38
rs568759263 Chr.10: 72132723-72132723 on Build GRCh38
rs1010963533 Chr.10: 72132770-72132770 on Build GRCh38
rs1007326025 Chr.10: 72132732-72132732 on Build GRCh38
rs750332602 Chr.10: 72133096-72133097 on Build GRCh38
rs753803060 Chr.10: 72133058-72133058 on Build GRCh38
rs760488500 Chr.10: 72133053-72133053 on Build GRCh38
rs756007889 Chr.10: 72133142-72133143 on Build GRCh38
rs893911218 Chr.10: 72132765-72132765 on Build GRCh38
rs765897386 Chr.10: 72133046-72133046 on Build GRCh38
rs141611243 Chr.10: 72133162-72133162 on Build GRCh38
rs188420196 Chr.10: 72132763-72132763 on Build GRCh38
rs574845267 Chr.10: 72133049-72133049 on Build GRCh38
rs202208603 Chr.10: 72133041-72133041 on Build GRCh38
rs929261854 Chr.10: 72132944-72132944 on Build GRCh38
rs141996717 Chr.10: 72133185-72133185 on Build GRCh38
rs377614205 Chr.10: 72132928-72132928 on Build GRCh38
rs747513880 Chr.10: 72132774-72132774 on Build GRCh38
rs1035266617 Chr.10: 72132796-72132796 on Build GRCh38
rs778054296 Chr.10: 72133144-72133144 on Build GRCh38
rs751635146 Chr.10: 72133082-72133082 on Build GRCh38
rs963105935 Chr.10: 72132852-72132852 on Build GRCh38
rs943236533 Chr.10: 72133036-72133036 on Build GRCh38
rs371450934 Chr.10: 72133150-72133150 on Build GRCh38
rs770374343 Chr.10: 72132800-72132800 on Build GRCh38
rs780735859 Chr.10: 72132943-72132943 on Build GRCh38
rs200721922 Chr.10: 72133088-72133088 on Build GRCh38
rs374955301 Chr.10: 72133050-72133050 on Build GRCh38
rs779557354 Chr.10: 72133145-72133146 on Build GRCh38
rs939430918 Chr.10: 72132924-72132924 on Build GRCh38
rs758996240 Chr.10: 72133154-72133154 on Build GRCh38
rs771359121 Chr.10: 72133149-72133149 on Build GRCh38
rs377388692 Chr.10: 72133200-72133200 on Build GRCh38
rs142117888 Chr.10: 72133013-72133013 on Build GRCh38
rs192323420 Chr.10: 72132833-72132833 on Build GRCh38
rs773734112 Chr.10: 72133042-72133042 on Build GRCh38
rs77464863 Chr.10: 72132743-72132744 on Build GRCh38
rs567123290 Chr.10: 72132827-72132827 on Build GRCh38
rs1007543797 Chr.10: 72132732-72132732 on Build GRCh38
rs372834128 Chr.10: 72133043-72133043 on Build GRCh38
rs755134952 Chr.10: 72133099-72133099 on Build GRCh38
rs991190120 Chr.10: 72132836-72132836 on Build GRCh38
rs112217483 Chr.10: 72132982-72132982 on Build GRCh38
rs745569621 Chr.10: 72132961-72132961 on Build GRCh38
rs547877756 Chr.10: 72132793-72132793 on Build GRCh38
rs997267683 Chr.10: 72133060-72133060 on Build GRCh38
rs756286851 Chr.10: 72133222-72133223 on Build GRCh38
rs918992057 Chr.10: 72133220-72133220 on Build GRCh38
rs573849264 Chr.10: 72132731-72132731 on Build GRCh38
rs777009724 Chr.10: 72133026-72133026 on Build GRCh38
rs770170337 Chr.10: 72133134-72133134 on Build GRCh38
rs146370051 Chr.10: 72133059-72133059 on Build GRCh38
rs753511548 Chr.10: 72133187-72133187 on Build GRCh38
rs184912932 Chr.10: 72132984-72132984 on Build GRCh38
rs1011036993 Chr.10: 72133068-72133068 on Build GRCh38
rs1031332876 Chr.10: 72132789-72132789 on Build GRCh38
rs565054047 Chr.10: 72133210-72133210 on Build GRCh38
rs968224915 Chr.10: 72132848-72132848 on Build GRCh38
rs988398980 Chr.10: 72132798-72132798 on Build GRCh38
rs957211336 Chr.10: 72132795-72132795 on Build GRCh38
rs768398422 Chr.10: 72133191-72133191 on Build GRCh38
rs376799409 Chr.10: 72133076-72133076 on Build GRCh38
rs1033847535 Chr.10: 72133203-72133203 on Build GRCh38
rs763845791 Chr.10: 72133056-72133056 on Build GRCh38
rs781528301 Chr.10: 72133116-72133116 on Build GRCh38
rs909106068 Chr.10: 72132988-72132988 on Build GRCh38
rs748499250 Chr.10: 72133030-72133030 on Build GRCh38
rs769419047 Chr.10: 72133016-72133016 on Build GRCh38
rs748854995 Chr.10: 72133147-72133148 on Build GRCh38
rs748391019 Chr.10: 72132728-72132729 on Build GRCh38

View Full Product Details