Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124014547-124015055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF664
Entrez Gene ID
144348
Gene Name
zinc finger protein 664
Gene Aliases
ZFOC1, ZNF176
UniGene
Hs.524828
Gene Chromosome Location
Chr.12: 123973215-124015439 on Build GRCh38
Transcripts
NM_001204298.1 NM_152437.2 NM_001204299.1

Gene Symbol
ZNF664-FAM101A
Entrez Gene ID
100533183
Gene Name
filamin-interacting protein FAM101A
Gene Aliases
-
UniGene
Hs.432901
Gene Chromosome Location
Chr.12: 123973215-124316024 on Build GRCh38
Transcripts
NM_001204298.1 NM_152437.2 NM_001204299.1

SNP Detail

SNP ID Chromosome Location
rs948560773 Chr.12: 124014812-124014812 on Build GRCh38
rs1008444550 Chr.12: 124014753-124014753 on Build GRCh38
rs556370864 Chr.12: 124014631-124014631 on Build GRCh38
rs575464838 Chr.12: 124014759-124014762 on Build GRCh38
rs906841384 Chr.12: 124014649-124014649 on Build GRCh38
rs1048497 Chr.12: 124014995-124014995 on Build GRCh38
rs547850990 Chr.12: 124014933-124014933 on Build GRCh38
rs529723582 Chr.12: 124014903-124014903 on Build GRCh38
rs561257601 Chr.12: 124014725-124014725 on Build GRCh38
rs1045572332 Chr.12: 124014829-124014829 on Build GRCh38
rs757136313 Chr.12: 124014965-124014965 on Build GRCh38
rs931054027 Chr.12: 124014901-124014901 on Build GRCh38
rs1054284186 Chr.12: 124014583-124014583 on Build GRCh38
rs758381066 Chr.12: 124014686-124014686 on Build GRCh38
rs189254346 Chr.12: 124014626-124014626 on Build GRCh38
rs971337904 Chr.12: 124014928-124014928 on Build GRCh38
rs370474871 Chr.12: 124014758-124014758 on Build GRCh38
rs147146939 Chr.12: 124014820-124014820 on Build GRCh38
rs11549100 Chr.12: 124014784-124014784 on Build GRCh38
rs913424336 Chr.12: 124014723-124014723 on Build GRCh38
rs939963006 Chr.12: 124014857-124014857 on Build GRCh38
rs746910420 Chr.12: 124014786-124014786 on Build GRCh38
rs576925490 Chr.12: 124014616-124014616 on Build GRCh38
rs1034352175 Chr.12: 124015003-124015003 on Build GRCh38
rs928551426 Chr.12: 124014852-124014852 on Build GRCh38
rs149457507 Chr.12: 124014743-124014743 on Build GRCh38
rs184447845 Chr.12: 124014882-124014882 on Build GRCh38
rs777510899 Chr.12: 124014924-124014924 on Build GRCh38
rs546909772 Chr.12: 124014663-124014663 on Build GRCh38
rs990331467 Chr.12: 124014711-124014711 on Build GRCh38
rs1028461364 Chr.12: 124014693-124014693 on Build GRCh38
rs1028111637 Chr.12: 124014600-124014600 on Build GRCh38
rs34578644 Chr.12: 124014582-124014583 on Build GRCh38
rs574808185 Chr.12: 124014636-124014636 on Build GRCh38
rs759244279 Chr.12: 124014684-124014684 on Build GRCh38
rs898224289 Chr.12: 124014683-124014683 on Build GRCh38
rs532344281 Chr.12: 124014842-124014842 on Build GRCh38
rs193125383 Chr.12: 124014642-124014642 on Build GRCh38
rs777617805 Chr.12: 124014776-124014776 on Build GRCh38
rs566842082 Chr.12: 124014985-124014985 on Build GRCh38
rs752377515 Chr.12: 124014775-124014775 on Build GRCh38
rs958120345 Chr.12: 124014779-124014779 on Build GRCh38
rs958279047 Chr.12: 124015022-124015022 on Build GRCh38
rs1019817352 Chr.12: 124014981-124014981 on Build GRCh38
rs1049945187 Chr.12: 124014919-124014919 on Build GRCh38
rs1036693609 Chr.12: 124014662-124014662 on Build GRCh38
rs781344564 Chr.12: 124014991-124014991 on Build GRCh38
rs115682148 Chr.12: 124014782-124014782 on Build GRCh38
rs999791798 Chr.12: 124015006-124015006 on Build GRCh38
rs957732658 Chr.12: 124014695-124014695 on Build GRCh38
rs889577883 Chr.12: 124014920-124014920 on Build GRCh38
rs11549102 Chr.12: 124014694-124014694 on Build GRCh38
rs925306743 Chr.12: 124014778-124014778 on Build GRCh38
rs112374470 Chr.12: 124014645-124014645 on Build GRCh38
rs751526791 Chr.12: 124014963-124014963 on Build GRCh38
rs971390362 Chr.12: 124014673-124014673 on Build GRCh38
rs11549101 Chr.12: 124014859-124014859 on Build GRCh38
rs189334278 Chr.12: 124014894-124014894 on Build GRCh38
rs867283770 Chr.12: 124014756-124014756 on Build GRCh38
rs1024357793 Chr.12: 124014873-124014873 on Build GRCh38
rs1003824313 Chr.12: 124014654-124014654 on Build GRCh38
rs1032318975 Chr.12: 124015014-124015014 on Build GRCh38
rs1046881862 Chr.12: 124014828-124014828 on Build GRCh38
rs1050854343 Chr.12: 124014749-124014749 on Build GRCh38
rs34870150 Chr.12: 124014888-124014890 on Build GRCh38
rs764734818 Chr.12: 124014774-124014774 on Build GRCh38

View Full Product Details