Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 131849668-131850160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP17
Entrez Gene ID
4326
Gene Name
matrix metallopeptidase 17
Gene Aliases
MMP-17, MT4-MMP, MT4MMP, MTMMP4
UniGene
Hs.709245
Gene Chromosome Location
Chr.12: 131828393-131851771 on Build GRCh38
Transcripts
NM_016155.5 XM_011538355.2 XM_011538356.2 XM_011538357.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6029037 c.1389C>T p.H463H Chr.12: 131849986-131849986 on Build GRCh38
1662816 c.1252C>T p.P418S Chr.12: 131849849-131849849 on Build GRCh38
2228831 c.1296C>T p.G432G Chr.12: 131849893-131849893 on Build GRCh38
7282772 c.1458C>A p.S486S Chr.12: 131850055-131850055 on Build GRCh38
7212125 c.1358G>A p.R453H Chr.12: 131849955-131849955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759280700 Chr.12: 131849859-131849859 on Build GRCh38
rs138985754 Chr.12: 131849959-131849959 on Build GRCh38
rs777146432 Chr.12: 131849871-131849871 on Build GRCh38
rs61942402 Chr.12: 131850128-131850128 on Build GRCh38
rs1042323781 Chr.12: 131850118-131850118 on Build GRCh38
rs753922425 Chr.12: 131849970-131849971 on Build GRCh38
rs770780629 Chr.12: 131849825-131849825 on Build GRCh38
rs1056170031 Chr.12: 131849713-131849713 on Build GRCh38
rs771294254 Chr.12: 131850071-131850071 on Build GRCh38
rs373834406 Chr.12: 131849884-131849884 on Build GRCh38
rs148740722 Chr.12: 131849824-131849824 on Build GRCh38
rs770120572 Chr.12: 131850076-131850076 on Build GRCh38
rs780900080 Chr.12: 131849833-131849833 on Build GRCh38
rs376752086 Chr.12: 131849793-131849793 on Build GRCh38
rs766999563 Chr.12: 131850041-131850041 on Build GRCh38
rs530441970 Chr.12: 131849946-131849946 on Build GRCh38
rs371193240 Chr.12: 131849923-131849923 on Build GRCh38
rs761126906 Chr.12: 131849870-131849886 on Build GRCh38
rs909377405 Chr.12: 131849714-131849714 on Build GRCh38
rs752214247 Chr.12: 131849812-131849812 on Build GRCh38
rs867479923 Chr.12: 131849801-131849801 on Build GRCh38
rs542923780 Chr.12: 131849756-131849756 on Build GRCh38
rs748925636 Chr.12: 131849762-131849762 on Build GRCh38
rs762447358 Chr.12: 131849955-131849955 on Build GRCh38
rs772654870 Chr.12: 131849917-131849917 on Build GRCh38
rs201494330 Chr.12: 131850048-131850048 on Build GRCh38
rs942161437 Chr.12: 131849727-131849727 on Build GRCh38
rs755256179 Chr.12: 131850055-131850055 on Build GRCh38
rs188513915 Chr.12: 131849773-131849773 on Build GRCh38
rs201986641 Chr.12: 131849787-131849787 on Build GRCh38
rs897891102 Chr.12: 131849750-131849750 on Build GRCh38
rs773917004 Chr.12: 131850004-131850004 on Build GRCh38
rs779153151 Chr.12: 131849987-131849987 on Build GRCh38
rs752274684 Chr.12: 131849864-131849864 on Build GRCh38
rs746865852 Chr.12: 131849779-131849779 on Build GRCh38
rs199948694 Chr.12: 131849869-131849869 on Build GRCh38
rs768594211 Chr.12: 131849852-131849852 on Build GRCh38
rs375097043 Chr.12: 131849766-131849766 on Build GRCh38
rs534877096 Chr.12: 131850030-131850030 on Build GRCh38
rs373705742 Chr.12: 131849761-131849761 on Build GRCh38
rs142421137 Chr.12: 131849834-131849834 on Build GRCh38
rs953901079 Chr.12: 131849822-131849822 on Build GRCh38
rs932236942 Chr.12: 131849691-131849691 on Build GRCh38
rs759464824 Chr.12: 131850107-131850107 on Build GRCh38
rs774046927 Chr.12: 131850078-131850078 on Build GRCh38
rs537443721 Chr.12: 131850038-131850038 on Build GRCh38
rs1016615677 Chr.12: 131850018-131850018 on Build GRCh38
rs573940179 Chr.12: 131850080-131850080 on Build GRCh38
rs982284232 Chr.12: 131850067-131850067 on Build GRCh38
rs772535826 Chr.12: 131850083-131850083 on Build GRCh38
rs749455826 Chr.12: 131849891-131849891 on Build GRCh38
rs984171158 Chr.12: 131850006-131850006 on Build GRCh38
rs766184246 Chr.12: 131850090-131850090 on Build GRCh38
rs760678952 Chr.12: 131850089-131850089 on Build GRCh38
rs753775447 Chr.12: 131850093-131850093 on Build GRCh38
rs369853750 Chr.12: 131849774-131849774 on Build GRCh38
rs34032150 Chr.12: 131849750-131849750 on Build GRCh38
rs1048827285 Chr.12: 131850024-131850024 on Build GRCh38
rs566139384 Chr.12: 131849742-131849742 on Build GRCh38
rs967809477 Chr.12: 131849847-131849847 on Build GRCh38
rs764211838 Chr.12: 131849932-131849933 on Build GRCh38
rs191276885 Chr.12: 131849995-131849995 on Build GRCh38
rs1009774126 Chr.12: 131849826-131849826 on Build GRCh38
rs1051140424 Chr.12: 131849709-131849709 on Build GRCh38
rs377440987 Chr.12: 131849853-131849853 on Build GRCh38
rs200355949 Chr.12: 131850031-131850031 on Build GRCh38
rs753525649 Chr.12: 131850053-131850053 on Build GRCh38
rs751976646 Chr.12: 131849795-131849795 on Build GRCh38
rs902058065 Chr.12: 131850070-131850070 on Build GRCh38
rs1012867498 Chr.12: 131849734-131849734 on Build GRCh38
rs145624993 Chr.12: 131849885-131849885 on Build GRCh38
rs777818507 Chr.12: 131849818-131849818 on Build GRCh38
rs758874268 Chr.12: 131849754-131849754 on Build GRCh38
rs1029003086 Chr.12: 131849938-131849938 on Build GRCh38
rs879200974 Chr.12: 131850043-131850043 on Build GRCh38
rs558467638 Chr.12: 131849759-131849759 on Build GRCh38
rs747565167 Chr.12: 131850064-131850064 on Build GRCh38
rs140213729 Chr.12: 131849858-131849858 on Build GRCh38
rs747470090 Chr.12: 131850011-131850011 on Build GRCh38
rs752130571 Chr.12: 131850117-131850117 on Build GRCh38
rs535362767 Chr.12: 131849745-131849745 on Build GRCh38
rs139162290 Chr.12: 131849915-131849915 on Build GRCh38
rs374439053 Chr.12: 131849954-131849954 on Build GRCh38
rs893690119 Chr.12: 131849957-131849957 on Build GRCh38
rs559536799 Chr.12: 131849870-131849870 on Build GRCh38
rs755994923 Chr.12: 131849975-131849975 on Build GRCh38
rs771335418 Chr.12: 131850022-131850022 on Build GRCh38
rs761152977 Chr.12: 131849942-131849942 on Build GRCh38
rs1028458373 Chr.12: 131849993-131849993 on Build GRCh38
rs775462466 Chr.12: 131849836-131849836 on Build GRCh38
rs748036032 Chr.12: 131849769-131849769 on Build GRCh38
rs1036646337 Chr.12: 131849897-131849897 on Build GRCh38
rs767977765 Chr.12: 131849798-131849798 on Build GRCh38
rs777937587 Chr.12: 131850059-131850059 on Build GRCh38
rs764985233 Chr.12: 131849863-131849863 on Build GRCh38
rs776900236 Chr.12: 131849786-131849786 on Build GRCh38
rs117508750 Chr.12: 131849983-131849983 on Build GRCh38
rs750337417 Chr.12: 131849971-131849971 on Build GRCh38
rs751504228 Chr.12: 131849877-131849877 on Build GRCh38
rs771573837 Chr.12: 131850081-131850081 on Build GRCh38
rs905186004 Chr.12: 131849920-131849920 on Build GRCh38
rs774503695 Chr.12: 131849893-131849893 on Build GRCh38
rs757130498 Chr.12: 131849878-131849878 on Build GRCh38
rs149055160 Chr.12: 131850037-131850037 on Build GRCh38
rs149877702 Chr.12: 131849970-131849970 on Build GRCh38
rs939083467 Chr.12: 131849700-131849700 on Build GRCh38
rs746259956 Chr.12: 131850075-131850075 on Build GRCh38
rs762316780 Chr.12: 131849796-131849796 on Build GRCh38
rs777087828 Chr.12: 131850074-131850074 on Build GRCh38
rs183469400 Chr.12: 131849698-131849698 on Build GRCh38
rs960613344 Chr.12: 131849989-131849989 on Build GRCh38
rs200132158 Chr.12: 131849808-131849808 on Build GRCh38
rs763674269 Chr.12: 131849956-131849956 on Build GRCh38
rs764380163 Chr.12: 131849755-131849755 on Build GRCh38
rs765122723 Chr.12: 131850108-131850108 on Build GRCh38
rs147503559 Chr.12: 131849851-131849851 on Build GRCh38
rs947772305 Chr.12: 131850019-131850019 on Build GRCh38
rs184269105 Chr.12: 131849996-131849996 on Build GRCh38
rs778295690 Chr.12: 131849809-131849809 on Build GRCh38
rs757465062 Chr.12: 131850111-131850111 on Build GRCh38
rs199551747 Chr.12: 131849800-131849800 on Build GRCh38
rs750425762 Chr.12: 131850047-131850047 on Build GRCh38
rs748369923 Chr.12: 131850058-131850058 on Build GRCh38
rs377623143 Chr.12: 131849890-131849890 on Build GRCh38
rs755093533 Chr.12: 131849986-131849986 on Build GRCh38
rs761360820 Chr.12: 131850040-131850040 on Build GRCh38
rs753213176 Chr.12: 131849752-131849752 on Build GRCh38
rs377417552 Chr.12: 131849794-131849794 on Build GRCh38
rs144808573 Chr.12: 131850056-131850056 on Build GRCh38
rs967934479 Chr.12: 131849974-131849974 on Build GRCh38
rs983813336 Chr.12: 131849875-131849875 on Build GRCh38
rs780987917 Chr.12: 131849879-131849879 on Build GRCh38
rs768090241 Chr.12: 131849794-131849795 on Build GRCh38
rs370586690 Chr.12: 131849857-131849857 on Build GRCh38
rs368841750 Chr.12: 131849960-131849960 on Build GRCh38
rs765307406 Chr.12: 131849792-131849792 on Build GRCh38
rs757820051 Chr.12: 131849816-131849816 on Build GRCh38
rs778478120 Chr.12: 131849767-131849767 on Build GRCh38
rs373295904 Chr.12: 131849780-131849780 on Build GRCh38
rs753279091 Chr.12: 131849814-131849814 on Build GRCh38

View Full Product Details