Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41379472-41379966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT34
Entrez Gene ID
3885
Gene Name
keratin 34
Gene Aliases
HA4, Ha-4, KRTHA4, hHa4
UniGene
Hs.296942
Gene Chromosome Location
Chr.17: 41377650-41383816 on Build GRCh38
Transcripts
NM_021013.3 XM_011524793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3517318 c.715-1G>A p.? Chr.17: 41379732-41379732 on Build GRCh38
2149951 c.757G>A p.V253M Chr.17: 41379689-41379689 on Build GRCh38
6556374 c.728T>A p.L243Q Chr.17: 41379718-41379718 on Build GRCh38
4066339 c.781G>A p.V261M Chr.17: 41379665-41379665 on Build GRCh38
4066338 c.879C>A p.T293T Chr.17: 41379476-41379476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778202313 Chr.17: 41379673-41379673 on Build GRCh38
rs752826260 Chr.17: 41379670-41379670 on Build GRCh38
rs555321847 Chr.17: 41379509-41379509 on Build GRCh38
rs778300807 Chr.17: 41379566-41379566 on Build GRCh38
rs369048328 Chr.17: 41379570-41379570 on Build GRCh38
rs773071645 Chr.17: 41379715-41379715 on Build GRCh38
rs772376036 Chr.17: 41379564-41379564 on Build GRCh38
rs763175166 Chr.17: 41379728-41379728 on Build GRCh38
rs369234151 Chr.17: 41379784-41379784 on Build GRCh38
rs2239710 Chr.17: 41379607-41379607 on Build GRCh38
rs773892443 Chr.17: 41379519-41379519 on Build GRCh38
rs146901800 Chr.17: 41379794-41379794 on Build GRCh38
rs768361702 Chr.17: 41379633-41379633 on Build GRCh38
rs760156674 Chr.17: 41379763-41379763 on Build GRCh38
rs750807257 Chr.17: 41379748-41379748 on Build GRCh38
rs781116002 Chr.17: 41379553-41379553 on Build GRCh38
rs939173039 Chr.17: 41379815-41379815 on Build GRCh38
rs371998384 Chr.17: 41379514-41379514 on Build GRCh38
rs149398304 Chr.17: 41379585-41379585 on Build GRCh38
rs150738879 Chr.17: 41379698-41379698 on Build GRCh38
rs139103580 Chr.17: 41379712-41379712 on Build GRCh38
rs780377995 Chr.17: 41379674-41379675 on Build GRCh38
rs909913394 Chr.17: 41379792-41379792 on Build GRCh38
rs748366260 Chr.17: 41379563-41379563 on Build GRCh38
rs545607533 Chr.17: 41379583-41379583 on Build GRCh38
rs772542576 Chr.17: 41379515-41379515 on Build GRCh38
rs764244872 Chr.17: 41379818-41379818 on Build GRCh38
rs763689125 Chr.17: 41379582-41379582 on Build GRCh38
rs747553598 Chr.17: 41379776-41379776 on Build GRCh38
rs770104112 Chr.17: 41379643-41379643 on Build GRCh38
rs200350721 Chr.17: 41379603-41379603 on Build GRCh38
rs202205358 Chr.17: 41379532-41379532 on Build GRCh38
rs773978057 Chr.17: 41379641-41379641 on Build GRCh38
rs758238341 Chr.17: 41379535-41379535 on Build GRCh38
rs749341338 Chr.17: 41379508-41379508 on Build GRCh38
rs781646309 Chr.17: 41379678-41379678 on Build GRCh38
rs774028684 Chr.17: 41379704-41379704 on Build GRCh38
rs376323832 Chr.17: 41379524-41379524 on Build GRCh38
rs369951520 Chr.17: 41379526-41379526 on Build GRCh38
rs751649477 Chr.17: 41379738-41379738 on Build GRCh38
rs756419013 Chr.17: 41379757-41379757 on Build GRCh38
rs550415508 Chr.17: 41379642-41379642 on Build GRCh38
rs1012034377 Chr.17: 41379877-41379877 on Build GRCh38
rs779076947 Chr.17: 41379599-41379599 on Build GRCh38
rs373836762 Chr.17: 41379554-41379554 on Build GRCh38
rs755052687 Chr.17: 41379592-41379592 on Build GRCh38
rs918491607 Chr.17: 41379804-41379804 on Build GRCh38
rs1044353874 Chr.17: 41379496-41379496 on Build GRCh38
rs749761044 Chr.17: 41379762-41379762 on Build GRCh38
rs368397378 Chr.17: 41379783-41379783 on Build GRCh38
rs560569006 Chr.17: 41379601-41379601 on Build GRCh38
rs763846445 Chr.17: 41379536-41379536 on Build GRCh38
rs370586990 Chr.17: 41379602-41379602 on Build GRCh38
rs758549987 Chr.17: 41379672-41379672 on Build GRCh38
rs774587309 Chr.17: 41379754-41379754 on Build GRCh38
rs767311692 Chr.17: 41379650-41379650 on Build GRCh38
rs757210175 Chr.17: 41379549-41379549 on Build GRCh38
rs748518457 Chr.17: 41379511-41379511 on Build GRCh38
rs140504517 Chr.17: 41379798-41379798 on Build GRCh38
rs747447820 Chr.17: 41379606-41379606 on Build GRCh38
rs774627374 Chr.17: 41379510-41379510 on Build GRCh38
rs768588112 Chr.17: 41379719-41379719 on Build GRCh38
rs201035931 Chr.17: 41379531-41379531 on Build GRCh38
rs866265957 Chr.17: 41379611-41379611 on Build GRCh38
rs138842774 Chr.17: 41379580-41379580 on Build GRCh38
rs147128610 Chr.17: 41379677-41379677 on Build GRCh38
rs760680685 Chr.17: 41379660-41379660 on Build GRCh38
rs759022880 Chr.17: 41379529-41379529 on Build GRCh38
rs192502620 Chr.17: 41379618-41379618 on Build GRCh38
rs144246640 Chr.17: 41379690-41379690 on Build GRCh38
rs201451902 Chr.17: 41379665-41379665 on Build GRCh38
rs775089425 Chr.17: 41379619-41379619 on Build GRCh38
rs764239666 Chr.17: 41379737-41379737 on Build GRCh38
rs537847537 Chr.17: 41379799-41379799 on Build GRCh38
rs759029766 Chr.17: 41379573-41379573 on Build GRCh38
rs767580018 Chr.17: 41379744-41379744 on Build GRCh38
rs753477854 Chr.17: 41379880-41379880 on Build GRCh38
rs140296098 Chr.17: 41379689-41379689 on Build GRCh38
rs747146219 Chr.17: 41379707-41379708 on Build GRCh38
rs781602979 Chr.17: 41379615-41379615 on Build GRCh38
rs556897929 Chr.17: 41379574-41379574 on Build GRCh38
rs1021935780 Chr.17: 41379683-41379683 on Build GRCh38
rs894913260 Chr.17: 41379848-41379848 on Build GRCh38
rs755071905 Chr.17: 41379557-41379557 on Build GRCh38
rs71383394 Chr.17: 41379905-41379905 on Build GRCh38
rs976855364 Chr.17: 41379751-41379751 on Build GRCh38
rs771574495 Chr.17: 41379713-41379713 on Build GRCh38
rs186796376 Chr.17: 41379579-41379579 on Build GRCh38
rs746359075 Chr.17: 41379617-41379617 on Build GRCh38
rs188110560 Chr.17: 41379759-41379759 on Build GRCh38
rs571289151 Chr.17: 41379908-41379908 on Build GRCh38
rs746047981 Chr.17: 41379820-41379820 on Build GRCh38
rs776698534 Chr.17: 41379721-41379721 on Build GRCh38
rs765996886 Chr.17: 41379835-41379835 on Build GRCh38
rs145049293 Chr.17: 41379578-41379578 on Build GRCh38
rs149921426 Chr.17: 41379716-41379716 on Build GRCh38
rs751810712 Chr.17: 41379676-41379676 on Build GRCh38
rs376395697 Chr.17: 41379569-41379569 on Build GRCh38
rs751316358 Chr.17: 41379537-41379537 on Build GRCh38
rs757526285 Chr.17: 41379743-41379743 on Build GRCh38
rs779053554 Chr.17: 41379561-41379561 on Build GRCh38
rs144798471 Chr.17: 41379586-41379586 on Build GRCh38
rs1031931561 Chr.17: 41379918-41379918 on Build GRCh38
rs778417368 Chr.17: 41379769-41379769 on Build GRCh38
rs750140886 Chr.17: 41379654-41379654 on Build GRCh38
rs903559388 Chr.17: 41379897-41379897 on Build GRCh38
rs777194157 Chr.17: 41379571-41379571 on Build GRCh38
rs780140580 Chr.17: 41379504-41379504 on Build GRCh38
rs144109786 Chr.17: 41379626-41379626 on Build GRCh38
rs748852948 Chr.17: 41379697-41379697 on Build GRCh38

View Full Product Details