Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130623476-130623987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUBP3
Entrez Gene ID
8939
Gene Name
far upstream element binding protein 3
Gene Aliases
FBP3
UniGene
Hs.673029
Gene Chromosome Location
Chr.9: 130579567-130638352 on Build GRCh38
Transcripts
NM_003934.1 XM_005272232.1 XM_006717313.2 XM_011519172.2 XM_011519173.2 XM_011519174.2 XM_017015269.1 XM_017015270.1 XM_017015271.1 XM_017015272.1 XM_017015273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685827 c.895A>C p.R299R Chr.9: 130623631-130623631 on Build GRCh38
6667733 c.925C>T p.R309W Chr.9: 130623661-130623661 on Build GRCh38
281263 c.887G>T p.S296I Chr.9: 130623623-130623623 on Build GRCh38
3780667 c.960G>A p.L320L Chr.9: 130623696-130623696 on Build GRCh38
6667737 c.712+3A>C p.? Chr.9: 130623631-130623631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020004567 Chr.9: 130623800-130623800 on Build GRCh38
rs767367068 Chr.9: 130623725-130623725 on Build GRCh38
rs375805534 Chr.9: 130623717-130623717 on Build GRCh38
rs761036333 Chr.9: 130623618-130623618 on Build GRCh38
rs1010384670 Chr.9: 130623562-130623562 on Build GRCh38
rs377086123 Chr.9: 130623646-130623646 on Build GRCh38
rs750187901 Chr.9: 130623659-130623659 on Build GRCh38
rs775045112 Chr.9: 130623705-130623705 on Build GRCh38
rs965460792 Chr.9: 130623915-130623915 on Build GRCh38
rs7043910 Chr.9: 130623570-130623570 on Build GRCh38
rs760450676 Chr.9: 130623727-130623727 on Build GRCh38
rs750508321 Chr.9: 130623596-130623597 on Build GRCh38
rs372531096 Chr.9: 130623597-130623597 on Build GRCh38
rs751494125 Chr.9: 130623562-130623562 on Build GRCh38
rs976815933 Chr.9: 130623927-130623927 on Build GRCh38
rs994806774 Chr.9: 130623519-130623519 on Build GRCh38
rs538761965 Chr.9: 130623757-130623757 on Build GRCh38
rs199887766 Chr.9: 130623743-130623743 on Build GRCh38
rs747392214 Chr.9: 130623691-130623691 on Build GRCh38
rs984888230 Chr.9: 130623643-130623643 on Build GRCh38
rs374085026 Chr.9: 130623756-130623756 on Build GRCh38
rs764077442 Chr.9: 130623560-130623560 on Build GRCh38
rs767448259 Chr.9: 130623657-130623657 on Build GRCh38
rs944048568 Chr.9: 130623505-130623505 on Build GRCh38
rs752840523 Chr.9: 130623735-130623735 on Build GRCh38
rs778211786 Chr.9: 130623737-130623737 on Build GRCh38
rs201855603 Chr.9: 130623690-130623690 on Build GRCh38
rs756320673 Chr.9: 130623588-130623588 on Build GRCh38
rs970582456 Chr.9: 130623913-130623913 on Build GRCh38
rs888633142 Chr.9: 130623543-130623543 on Build GRCh38
rs140357695 Chr.9: 130623518-130623518 on Build GRCh38
rs765351187 Chr.9: 130623812-130623812 on Build GRCh38
rs754103885 Chr.9: 130623662-130623662 on Build GRCh38
rs1001739869 Chr.9: 130623778-130623778 on Build GRCh38
rs757083479 Chr.9: 130623569-130623569 on Build GRCh38
rs576921954 Chr.9: 130623909-130623909 on Build GRCh38
rs200047797 Chr.9: 130623653-130623653 on Build GRCh38
rs768754951 Chr.9: 130623919-130623919 on Build GRCh38
rs145535460 Chr.9: 130623736-130623736 on Build GRCh38
rs187520242 Chr.9: 130623881-130623881 on Build GRCh38
rs762572286 Chr.9: 130623708-130623708 on Build GRCh38
rs779160859 Chr.9: 130623677-130623677 on Build GRCh38
rs918641905 Chr.9: 130623945-130623945 on Build GRCh38
rs761286072 Chr.9: 130623758-130623758 on Build GRCh38
rs762518172 Chr.9: 130623647-130623647 on Build GRCh38
rs763564658 Chr.9: 130623648-130623648 on Build GRCh38
rs911706005 Chr.9: 130623907-130623907 on Build GRCh38
rs1027181685 Chr.9: 130623521-130623521 on Build GRCh38
rs182842518 Chr.9: 130623779-130623779 on Build GRCh38
rs368455372 Chr.9: 130623517-130623517 on Build GRCh38
rs149024696 Chr.9: 130623858-130623858 on Build GRCh38
rs779214059 Chr.9: 130623734-130623734 on Build GRCh38
rs116711897 Chr.9: 130623726-130623726 on Build GRCh38
rs1041878280 Chr.9: 130623754-130623754 on Build GRCh38
rs11243999 Chr.9: 130623870-130623870 on Build GRCh38
rs749388894 Chr.9: 130623590-130623590 on Build GRCh38
rs971850983 Chr.9: 130623890-130623890 on Build GRCh38
rs772464787 Chr.9: 130623684-130623684 on Build GRCh38
rs764812675 Chr.9: 130623634-130623634 on Build GRCh38
rs763021421 Chr.9: 130623740-130623740 on Build GRCh38
rs768839853 Chr.9: 130623592-130623592 on Build GRCh38
rs145112455 Chr.9: 130623770-130623770 on Build GRCh38
rs143033281 Chr.9: 130623896-130623896 on Build GRCh38
rs771323314 Chr.9: 130623700-130623700 on Build GRCh38
rs1029694868 Chr.9: 130623822-130623822 on Build GRCh38
rs893292069 Chr.9: 130623526-130623526 on Build GRCh38
rs559530128 Chr.9: 130623958-130623958 on Build GRCh38
rs766162462 Chr.9: 130623728-130623728 on Build GRCh38
rs528138061 Chr.9: 130623656-130623656 on Build GRCh38
rs773484155 Chr.9: 130623614-130623614 on Build GRCh38
rs755878728 Chr.9: 130623661-130623661 on Build GRCh38
rs761244374 Chr.9: 130623718-130623718 on Build GRCh38
rs929977571 Chr.9: 130623954-130623954 on Build GRCh38
rs866771792 Chr.9: 130623925-130623925 on Build GRCh38
rs771249400 Chr.9: 130623738-130623738 on Build GRCh38
rs545676042 Chr.9: 130623952-130623952 on Build GRCh38
rs768300430 Chr.9: 130623750-130623750 on Build GRCh38
rs534731822 Chr.9: 130623787-130623787 on Build GRCh38
rs748202070 Chr.9: 130623678-130623678 on Build GRCh38
rs1016319869 Chr.9: 130623865-130623865 on Build GRCh38
rs368274658 Chr.9: 130623578-130623578 on Build GRCh38
rs1014000696 Chr.9: 130623791-130623791 on Build GRCh38
rs967466772 Chr.9: 130623811-130623811 on Build GRCh38
rs530394059 Chr.9: 130623733-130623733 on Build GRCh38
rs771603581 Chr.9: 130623763-130623763 on Build GRCh38
rs559087762 Chr.9: 130623605-130623605 on Build GRCh38
rs781559569 Chr.9: 130623742-130623742 on Build GRCh38
rs1006649842 Chr.9: 130623861-130623861 on Build GRCh38
rs780253616 Chr.9: 130623589-130623589 on Build GRCh38
rs1044741927 Chr.9: 130623504-130623504 on Build GRCh38

View Full Product Details