Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133098383-133098874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RALGDS
Entrez Gene ID
5900
Gene Name
ral guanine nucleotide dissociation stimulator
Gene Aliases
RGDS, RGF, RalGEF
UniGene
Hs.106185
Gene Chromosome Location
Chr.9: 133097720-133149220 on Build GRCh38
Transcripts
NM_001042368.2 NM_001271774.1 NM_001271775.1 NM_001271776.1 NM_006266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6313647 c.2589C>A p.N863K Chr.9: 133098743-133098743 on Build GRCh38
6762430 c.2503G>A p.A835T Chr.9: 133098742-133098742 on Build GRCh38
6313649 c.2586C>A p.N862K Chr.9: 133098743-133098743 on Build GRCh38
4382279 c.2809G>A p.V937I Chr.9: 133098736-133098736 on Build GRCh38
5824803 c.2487G>T p.L829L Chr.9: 133098758-133098758 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017364883 Chr.9: 133098797-133098797 on Build GRCh38
rs764693552 Chr.9: 133098600-133098600 on Build GRCh38
rs370951865 Chr.9: 133098606-133098606 on Build GRCh38
rs992090460 Chr.9: 133098437-133098437 on Build GRCh38
rs376030443 Chr.9: 133098642-133098642 on Build GRCh38
rs756428809 Chr.9: 133098719-133098719 on Build GRCh38
rs776277724 Chr.9: 133098572-133098572 on Build GRCh38
rs775223244 Chr.9: 133098768-133098768 on Build GRCh38
rs750439381 Chr.9: 133098718-133098718 on Build GRCh38
rs780791091 Chr.9: 133098745-133098745 on Build GRCh38
rs936978309 Chr.9: 133098537-133098537 on Build GRCh38
rs776063928 Chr.9: 133098684-133098684 on Build GRCh38
rs373844230 Chr.9: 133098541-133098541 on Build GRCh38
rs149083027 Chr.9: 133098666-133098666 on Build GRCh38
rs143209866 Chr.9: 133098667-133098667 on Build GRCh38
rs977864913 Chr.9: 133098840-133098840 on Build GRCh38
rs759013918 Chr.9: 133098597-133098597 on Build GRCh38
rs375607757 Chr.9: 133098710-133098710 on Build GRCh38
rs746897111 Chr.9: 133098742-133098742 on Build GRCh38
rs370328313 Chr.9: 133098563-133098563 on Build GRCh38
rs996566211 Chr.9: 133098628-133098628 on Build GRCh38
rs190743751 Chr.9: 133098602-133098602 on Build GRCh38
rs576652586 Chr.9: 133098802-133098802 on Build GRCh38
rs928626961 Chr.9: 133098842-133098842 on Build GRCh38
rs925436068 Chr.9: 133098482-133098482 on Build GRCh38
rs780129654 Chr.9: 133098734-133098734 on Build GRCh38
rs889701434 Chr.9: 133098461-133098461 on Build GRCh38
rs573407742 Chr.9: 133098407-133098407 on Build GRCh38
rs374088319 Chr.9: 133098582-133098582 on Build GRCh38
rs985963780 Chr.9: 133098512-133098512 on Build GRCh38
rs747935608 Chr.9: 133098652-133098652 on Build GRCh38
rs957917376 Chr.9: 133098559-133098559 on Build GRCh38
rs368805508 Chr.9: 133098775-133098775 on Build GRCh38
rs753085695 Chr.9: 133098596-133098596 on Build GRCh38
rs568299176 Chr.9: 133098622-133098622 on Build GRCh38
rs772780500 Chr.9: 133098546-133098546 on Build GRCh38
rs745773402 Chr.9: 133098737-133098737 on Build GRCh38
rs370690294 Chr.9: 133098645-133098645 on Build GRCh38
rs748133573 Chr.9: 133098536-133098536 on Build GRCh38
rs773295357 Chr.9: 133098558-133098558 on Build GRCh38
rs746054003 Chr.9: 133098610-133098610 on Build GRCh38
rs45512701 Chr.9: 133098455-133098455 on Build GRCh38
rs551848535 Chr.9: 133098514-133098514 on Build GRCh38
rs1040743629 Chr.9: 133098826-133098826 on Build GRCh38
rs886017 Chr.9: 133098713-133098713 on Build GRCh38
rs1026856324 Chr.9: 133098631-133098631 on Build GRCh38
rs752074040 Chr.9: 133098712-133098712 on Build GRCh38
rs147457502 Chr.9: 133098827-133098827 on Build GRCh38
rs774834956 Chr.9: 133098796-133098796 on Build GRCh38
rs761749654 Chr.9: 133098410-133098410 on Build GRCh38
rs115989046 Chr.9: 133098805-133098805 on Build GRCh38
rs531302066 Chr.9: 133098583-133098583 on Build GRCh38
rs781302534 Chr.9: 133098609-133098609 on Build GRCh38
rs777715610 Chr.9: 133098740-133098740 on Build GRCh38
rs764765524 Chr.9: 133098680-133098680 on Build GRCh38
rs760651358 Chr.9: 133098573-133098587 on Build GRCh38
rs899494916 Chr.9: 133098836-133098836 on Build GRCh38
rs769095384 Chr.9: 133098783-133098783 on Build GRCh38
rs145020652 Chr.9: 133098653-133098653 on Build GRCh38
rs377667079 Chr.9: 133098683-133098683 on Build GRCh38
rs993242065 Chr.9: 133098438-133098438 on Build GRCh38
rs745365387 Chr.9: 133098761-133098761 on Build GRCh38
rs560161218 Chr.9: 133098453-133098453 on Build GRCh38
rs760169037 Chr.9: 133098558-133098558 on Build GRCh38
rs1062356 Chr.9: 133098692-133098692 on Build GRCh38
rs773078165 Chr.9: 133098654-133098654 on Build GRCh38
rs375124486 Chr.9: 133098661-133098661 on Build GRCh38
rs766914011 Chr.9: 133098609-133098610 on Build GRCh38
rs1006486287 Chr.9: 133098406-133098406 on Build GRCh38
rs767685442 Chr.9: 133098716-133098716 on Build GRCh38
rs759279035 Chr.9: 133098677-133098677 on Build GRCh38
rs539826984 Chr.9: 133098736-133098736 on Build GRCh38
rs146412006 Chr.9: 133098743-133098743 on Build GRCh38
rs878911634 Chr.9: 133098436-133098436 on Build GRCh38
rs776167423 Chr.9: 133098673-133098673 on Build GRCh38
rs376268091 Chr.9: 133098766-133098766 on Build GRCh38
rs1021319433 Chr.9: 133098806-133098806 on Build GRCh38
rs1045217672 Chr.9: 133098464-133098464 on Build GRCh38
rs866093295 Chr.9: 133098839-133098839 on Build GRCh38

View Full Product Details