Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 63622637-63623171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
REEP3
Entrez Gene ID
221035
Gene Name
receptor accessory protein 3
Gene Aliases
C10orf74, Yip2b
UniGene
Hs.499833
Gene Chromosome Location
Chr.10: 63521353-63625123 on Build GRCh38
Transcripts
NM_001001330.2

SNP Detail

SNP ID Chromosome Location
rs575955403 Chr.10: 63622921-63622921 on Build GRCh38
rs913992469 Chr.10: 63623013-63623013 on Build GRCh38
rs561401306 Chr.10: 63622957-63622957 on Build GRCh38
rs797001549 Chr.10: 63622706-63622710 on Build GRCh38
rs763595790 Chr.10: 63623045-63623045 on Build GRCh38
rs564595176 Chr.10: 63623017-63623017 on Build GRCh38
rs920008697 Chr.10: 63622970-63622970 on Build GRCh38
rs907411808 Chr.10: 63623147-63623147 on Build GRCh38
rs931925018 Chr.10: 63623065-63623065 on Build GRCh38
rs72840212 Chr.10: 63623072-63623072 on Build GRCh38
rs533246032 Chr.10: 63623121-63623121 on Build GRCh38
rs983701628 Chr.10: 63622995-63622995 on Build GRCh38
rs1034349490 Chr.10: 63622952-63622952 on Build GRCh38
rs1005847987 Chr.10: 63623094-63623094 on Build GRCh38
rs925502490 Chr.10: 63622981-63622981 on Build GRCh38
rs1025560703 Chr.10: 63622956-63622956 on Build GRCh38
rs560776239 Chr.10: 63623132-63623132 on Build GRCh38
rs948265252 Chr.10: 63622739-63622739 on Build GRCh38
rs960789334 Chr.10: 63622936-63622936 on Build GRCh38
rs989067458 Chr.10: 63622667-63622667 on Build GRCh38
rs572762920 Chr.10: 63622806-63622806 on Build GRCh38
rs971401909 Chr.10: 63622769-63622769 on Build GRCh38
rs1052540707 Chr.10: 63622803-63622803 on Build GRCh38
rs1032755817 Chr.10: 63622853-63622853 on Build GRCh38
rs770024988 Chr.10: 63622836-63622836 on Build GRCh38
rs185339356 Chr.10: 63623035-63623035 on Build GRCh38
rs1010018159 Chr.10: 63623110-63623110 on Build GRCh38
rs944606894 Chr.10: 63623024-63623024 on Build GRCh38
rs981456478 Chr.10: 63622775-63622775 on Build GRCh38
rs1047600140 Chr.10: 63622823-63622823 on Build GRCh38
rs67568146 Chr.10: 63622706-63622707 on Build GRCh38
rs1038504070 Chr.10: 63622793-63622793 on Build GRCh38
rs565057580 Chr.10: 63622997-63622997 on Build GRCh38
rs920433754 Chr.10: 63622794-63622794 on Build GRCh38
rs1020508450 Chr.10: 63622929-63622929 on Build GRCh38
rs913480056 Chr.10: 63622688-63622688 on Build GRCh38
rs565145498 Chr.10: 63623105-63623105 on Build GRCh38
rs1010841449 Chr.10: 63622912-63622912 on Build GRCh38
rs1040002892 Chr.10: 63622832-63622832 on Build GRCh38
rs779781396 Chr.10: 63622705-63622706 on Build GRCh38
rs141425770 Chr.10: 63622947-63622947 on Build GRCh38
rs962061720 Chr.10: 63622946-63622946 on Build GRCh38
rs1014962660 Chr.10: 63623101-63623101 on Build GRCh38
rs148267458 Chr.10: 63622854-63622854 on Build GRCh38
rs1014687780 Chr.10: 63622946-63622946 on Build GRCh38
rs868860680 Chr.10: 63623068-63623068 on Build GRCh38
rs879118315 Chr.10: 63622703-63622703 on Build GRCh38
rs563897974 Chr.10: 63622824-63622824 on Build GRCh38
rs918181991 Chr.10: 63622697-63622697 on Build GRCh38
rs1028645848 Chr.10: 63622880-63622880 on Build GRCh38
rs1255454 Chr.10: 63622958-63622958 on Build GRCh38
rs181258901 Chr.10: 63622788-63622788 on Build GRCh38
rs1005816702 Chr.10: 63622923-63622928 on Build GRCh38
rs930468966 Chr.10: 63622971-63622971 on Build GRCh38
rs867306873 Chr.10: 63622731-63622731 on Build GRCh38
rs977918207 Chr.10: 63622779-63622779 on Build GRCh38
rs1013745006 Chr.10: 63623133-63623133 on Build GRCh38
rs980807354 Chr.10: 63622770-63622770 on Build GRCh38
rs900166910 Chr.10: 63623057-63623057 on Build GRCh38
rs1040243961 Chr.10: 63623043-63623043 on Build GRCh38
rs188639839 Chr.10: 63623136-63623136 on Build GRCh38
rs771611404 Chr.10: 63622706-63622711 on Build GRCh38
rs995877643 Chr.10: 63622850-63622850 on Build GRCh38
rs931656468 Chr.10: 63623062-63623062 on Build GRCh38
rs944346835 Chr.10: 63622826-63622826 on Build GRCh38
rs972045573 Chr.10: 63622941-63622941 on Build GRCh38

View Full Product Details