Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 21540484-21541031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBMY1A1
Entrez Gene ID
5940
Gene Name
RNA binding motif protein, Y-linked, family 1, member A1
Gene Aliases
RBM, RBM1, RBM2, RBMY, RBMY1C, YRRM1, YRRM2
UniGene
Hs.380450
Gene Chromosome Location
Chr.Y: 21534879-21559683 on Build GRCh38
Transcripts
NM_001320944.1 NM_001320945.1 NM_005058.3 XM_011531437.2 XM_011531438.1 XM_017030060.1

SNP Detail

SNP ID Chromosome Location
rs200614345 Chr.Y: 21540939-21540939 on Build GRCh38
rs760457230 Chr.Y: 21540825-21540825 on Build GRCh38
rs3868239 Chr.Y: 21540730-21540730 on Build GRCh38
rs334649 Chr.Y: 21541008-21541008 on Build GRCh38
rs3868238 Chr.Y: 21540566-21540566 on Build GRCh38
rs773045632 Chr.Y: 21540731-21540731 on Build GRCh38
rs767379779 Chr.Y: 21540717-21540717 on Build GRCh38

View Full Product Details