Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4544200-4544601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA6B
Entrez Gene ID
10501
Gene Name
semaphorin 6B
Gene Aliases
SEM-SEMA-Y, SEMA-VIB, SEMAN, semaZ
UniGene
Hs.465642
Gene Chromosome Location
Chr.19: 4542588-4581484 on Build GRCh38
Transcripts
NM_032108.3 XM_011527639.2 XM_011527640.2 XM_011527641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4405513 c.1883G>A p.R628Q Chr.19: 4544385-4544385 on Build GRCh38
6774881 c.1814C>T p.A605V Chr.19: 4544454-4544454 on Build GRCh38
4420116 c.1894C>T p.R632W Chr.19: 4544374-4544374 on Build GRCh38
6774882 c.1802C>T p.T601M Chr.19: 4544466-4544466 on Build GRCh38
6017138 c.1985C>T p.P662L Chr.19: 4544283-4544283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777859187 Chr.19: 4544538-4544538 on Build GRCh38
rs371352629 Chr.19: 4544289-4544289 on Build GRCh38
rs1033085752 Chr.19: 4544341-4544341 on Build GRCh38
rs776582297 Chr.19: 4544564-4544564 on Build GRCh38
rs771924909 Chr.19: 4544453-4544453 on Build GRCh38
rs1015238601 Chr.19: 4544271-4544271 on Build GRCh38
rs768709230 Chr.19: 4544473-4544473 on Build GRCh38
rs1041531752 Chr.19: 4544238-4544238 on Build GRCh38
rs761365296 Chr.19: 4544484-4544484 on Build GRCh38
rs1014377568 Chr.19: 4544480-4544480 on Build GRCh38
rs777997784 Chr.19: 4544447-4544447 on Build GRCh38
rs1049623909 Chr.19: 4544263-4544263 on Build GRCh38
rs776321997 Chr.19: 4544482-4544482 on Build GRCh38
rs1007917533 Chr.19: 4544579-4544579 on Build GRCh38
rs753385574 Chr.19: 4544377-4544377 on Build GRCh38
rs932429745 Chr.19: 4544563-4544563 on Build GRCh38
rs758743385 Chr.19: 4544388-4544388 on Build GRCh38
rs747645668 Chr.19: 4544567-4544567 on Build GRCh38
rs749467548 Chr.19: 4544539-4544539 on Build GRCh38
rs771923704 Chr.19: 4544334-4544334 on Build GRCh38
rs148753710 Chr.19: 4544435-4544435 on Build GRCh38
rs528232558 Chr.19: 4544257-4544257 on Build GRCh38
rs751999514 Chr.19: 4544414-4544414 on Build GRCh38
rs763869846 Chr.19: 4544227-4544227 on Build GRCh38
rs560845019 Chr.19: 4544518-4544518 on Build GRCh38
rs770168650 Chr.19: 4544508-4544509 on Build GRCh38
rs888394318 Chr.19: 4544269-4544269 on Build GRCh38
rs1005158963 Chr.19: 4544270-4544270 on Build GRCh38
rs750796918 Chr.19: 4544277-4544277 on Build GRCh38
rs768685185 Chr.19: 4544307-4544307 on Build GRCh38
rs932523803 Chr.19: 4544261-4544261 on Build GRCh38
rs981283655 Chr.19: 4544444-4544444 on Build GRCh38
rs564925636 Chr.19: 4544301-4544301 on Build GRCh38
rs764821155 Chr.19: 4544493-4544493 on Build GRCh38
rs764913336 Chr.19: 4544386-4544386 on Build GRCh38
rs767987328 Chr.19: 4544512-4544512 on Build GRCh38
rs775526111 Chr.19: 4544337-4544337 on Build GRCh38
rs755550248 Chr.19: 4544507-4544507 on Build GRCh38
rs747115338 Chr.19: 4544302-4544302 on Build GRCh38
rs761439397 Chr.19: 4544385-4544385 on Build GRCh38
rs763638386 Chr.19: 4544371-4544371 on Build GRCh38
rs560506671 Chr.19: 4544231-4544231 on Build GRCh38
rs756177194 Chr.19: 4544536-4544536 on Build GRCh38
rs775579653 Chr.19: 4544470-4544470 on Build GRCh38
rs748258087 Chr.19: 4544432-4544432 on Build GRCh38
rs760752799 Chr.19: 4544471-4544471 on Build GRCh38
rs768764268 Chr.19: 4544553-4544553 on Build GRCh38
rs915931321 Chr.19: 4544378-4544378 on Build GRCh38
rs781312157 Chr.19: 4544426-4544426 on Build GRCh38
rs970883516 Chr.19: 4544278-4544278 on Build GRCh38
rs781400191 Chr.19: 4544311-4544311 on Build GRCh38
rs1013970495 Chr.19: 4544276-4544276 on Build GRCh38
rs755066405 Chr.19: 4544420-4544420 on Build GRCh38
rs780571049 Chr.19: 4544393-4544393 on Build GRCh38
rs766674899 Chr.19: 4544503-4544503 on Build GRCh38
rs991503722 Chr.19: 4544374-4544374 on Build GRCh38
rs757537761 Chr.19: 4544547-4544547 on Build GRCh38
rs550558046 Chr.19: 4544333-4544333 on Build GRCh38
rs748265038 Chr.19: 4544317-4544317 on Build GRCh38
rs200785087 Chr.19: 4544345-4544345 on Build GRCh38
rs898224051 Chr.19: 4544562-4544562 on Build GRCh38
rs924672944 Chr.19: 4544570-4544570 on Build GRCh38
rs945451920 Chr.19: 4544234-4544234 on Build GRCh38
rs770046891 Chr.19: 4544326-4544326 on Build GRCh38
rs769282597 Chr.19: 4544568-4544568 on Build GRCh38
rs531242138 Chr.19: 4544546-4544546 on Build GRCh38
rs977920134 Chr.19: 4544382-4544382 on Build GRCh38
rs200580601 Chr.19: 4544330-4544330 on Build GRCh38
rs763234419 Chr.19: 4544497-4544497 on Build GRCh38
rs750054428 Chr.19: 4544495-4544495 on Build GRCh38
rs745679612 Chr.19: 4544450-4544450 on Build GRCh38
rs779203344 Chr.19: 4544274-4544274 on Build GRCh38
rs752738677 Chr.19: 4544534-4544534 on Build GRCh38

View Full Product Details