Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44240896-44241431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICOSLG
Entrez Gene ID
23308
Gene Name
inducible T-cell costimulator ligand
Gene Aliases
B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, LICOS
UniGene
Hs.14155
Gene Chromosome Location
Chr.21: 44216980-44241447 on Build GRCh38
Transcripts
NM_001283050.1 NM_001283051.1 NM_001283052.1 NM_015259.5 XM_011529514.2 XM_011529515.2 XM_017028310.1 XM_006723899.2 XM_006723900.2 XM_011546078.2 XM_011546079.1

Gene Symbol
LOC102723996
Entrez Gene ID
102723996
Gene Name
ICOS ligand
Gene Aliases
-
UniGene
Hs.14155
Gene Chromosome Location
Chr.21: 5022044-5046678 on Build GRCh38
Transcripts
NM_001283050.1 NM_001283051.1 NM_001283052.1 NM_015259.5 XM_011529514.2 XM_011529515.2 XM_017028310.1 XM_006723899.2 XM_006723900.2 XM_011546078.2 XM_011546079.1

SNP Detail

SNP ID Chromosome Location
rs563599848 Chr.21: 44241184-44241184 on Build GRCh38
rs533202426 Chr.21: 44241128-44241128 on Build GRCh38
rs370529368 Chr.21: 44241056-44241056 on Build GRCh38
rs866905055 Chr.21: 44241118-44241118 on Build GRCh38
rs953430338 Chr.21: 44241212-44241213 on Build GRCh38
rs540319342 Chr.21: 44241045-44241045 on Build GRCh38
rs528194677 Chr.21: 44241407-44241407 on Build GRCh38
rs567121926 Chr.21: 44241385-44241385 on Build GRCh38
rs559508028 Chr.21: 44241079-44241079 on Build GRCh38
rs943938239 Chr.21: 44241239-44241239 on Build GRCh38
rs1053850100 Chr.21: 44241286-44241286 on Build GRCh38
rs746118567 Chr.21: 44241400-44241400 on Build GRCh38
rs894906463 Chr.21: 44241299-44241299 on Build GRCh38
rs963948521 Chr.21: 44241379-44241379 on Build GRCh38
rs973810743 Chr.21: 44241186-44241186 on Build GRCh38
rs187365981 Chr.21: 44241305-44241305 on Build GRCh38
rs373126838 Chr.21: 44241312-44241312 on Build GRCh38
rs35622408 Chr.21: 44240935-44240936 on Build GRCh38
rs557275075 Chr.21: 44240934-44240934 on Build GRCh38
rs912376794 Chr.21: 44241220-44241220 on Build GRCh38
rs1042094493 Chr.21: 44241043-44241043 on Build GRCh38
rs939062853 Chr.21: 44241002-44241012 on Build GRCh38
rs923852011 Chr.21: 44241265-44241265 on Build GRCh38
rs922005399 Chr.21: 44240967-44240967 on Build GRCh38
rs1012396619 Chr.21: 44241116-44241116 on Build GRCh38
rs182699570 Chr.21: 44241163-44241163 on Build GRCh38
rs963446365 Chr.21: 44241180-44241180 on Build GRCh38
rs1005049382 Chr.21: 44241169-44241169 on Build GRCh38
rs575529616 Chr.21: 44240936-44240936 on Build GRCh38
rs970938401 Chr.21: 44240932-44240932 on Build GRCh38
rs75939338 Chr.21: 44240946-44240946 on Build GRCh38
rs554651249 Chr.21: 44241006-44241006 on Build GRCh38
rs906674245 Chr.21: 44241324-44241324 on Build GRCh38
rs936373173 Chr.21: 44241285-44241285 on Build GRCh38
rs942109193 Chr.21: 44240948-44240948 on Build GRCh38
rs573303731 Chr.21: 44241033-44241033 on Build GRCh38
rs1000931828 Chr.21: 44241060-44241060 on Build GRCh38
rs1015217428 Chr.21: 44241371-44241371 on Build GRCh38
rs1049572676 Chr.21: 44240990-44240990 on Build GRCh38
rs1015350159 Chr.21: 44241171-44241171 on Build GRCh38
rs1005001569 Chr.21: 44241345-44241345 on Build GRCh38
rs1042342067 Chr.21: 44241251-44241251 on Build GRCh38
rs985305062 Chr.21: 44241218-44241218 on Build GRCh38
rs922065495 Chr.21: 44241208-44241208 on Build GRCh38
rs932514200 Chr.21: 44240976-44240977 on Build GRCh38
rs111946962 Chr.21: 44240979-44240979 on Build GRCh38
rs2847224 Chr.21: 44240952-44240952 on Build GRCh38

View Full Product Details