Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159169204-159169790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC1
Entrez Gene ID
84624
Gene Name
fibronectin type III domain containing 1
Gene Aliases
AGS8, FNDC2, MEL4B3, bA243O10.1, dJ322A24.1
UniGene
Hs.520525
Gene Chromosome Location
Chr.6: 159169397-159272108 on Build GRCh38
Transcripts
NM_032532.2 XM_011536190.2 XM_011536191.2 NR_121668.1

Gene Symbol
LOC101929122
Entrez Gene ID
101929122
Gene Name
uncharacterized LOC101929122
Gene Aliases
-
UniGene
Hs.134215
Gene Chromosome Location
Chr.6: 159165899-159170007 on Build GRCh38
Transcripts
NM_032532.2 XM_011536190.2 XM_011536191.2 NR_121668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6284603 c.10G>T p.E4* Chr.6: 159169606-159169606 on Build GRCh38
5480010 c.106T>C p.S36P Chr.6: 159169702-159169702 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs538064187 Chr.6: 159169385-159169385 on Build GRCh38
rs1029962300 Chr.6: 159169228-159169228 on Build GRCh38
rs970813202 Chr.6: 159169292-159169292 on Build GRCh38
rs544425804 Chr.6: 159169769-159169769 on Build GRCh38
rs924021133 Chr.6: 159169307-159169307 on Build GRCh38
rs184501248 Chr.6: 159169261-159169261 on Build GRCh38
rs1039886646 Chr.6: 159169352-159169352 on Build GRCh38
rs901316656 Chr.6: 159169364-159169364 on Build GRCh38
rs545005095 Chr.6: 159169753-159169753 on Build GRCh38
rs558701793 Chr.6: 159169758-159169758 on Build GRCh38
rs997224263 Chr.6: 159169369-159169369 on Build GRCh38
rs867510931 Chr.6: 159169346-159169346 on Build GRCh38
rs568662089 Chr.6: 159169681-159169681 on Build GRCh38
rs975431005 Chr.6: 159169721-159169721 on Build GRCh38
rs940409585 Chr.6: 159169612-159169612 on Build GRCh38
rs295332 Chr.6: 159169702-159169702 on Build GRCh38
rs981935757 Chr.6: 159169480-159169480 on Build GRCh38
rs963342654 Chr.6: 159169413-159169413 on Build GRCh38
rs189260029 Chr.6: 159169605-159169605 on Build GRCh38
rs1051500567 Chr.6: 159169406-159169406 on Build GRCh38
rs1023592926 Chr.6: 159169651-159169651 on Build GRCh38
rs534463114 Chr.6: 159169697-159169697 on Build GRCh38
rs963612209 Chr.6: 159169285-159169293 on Build GRCh38
rs1047204980 Chr.6: 159169599-159169599 on Build GRCh38
rs976031875 Chr.6: 159169534-159169534 on Build GRCh38
rs1037381998 Chr.6: 159169620-159169620 on Build GRCh38
rs916373331 Chr.6: 159169287-159169287 on Build GRCh38
rs879058235 Chr.6: 159169361-159169361 on Build GRCh38
rs989515398 Chr.6: 159169508-159169508 on Build GRCh38
rs942974862 Chr.6: 159169523-159169523 on Build GRCh38
rs1022298612 Chr.6: 159169717-159169717 on Build GRCh38
rs970590593 Chr.6: 159169457-159169457 on Build GRCh38
rs552278238 Chr.6: 159169343-159169343 on Build GRCh38
rs964431964 Chr.6: 159169720-159169720 on Build GRCh38
rs922853757 Chr.6: 159169578-159169578 on Build GRCh38
rs1016215827 Chr.6: 159169263-159169263 on Build GRCh38
rs193207381 Chr.6: 159169747-159169747 on Build GRCh38
rs915198514 Chr.6: 159169512-159169512 on Build GRCh38
rs1012547394 Chr.6: 159169425-159169425 on Build GRCh38
rs1004581605 Chr.6: 159169410-159169410 on Build GRCh38
rs1015968632 Chr.6: 159169505-159169505 on Build GRCh38
rs906436665 Chr.6: 159169652-159169652 on Build GRCh38
rs898791029 Chr.6: 159169624-159169624 on Build GRCh38
rs982930641 Chr.6: 159169766-159169766 on Build GRCh38
rs1023479950 Chr.6: 159169427-159169427 on Build GRCh38
rs1003404090 Chr.6: 159169680-159169680 on Build GRCh38
rs1012166358 Chr.6: 159169628-159169628 on Build GRCh38
rs1012892943 Chr.6: 159169772-159169772 on Build GRCh38
rs956759621 Chr.6: 159169496-159169496 on Build GRCh38
rs934193740 Chr.6: 159169582-159169582 on Build GRCh38
rs295333 Chr.6: 159169344-159169344 on Build GRCh38
rs886180351 Chr.6: 159169407-159169407 on Build GRCh38
rs990689799 Chr.6: 159169286-159169286 on Build GRCh38
rs982156749 Chr.6: 159169300-159169300 on Build GRCh38
rs575385762 Chr.6: 159169767-159169767 on Build GRCh38
rs745445002 Chr.6: 159169633-159169633 on Build GRCh38
rs1031123339 Chr.6: 159169490-159169490 on Build GRCh38
rs935355458 Chr.6: 159169333-159169333 on Build GRCh38

View Full Product Details