Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42780399-42780904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WNK4
Entrez Gene ID
65266
Gene Name
WNK lysine deficient protein kinase 4
Gene Aliases
PHA2B, PRKWNK4
UniGene
Hs.105448
Gene Chromosome Location
Chr.17: 42779701-42797066 on Build GRCh38
Transcripts
NM_001321299.1 NM_032387.4 XM_011525135.2 XM_017024962.1 XM_017024963.1 XM_017024964.1 XM_017024965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3517758 c.68G>A p.R23Q Chr.17: 42780766-42780766 on Build GRCh38
6391543 c.8C>T p.A3V Chr.17: 42780706-42780706 on Build GRCh38
6823127 c.26C>T p.T9I Chr.17: 42780724-42780724 on Build GRCh38
4914818 c.32T>C p.V11A Chr.17: 42780730-42780730 on Build GRCh38
6375215 c.104G>A p.R35H Chr.17: 42780802-42780802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200318202 Chr.17: 42780677-42780677 on Build GRCh38
rs202057094 Chr.17: 42780692-42780692 on Build GRCh38
rs756995575 Chr.17: 42780774-42780774 on Build GRCh38
rs759170365 Chr.17: 42780819-42780819 on Build GRCh38
rs749459405 Chr.17: 42780777-42780777 on Build GRCh38
rs1012246606 Chr.17: 42780814-42780814 on Build GRCh38
rs936723484 Chr.17: 42780480-42780480 on Build GRCh38
rs752056395 Chr.17: 42780661-42780661 on Build GRCh38
rs748667566 Chr.17: 42780791-42780791 on Build GRCh38
rs375444500 Chr.17: 42780880-42780880 on Build GRCh38
rs750387045 Chr.17: 42780665-42780666 on Build GRCh38
rs746782601 Chr.17: 42780770-42780770 on Build GRCh38
rs755221021 Chr.17: 42780849-42780849 on Build GRCh38
rs778741303 Chr.17: 42780684-42780684 on Build GRCh38
rs915819165 Chr.17: 42780431-42780431 on Build GRCh38
rs751457281 Chr.17: 42780839-42780839 on Build GRCh38
rs370313183 Chr.17: 42780656-42780656 on Build GRCh38
rs766110219 Chr.17: 42780741-42780741 on Build GRCh38
rs900799207 Chr.17: 42780486-42780486 on Build GRCh38
rs959813875 Chr.17: 42780597-42780597 on Build GRCh38
rs754554663 Chr.17: 42780791-42780791 on Build GRCh38
rs752591503 Chr.17: 42780762-42780762 on Build GRCh38
rs537127664 Chr.17: 42780662-42780662 on Build GRCh38
rs371972656 Chr.17: 42780768-42780768 on Build GRCh38
rs1024175873 Chr.17: 42780815-42780815 on Build GRCh38
rs970358476 Chr.17: 42780649-42780649 on Build GRCh38
rs565987446 Chr.17: 42780530-42780530 on Build GRCh38
rs780170215 Chr.17: 42780776-42780776 on Build GRCh38
rs775594265 Chr.17: 42780709-42780709 on Build GRCh38
rs748218580 Chr.17: 42780853-42780853 on Build GRCh38
rs897402948 Chr.17: 42780588-42780588 on Build GRCh38
rs755811535 Chr.17: 42780676-42780676 on Build GRCh38
rs779978421 Chr.17: 42780768-42780768 on Build GRCh38
rs769877352 Chr.17: 42780651-42780651 on Build GRCh38
rs746869790 Chr.17: 42780768-42780769 on Build GRCh38
rs146248553 Chr.17: 42780435-42780435 on Build GRCh38
rs768049262 Chr.17: 42780728-42780728 on Build GRCh38
rs764485950 Chr.17: 42780658-42780658 on Build GRCh38
rs576090257 Chr.17: 42780644-42780644 on Build GRCh38
rs969905249 Chr.17: 42780801-42780801 on Build GRCh38
rs370527167 Chr.17: 42780790-42780790 on Build GRCh38
rs766814362 Chr.17: 42780808-42780808 on Build GRCh38
rs773763949 Chr.17: 42780797-42780797 on Build GRCh38
rs529976001 Chr.17: 42780846-42780846 on Build GRCh38
rs925872814 Chr.17: 42780454-42780454 on Build GRCh38
rs892579963 Chr.17: 42780516-42780516 on Build GRCh38
rs758373345 Chr.17: 42780834-42780834 on Build GRCh38
rs753485187 Chr.17: 42780743-42780743 on Build GRCh38
rs889978315 Chr.17: 42780583-42780583 on Build GRCh38
rs772673471 Chr.17: 42780796-42780796 on Build GRCh38
rs1005697978 Chr.17: 42780585-42780585 on Build GRCh38
rs547695680 Chr.17: 42780489-42780489 on Build GRCh38
rs61754323 Chr.17: 42780681-42780681 on Build GRCh38
rs773704739 Chr.17: 42780731-42780731 on Build GRCh38
rs915747607 Chr.17: 42780628-42780628 on Build GRCh38
rs764738132 Chr.17: 42780756-42780756 on Build GRCh38
rs761145984 Chr.17: 42780802-42780802 on Build GRCh38
rs777488350 Chr.17: 42780690-42780690 on Build GRCh38
rs757050478 Chr.17: 42780840-42780840 on Build GRCh38
rs768740797 Chr.17: 42780713-42780713 on Build GRCh38
rs938499880 Chr.17: 42780422-42780422 on Build GRCh38
rs1049460178 Chr.17: 42780491-42780491 on Build GRCh38
rs1017236346 Chr.17: 42780587-42780587 on Build GRCh38
rs375681809 Chr.17: 42780652-42780652 on Build GRCh38
rs764712635 Chr.17: 42780820-42780820 on Build GRCh38
rs997682411 Chr.17: 42780532-42780532 on Build GRCh38
rs762319507 Chr.17: 42780722-42780722 on Build GRCh38
rs374001790 Chr.17: 42780823-42780823 on Build GRCh38
rs1035020595 Chr.17: 42780601-42780601 on Build GRCh38
rs1007813318 Chr.17: 42780573-42780573 on Build GRCh38
rs538814418 Chr.17: 42780455-42780455 on Build GRCh38
rs955842877 Chr.17: 42780862-42780862 on Build GRCh38
rs201419013 Chr.17: 42780861-42780861 on Build GRCh38
rs763553713 Chr.17: 42780657-42780657 on Build GRCh38
rs992658043 Chr.17: 42780428-42780428 on Build GRCh38
rs542693543 Chr.17: 42780810-42780810 on Build GRCh38
rs764040354 Chr.17: 42780838-42780838 on Build GRCh38
rs991341609 Chr.17: 42780623-42780623 on Build GRCh38
rs61754322 Chr.17: 42780596-42780596 on Build GRCh38
rs1030538955 Chr.17: 42780535-42780535 on Build GRCh38
rs758319975 Chr.17: 42780764-42780764 on Build GRCh38
rs778456079 Chr.17: 42780870-42780870 on Build GRCh38
rs1045429908 Chr.17: 42780444-42780446 on Build GRCh38
rs889333871 Chr.17: 42780555-42780555 on Build GRCh38
rs61754324 Chr.17: 42780714-42780714 on Build GRCh38
rs960713079 Chr.17: 42780611-42780611 on Build GRCh38
rs61754321 Chr.17: 42780557-42780557 on Build GRCh38
rs748082300 Chr.17: 42780685-42780685 on Build GRCh38
rs771768583 Chr.17: 42780686-42780686 on Build GRCh38
rs61754325 Chr.17: 42780745-42780745 on Build GRCh38
rs933656010 Chr.17: 42780732-42780732 on Build GRCh38
rs374744005 Chr.17: 42780671-42780671 on Build GRCh38
rs933401331 Chr.17: 42780695-42780695 on Build GRCh38
rs1016314246 Chr.17: 42780578-42780578 on Build GRCh38
rs577125376 Chr.17: 42780712-42780712 on Build GRCh38
rs778896525 Chr.17: 42780852-42780852 on Build GRCh38
rs368683340 Chr.17: 42780765-42780765 on Build GRCh38
rs1023702090 Chr.17: 42780641-42780641 on Build GRCh38
rs1052728868 Chr.17: 42780485-42780485 on Build GRCh38
rs758474340 Chr.17: 42780667-42780668 on Build GRCh38
rs760446306 Chr.17: 42780735-42780735 on Build GRCh38
rs774472658 Chr.17: 42780780-42780780 on Build GRCh38

View Full Product Details