Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31665872-31666374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK2B
Entrez Gene ID
1460
Gene Name
casein kinase 2 beta
Gene Aliases
CK2B, CK2N, CSK2B, G5A
UniGene
Hs.73527
Gene Chromosome Location
Chr.6: 31665880-31670070 on Build GRCh38
Transcripts
NM_001282385.1 NM_001320.6 NM_001199237.1 XM_005249403.3

Gene Symbol
GPANK1
Entrez Gene ID
7918
Gene Name
G-patch domain and ankyrin repeats 1
Gene Aliases
ANKRD59, BAT4, D6S54E, G5, GPATCH10
UniGene
Hs.247478
Gene Chromosome Location
Chr.6: 31661229-31666283 on Build GRCh38
Transcripts
NM_001282385.1 NM_001320.6 NM_001199237.1 XM_005249403.3

SNP Detail

SNP ID Chromosome Location
rs567067253 Chr.6: 31666176-31666176 on Build GRCh38
rs887190990 Chr.6: 31666199-31666199 on Build GRCh38
rs551629267 Chr.6: 31666173-31666173 on Build GRCh38
rs1051381410 Chr.6: 31666120-31666120 on Build GRCh38
rs535268272 Chr.6: 31666257-31666257 on Build GRCh38
rs867380870 Chr.6: 31665900-31665900 on Build GRCh38
rs987886261 Chr.6: 31666112-31666112 on Build GRCh38
rs1049205319 Chr.6: 31666160-31666160 on Build GRCh38
rs753492791 Chr.6: 31665899-31665899 on Build GRCh38
rs147357294 Chr.6: 31666056-31666057 on Build GRCh38
rs898004634 Chr.6: 31666126-31666126 on Build GRCh38
rs1052517348 Chr.6: 31665901-31665901 on Build GRCh38
rs956503813 Chr.6: 31666060-31666060 on Build GRCh38
rs146452657 Chr.6: 31666030-31666030 on Build GRCh38
rs987084218 Chr.6: 31666338-31666338 on Build GRCh38
rs1033027469 Chr.6: 31666307-31666307 on Build GRCh38
rs1030017705 Chr.6: 31666089-31666089 on Build GRCh38
rs1030942354 Chr.6: 31666217-31666217 on Build GRCh38
rs1051602624 Chr.6: 31665934-31665934 on Build GRCh38
rs28986180 Chr.6: 31666058-31666059 on Build GRCh38
rs758350057 Chr.6: 31666047-31666047 on Build GRCh38
rs943318306 Chr.6: 31666141-31666141 on Build GRCh38
rs141052842 Chr.6: 31666181-31666181 on Build GRCh38
rs925505242 Chr.6: 31666106-31666106 on Build GRCh38
rs535535721 Chr.6: 31666289-31666289 on Build GRCh38
rs1026667002 Chr.6: 31665963-31665963 on Build GRCh38
rs539448235 Chr.6: 31666151-31666151 on Build GRCh38
rs918131838 Chr.6: 31666045-31666045 on Build GRCh38
rs568249646 Chr.6: 31666184-31666184 on Build GRCh38
rs982017903 Chr.6: 31666023-31666023 on Build GRCh38
rs956598113 Chr.6: 31666226-31666226 on Build GRCh38
rs951330312 Chr.6: 31666033-31666033 on Build GRCh38
rs1007344306 Chr.6: 31665940-31665940 on Build GRCh38
rs889040698 Chr.6: 31665935-31665935 on Build GRCh38
rs936400405 Chr.6: 31666108-31666108 on Build GRCh38
rs889024432 Chr.6: 31666139-31666139 on Build GRCh38
rs752797162 Chr.6: 31665953-31665953 on Build GRCh38
rs989296940 Chr.6: 31666074-31666074 on Build GRCh38
rs1036938801 Chr.6: 31665906-31665906 on Build GRCh38
rs550699129 Chr.6: 31666099-31666099 on Build GRCh38
rs570478106 Chr.6: 31666014-31666014 on Build GRCh38
rs915359252 Chr.6: 31666076-31666076 on Build GRCh38
rs995856712 Chr.6: 31666071-31666071 on Build GRCh38
rs568868694 Chr.6: 31666118-31666118 on Build GRCh38
rs574222918 Chr.6: 31666240-31666240 on Build GRCh38
rs954580020 Chr.6: 31666096-31666096 on Build GRCh38
rs888176418 Chr.6: 31665973-31665974 on Build GRCh38
rs754126879 Chr.6: 31666320-31666320 on Build GRCh38
rs986172611 Chr.6: 31666087-31666087 on Build GRCh38
rs997157981 Chr.6: 31665922-31665922 on Build GRCh38
rs140631569 Chr.6: 31666345-31666346 on Build GRCh38
rs910240352 Chr.6: 31666344-31666344 on Build GRCh38
rs561954738 Chr.6: 31665992-31665992 on Build GRCh38
rs936466416 Chr.6: 31665895-31665895 on Build GRCh38
rs906170252 Chr.6: 31666191-31666191 on Build GRCh38
rs557101961 Chr.6: 31666309-31666309 on Build GRCh38
rs1016132253 Chr.6: 31665975-31665975 on Build GRCh38
rs546666376 Chr.6: 31666124-31666124 on Build GRCh38
rs1023291569 Chr.6: 31665985-31665985 on Build GRCh38
rs913654982 Chr.6: 31666114-31666114 on Build GRCh38
rs995626641 Chr.6: 31665996-31665996 on Build GRCh38
rs989349190 Chr.6: 31666235-31666235 on Build GRCh38
rs932048178 Chr.6: 31666081-31666081 on Build GRCh38
rs988723342 Chr.6: 31666310-31666310 on Build GRCh38
rs933104971 Chr.6: 31666334-31666334 on Build GRCh38
rs1022137190 Chr.6: 31666255-31666255 on Build GRCh38
rs969617183 Chr.6: 31666281-31666281 on Build GRCh38
rs980930391 Chr.6: 31666090-31666090 on Build GRCh38
rs992165121 Chr.6: 31666039-31666039 on Build GRCh38
rs913615249 Chr.6: 31666318-31666318 on Build GRCh38
rs1003888690 Chr.6: 31666216-31666216 on Build GRCh38
rs16899941 Chr.6: 31665912-31665912 on Build GRCh38
rs969670642 Chr.6: 31666077-31666077 on Build GRCh38
rs528261758 Chr.6: 31665911-31665911 on Build GRCh38

View Full Product Details