Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150338902-150339396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RARRES2
Entrez Gene ID
5919
Gene Name
retinoic acid receptor responder 2
Gene Aliases
HP10433, TIG2
UniGene
Hs.647064
Gene Chromosome Location
Chr.7: 150338318-150341685 on Build GRCh38
Transcripts
NM_002889.3 XM_017012491.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7274211 c.351C>T p.C117C Chr.7: 150339010-150339010 on Build GRCh38
5649568 c.293_294insT p.L99fs*9 Chr.7: 150339067-150339068 on Build GRCh38
1728607 c.335G>A p.G112D Chr.7: 150339026-150339026 on Build GRCh38
392167 c.355_356insA p.I119fs*10 Chr.7: 150339005-150339006 on Build GRCh38
5991437 c.350G>A p.C117Y Chr.7: 150339011-150339011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977709149 Chr.7: 150338964-150338964 on Build GRCh38
rs774218646 Chr.7: 150339037-150339037 on Build GRCh38
rs758712774 Chr.7: 150338970-150338970 on Build GRCh38
rs990875244 Chr.7: 150339354-150339354 on Build GRCh38
rs201989291 Chr.7: 150339074-150339074 on Build GRCh38
rs117067760 Chr.7: 150339334-150339334 on Build GRCh38
rs759487691 Chr.7: 150339122-150339122 on Build GRCh38
rs769214253 Chr.7: 150339086-150339086 on Build GRCh38
rs770192458 Chr.7: 150338989-150338989 on Build GRCh38
rs757531422 Chr.7: 150339075-150339075 on Build GRCh38
rs138193154 Chr.7: 150338987-150338987 on Build GRCh38
rs375183992 Chr.7: 150339015-150339015 on Build GRCh38
rs997597640 Chr.7: 150338961-150338961 on Build GRCh38
rs1000717158 Chr.7: 150339290-150339290 on Build GRCh38
rs771999265 Chr.7: 150339272-150339272 on Build GRCh38
rs150618712 Chr.7: 150339077-150339077 on Build GRCh38
rs766137842 Chr.7: 150339115-150339115 on Build GRCh38
rs752705936 Chr.7: 150338963-150338963 on Build GRCh38
rs760662057 Chr.7: 150339036-150339036 on Build GRCh38
rs555156802 Chr.7: 150339085-150339085 on Build GRCh38
rs901404359 Chr.7: 150339260-150339260 on Build GRCh38
rs773317212 Chr.7: 150339302-150339302 on Build GRCh38
rs923026091 Chr.7: 150338931-150338931 on Build GRCh38
rs774373551 Chr.7: 150339161-150339161 on Build GRCh38
rs772969637 Chr.7: 150339024-150339024 on Build GRCh38
rs749752401 Chr.7: 150339090-150339090 on Build GRCh38
rs1051850456 Chr.7: 150339222-150339222 on Build GRCh38
rs369898714 Chr.7: 150339094-150339094 on Build GRCh38
rs762377599 Chr.7: 150339010-150339010 on Build GRCh38
rs959061154 Chr.7: 150339317-150339317 on Build GRCh38
rs200065559 Chr.7: 150339018-150339018 on Build GRCh38
rs761726877 Chr.7: 150339086-150339086 on Build GRCh38
rs998049720 Chr.7: 150339219-150339219 on Build GRCh38
rs761608004 Chr.7: 150338935-150338935 on Build GRCh38
rs747137713 Chr.7: 150339073-150339073 on Build GRCh38
rs150257545 Chr.7: 150339128-150339128 on Build GRCh38
rs767247304 Chr.7: 150338938-150338938 on Build GRCh38
rs567505283 Chr.7: 150339066-150339066 on Build GRCh38
rs747182020 Chr.7: 150338980-150338980 on Build GRCh38
rs750388554 Chr.7: 150338940-150338940 on Build GRCh38
rs773057585 Chr.7: 150339103-150339103 on Build GRCh38
rs201032822 Chr.7: 150339093-150339093 on Build GRCh38
rs950494861 Chr.7: 150339358-150339358 on Build GRCh38
rs80070869 Chr.7: 150339199-150339199 on Build GRCh38
rs541742873 Chr.7: 150339267-150339267 on Build GRCh38
rs531458324 Chr.7: 150339258-150339258 on Build GRCh38
rs200967332 Chr.7: 150339049-150339049 on Build GRCh38
rs115799842 Chr.7: 150339245-150339245 on Build GRCh38
rs900483413 Chr.7: 150339203-150339203 on Build GRCh38
rs537665173 Chr.7: 150339097-150339097 on Build GRCh38
rs900602371 Chr.7: 150338932-150338932 on Build GRCh38
rs942230881 Chr.7: 150339198-150339198 on Build GRCh38
rs753740724 Chr.7: 150339046-150339046 on Build GRCh38
rs527467984 Chr.7: 150339373-150339373 on Build GRCh38
rs1047886885 Chr.7: 150339164-150339164 on Build GRCh38
rs1023342051 Chr.7: 150339357-150339357 on Build GRCh38
rs1804764 Chr.7: 150338988-150338988 on Build GRCh38
rs761411546 Chr.7: 150339021-150339021 on Build GRCh38
rs369499714 Chr.7: 150339023-150339023 on Build GRCh38
rs924912088 Chr.7: 150339118-150339118 on Build GRCh38
rs916393928 Chr.7: 150339005-150339005 on Build GRCh38
rs957814201 Chr.7: 150339337-150339337 on Build GRCh38
rs3210475 Chr.7: 150339030-150339030 on Build GRCh38
rs766220999 Chr.7: 150339039-150339039 on Build GRCh38
rs992199267 Chr.7: 150339329-150339329 on Build GRCh38
rs767283380 Chr.7: 150339102-150339103 on Build GRCh38
rs11558529 Chr.7: 150339027-150339027 on Build GRCh38
rs977818928 Chr.7: 150339110-150339110 on Build GRCh38
rs755447100 Chr.7: 150338930-150338930 on Build GRCh38
rs764195333 Chr.7: 150339059-150339059 on Build GRCh38
rs553561665 Chr.7: 150339159-150339159 on Build GRCh38
rs1020838583 Chr.7: 150339327-150339327 on Build GRCh38
rs768609534 Chr.7: 150338975-150338975 on Build GRCh38
rs756010640 Chr.7: 150338954-150338954 on Build GRCh38
rs949356604 Chr.7: 150339131-150339131 on Build GRCh38
rs748545317 Chr.7: 150339223-150339223 on Build GRCh38
rs760389743 Chr.7: 150339105-150339105 on Build GRCh38
rs200876936 Chr.7: 150338979-150338979 on Build GRCh38
rs138940225 Chr.7: 150339176-150339176 on Build GRCh38
rs779099036 Chr.7: 150338958-150338958 on Build GRCh38
rs1009407661 Chr.7: 150339254-150339254 on Build GRCh38
rs374620652 Chr.7: 150339121-150339121 on Build GRCh38
rs540698063 Chr.7: 150339120-150339121 on Build GRCh38
rs1129497 Chr.7: 150338991-150338991 on Build GRCh38

View Full Product Details