Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 72130020-72130529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHSL2
Entrez Gene ID
340527
Gene Name
NHS like 2
Gene Aliases
-
UniGene
Hs.397836
Gene Chromosome Location
Chr.X: 71911073-72153286 on Build GRCh38
Transcripts
NM_001013627.2 XM_011530932.1 XM_011530933.1 XM_011530934.2 XM_017029473.1 XM_017029474.1 XM_017029475.1 XM_017029476.1 XM_017029477.1 NM_001024455.3

Gene Symbol
RGAG4
Entrez Gene ID
340526
Gene Name
retrotransposon gag domain containing 4
Gene Aliases
6430402L03Rik, MAR5, MART5
UniGene
Hs.512180
Gene Chromosome Location
Chr.X: 72127110-72131901 on Build GRCh38
Transcripts
NM_001013627.2 XM_011530932.1 XM_011530933.1 XM_011530934.2 XM_017029473.1 XM_017029474.1 XM_017029475.1 XM_017029476.1 XM_017029477.1 NM_001024455.3

SNP Detail

SNP ID Chromosome Location
rs764230784 Chr.X: 72130172-72130172 on Build GRCh38
rs6624595 Chr.X: 72130283-72130283 on Build GRCh38
rs781715793 Chr.X: 72130078-72130078 on Build GRCh38
rs749168615 Chr.X: 72130371-72130373 on Build GRCh38
rs757021429 Chr.X: 72130125-72130125 on Build GRCh38
rs780558080 Chr.X: 72130187-72130187 on Build GRCh38
rs757911792 Chr.X: 72130348-72130353 on Build GRCh38
rs372733180 Chr.X: 72130113-72130113 on Build GRCh38
rs777354507 Chr.X: 72130205-72130205 on Build GRCh38
rs763523522 Chr.X: 72130366-72130383 on Build GRCh38
rs773585535 Chr.X: 72130326-72130326 on Build GRCh38
rs367678202 Chr.X: 72130198-72130198 on Build GRCh38
rs763116118 Chr.X: 72130071-72130071 on Build GRCh38
rs777103915 Chr.X: 72130314-72130316 on Build GRCh38
rs760027218 Chr.X: 72130326-72130329 on Build GRCh38
rs760970544 Chr.X: 72130328-72130328 on Build GRCh38
rs1028453688 Chr.X: 72130448-72130448 on Build GRCh38
rs759755785 Chr.X: 72130055-72130055 on Build GRCh38
rs749231105 Chr.X: 72130460-72130460 on Build GRCh38
rs755935647 Chr.X: 72130447-72130447 on Build GRCh38
rs60726595 Chr.X: 72130143-72130143 on Build GRCh38
rs774516982 Chr.X: 72130480-72130480 on Build GRCh38
rs752160610 Chr.X: 72130189-72130189 on Build GRCh38
rs762226176 Chr.X: 72130175-72130175 on Build GRCh38
rs745654595 Chr.X: 72130394-72130395 on Build GRCh38
rs773197783 Chr.X: 72130494-72130495 on Build GRCh38
rs775454517 Chr.X: 72130151-72130151 on Build GRCh38
rs776641896 Chr.X: 72130049-72130049 on Build GRCh38
rs766799770 Chr.X: 72130355-72130355 on Build GRCh38
rs770843646 Chr.X: 72130126-72130126 on Build GRCh38
rs995909319 Chr.X: 72130127-72130127 on Build GRCh38
rs745766699 Chr.X: 72130169-72130169 on Build GRCh38
rs751171435 Chr.X: 72130367-72130370 on Build GRCh38
rs780731250 Chr.X: 72130348-72130348 on Build GRCh38
rs769867866 Chr.X: 72130244-72130244 on Build GRCh38
rs758470258 Chr.X: 72130250-72130250 on Build GRCh38
rs749400236 Chr.X: 72130274-72130274 on Build GRCh38
rs755133990 Chr.X: 72130059-72130059 on Build GRCh38
rs866914625 Chr.X: 72130260-72130260 on Build GRCh38
rs763629425 Chr.X: 72130404-72130404 on Build GRCh38
rs751074420 Chr.X: 72130415-72130415 on Build GRCh38
rs755973703 Chr.X: 72130395-72130418 on Build GRCh38
rs768319540 Chr.X: 72130331-72130332 on Build GRCh38
rs369299268 Chr.X: 72130366-72130366 on Build GRCh38
rs267606505 Chr.X: 72130298-72130298 on Build GRCh38
rs760830014 Chr.X: 72130258-72130263 on Build GRCh38
rs778358543 Chr.X: 72130111-72130111 on Build GRCh38
rs759517559 Chr.X: 72130135-72130135 on Build GRCh38
rs369686441 Chr.X: 72130484-72130484 on Build GRCh38
rs750913102 Chr.X: 72130092-72130092 on Build GRCh38
rs770892372 Chr.X: 72130226-72130226 on Build GRCh38
rs370818026 Chr.X: 72130070-72130070 on Build GRCh38
rs183714924 Chr.X: 72130403-72130403 on Build GRCh38
rs756994530 Chr.X: 72130422-72130422 on Build GRCh38
rs146618350 Chr.X: 72130444-72130444 on Build GRCh38
rs756640557 Chr.X: 72130186-72130186 on Build GRCh38
rs753322291 Chr.X: 72130348-72130350 on Build GRCh38
rs987051099 Chr.X: 72130096-72130096 on Build GRCh38
rs182079718 Chr.X: 72130288-72130288 on Build GRCh38
rs754364559 Chr.X: 72130364-72130364 on Build GRCh38
rs780935401 Chr.X: 72130423-72130423 on Build GRCh38
rs753625204 Chr.X: 72130112-72130112 on Build GRCh38
rs767842591 Chr.X: 72130183-72130183 on Build GRCh38
rs200189694 Chr.X: 72130073-72130073 on Build GRCh38
rs201171574 Chr.X: 72130110-72130110 on Build GRCh38
rs776759820 Chr.X: 72130128-72130128 on Build GRCh38
rs750966235 Chr.X: 72130185-72130185 on Build GRCh38
rs768655277 Chr.X: 72130473-72130473 on Build GRCh38
rs768771267 Chr.X: 72130277-72130277 on Build GRCh38
rs779901827 Chr.X: 72130401-72130403 on Build GRCh38
rs780950658 Chr.X: 72130393-72130394 on Build GRCh38
rs983578517 Chr.X: 72130445-72130445 on Build GRCh38
rs781166664 Chr.X: 72130231-72130231 on Build GRCh38
rs772143269 Chr.X: 72130419-72130421 on Build GRCh38
rs746644853 Chr.X: 72130217-72130217 on Build GRCh38
rs766726937 Chr.X: 72130291-72130320 on Build GRCh38
rs936438936 Chr.X: 72130197-72130197 on Build GRCh38
rs767637198 Chr.X: 72130074-72130074 on Build GRCh38
rs778540620 Chr.X: 72130416-72130418 on Build GRCh38
rs35968794 Chr.X: 72130267-72130268 on Build GRCh38
rs745609510 Chr.X: 72130238-72130238 on Build GRCh38
rs868311232 Chr.X: 72130290-72130290 on Build GRCh38

View Full Product Details