Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75299579-75300126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPRT
Entrez Gene ID
139596
Gene Name
uracil phosphoribosyltransferase homolog
Gene Aliases
FUR1, UPP
UniGene
Hs.91612
Gene Chromosome Location
Chr.X: 75274059-75304897 on Build GRCh38
Transcripts
NM_001307944.1 NM_145052.3 NR_030774.1

SNP Detail

SNP ID Chromosome Location
rs192578725 Chr.X: 75299692-75299692 on Build GRCh38
rs1054230096 Chr.X: 75299914-75299914 on Build GRCh38
rs772401368 Chr.X: 75299927-75299927 on Build GRCh38
rs937246756 Chr.X: 75299909-75299909 on Build GRCh38
rs781319191 Chr.X: 75299923-75299923 on Build GRCh38
rs755231771 Chr.X: 75299918-75299918 on Build GRCh38
rs767516008 Chr.X: 75299808-75299808 on Build GRCh38
rs1041769902 Chr.X: 75300015-75300015 on Build GRCh38
rs918256628 Chr.X: 75299701-75299701 on Build GRCh38
rs544003446 Chr.X: 75299660-75299660 on Build GRCh38
rs749555172 Chr.X: 75299706-75299716 on Build GRCh38
rs772661687 Chr.X: 75299707-75299707 on Build GRCh38
rs375878184 Chr.X: 75300038-75300038 on Build GRCh38
rs778851738 Chr.X: 75299636-75299636 on Build GRCh38
rs930799881 Chr.X: 75299728-75299728 on Build GRCh38
rs957269426 Chr.X: 75299765-75299765 on Build GRCh38
rs988807337 Chr.X: 75299617-75299617 on Build GRCh38
rs764914390 Chr.X: 75299714-75299714 on Build GRCh38
rs1036724638 Chr.X: 75299952-75299952 on Build GRCh38
rs763677284 Chr.X: 75299759-75299759 on Build GRCh38
rs774884791 Chr.X: 75300067-75300067 on Build GRCh38
rs755220926 Chr.X: 75300100-75300100 on Build GRCh38
rs780088630 Chr.X: 75299688-75299691 on Build GRCh38
rs997563492 Chr.X: 75300054-75300054 on Build GRCh38
rs200426086 Chr.X: 75299879-75299879 on Build GRCh38
rs1044664869 Chr.X: 75299674-75299674 on Build GRCh38
rs779893995 Chr.X: 75299868-75299868 on Build GRCh38
rs757103325 Chr.X: 75299793-75299793 on Build GRCh38
rs771349467 Chr.X: 75300031-75300031 on Build GRCh38
rs950377554 Chr.X: 75299647-75299647 on Build GRCh38
rs778273866 Chr.X: 75300008-75300008 on Build GRCh38
rs1003876010 Chr.X: 75299864-75299864 on Build GRCh38
rs745390040 Chr.X: 75300076-75300076 on Build GRCh38
rs5938015 Chr.X: 75300004-75300004 on Build GRCh38
rs748349582 Chr.X: 75299953-75299953 on Build GRCh38
rs946136541 Chr.X: 75300008-75300009 on Build GRCh38
rs185025369 Chr.X: 75300025-75300025 on Build GRCh38
rs1006457095 Chr.X: 75300081-75300081 on Build GRCh38
rs761114236 Chr.X: 75299713-75299713 on Build GRCh38
rs755944011 Chr.X: 75299863-75299863 on Build GRCh38
rs768800532 Chr.X: 75299718-75299719 on Build GRCh38
rs371136388 Chr.X: 75299928-75299928 on Build GRCh38
rs368655148 Chr.X: 75299779-75299779 on Build GRCh38
rs145348422 Chr.X: 75299845-75299845 on Build GRCh38
rs1048138010 Chr.X: 75299826-75299826 on Build GRCh38
rs998483290 Chr.X: 75300011-75300011 on Build GRCh38
rs751120673 Chr.X: 75299785-75299785 on Build GRCh38
rs775276745 Chr.X: 75299718-75299718 on Build GRCh38
rs757173078 Chr.X: 75299623-75299623 on Build GRCh38
rs34875647 Chr.X: 75300043-75300044 on Build GRCh38
rs778920403 Chr.X: 75299921-75299921 on Build GRCh38
rs747535190 Chr.X: 75299934-75299934 on Build GRCh38
rs147078352 Chr.X: 75299613-75299613 on Build GRCh38
rs927256284 Chr.X: 75299799-75299799 on Build GRCh38
rs762753901 Chr.X: 75299725-75299725 on Build GRCh38
rs774843210 Chr.X: 75299906-75299907 on Build GRCh38
rs901915031 Chr.X: 75300017-75300017 on Build GRCh38
rs374264662 Chr.X: 75299693-75299693 on Build GRCh38

View Full Product Details