Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75304116-75304621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPRT
Entrez Gene ID
139596
Gene Name
uracil phosphoribosyltransferase homolog
Gene Aliases
FUR1, UPP
UniGene
Hs.91612
Gene Chromosome Location
Chr.X: 75274059-75304897 on Build GRCh38
Transcripts
NM_001307944.1 NM_145052.3 NR_030774.1

SNP Detail

SNP ID Chromosome Location
rs776782677 Chr.X: 75304214-75304214 on Build GRCh38
rs761732054 Chr.X: 75304226-75304226 on Build GRCh38
rs987203118 Chr.X: 75304387-75304387 on Build GRCh38
rs972346259 Chr.X: 75304430-75304430 on Build GRCh38
rs987584157 Chr.X: 75304465-75304465 on Build GRCh38
rs866011546 Chr.X: 75304306-75304306 on Build GRCh38
rs1020026070 Chr.X: 75304154-75304154 on Build GRCh38
rs963199861 Chr.X: 75304422-75304422 on Build GRCh38
rs768911259 Chr.X: 75304169-75304169 on Build GRCh38
rs753025590 Chr.X: 75304295-75304295 on Build GRCh38
rs759953182 Chr.X: 75304183-75304183 on Build GRCh38
rs763350522 Chr.X: 75304494-75304494 on Build GRCh38
rs978679118 Chr.X: 75304221-75304221 on Build GRCh38
rs911636115 Chr.X: 75304443-75304443 on Build GRCh38
rs968260524 Chr.X: 75304168-75304168 on Build GRCh38
rs1035793842 Chr.X: 75304285-75304285 on Build GRCh38
rs763050811 Chr.X: 75304330-75304330 on Build GRCh38
rs6647687 Chr.X: 75304445-75304445 on Build GRCh38
rs943054527 Chr.X: 75304527-75304527 on Build GRCh38
rs1009701238 Chr.X: 75304150-75304150 on Build GRCh38
rs910527066 Chr.X: 75304582-75304582 on Build GRCh38
rs919401122 Chr.X: 75304151-75304151 on Build GRCh38
rs933428040 Chr.X: 75304463-75304463 on Build GRCh38
rs199876551 Chr.X: 75304437-75304438 on Build GRCh38
rs991381921 Chr.X: 75304326-75304326 on Build GRCh38
rs764279485 Chr.X: 75304377-75304377 on Build GRCh38

View Full Product Details