Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 78747624-78748133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LPAR4
Entrez Gene ID
2846
Gene Name
lysophosphatidic acid receptor 4
Gene Aliases
GPR23, LPA4, P2RY9, P2Y5-LIKE, P2Y9
UniGene
Hs.522701
Gene Chromosome Location
Chr.X: 78747658-78758714 on Build GRCh38
Transcripts
NM_001278000.1 NM_005296.2 XM_005262126.3 XM_006724639.3 XM_017029437.1 XM_017029438.1

SNP Detail

SNP ID Chromosome Location
rs910753149 Chr.X: 78748084-78748084 on Build GRCh38
rs190688078 Chr.X: 78747673-78747673 on Build GRCh38
rs1050191534 Chr.X: 78748058-78748058 on Build GRCh38
rs927144463 Chr.X: 78748076-78748076 on Build GRCh38
rs958610843 Chr.X: 78747769-78747770 on Build GRCh38
rs924734141 Chr.X: 78747969-78747969 on Build GRCh38
rs1035569147 Chr.X: 78747708-78747708 on Build GRCh38
rs1037980591 Chr.X: 78748093-78748093 on Build GRCh38
rs940068499 Chr.X: 78747945-78747945 on Build GRCh38
rs750335715 Chr.X: 78747750-78747750 on Build GRCh38
rs868043775 Chr.X: 78747931-78747931 on Build GRCh38
rs933458166 Chr.X: 78748025-78748025 on Build GRCh38
rs866143994 Chr.X: 78747919-78747919 on Build GRCh38
rs995643616 Chr.X: 78747694-78747694 on Build GRCh38
rs779194431 Chr.X: 78747947-78747947 on Build GRCh38
rs1028543166 Chr.X: 78747669-78747669 on Build GRCh38
rs898585333 Chr.X: 78747954-78747954 on Build GRCh38
rs943611198 Chr.X: 78748085-78748085 on Build GRCh38
rs569900559 Chr.X: 78747926-78747926 on Build GRCh38
rs898188304 Chr.X: 78747686-78747686 on Build GRCh38
rs987596087 Chr.X: 78748077-78748077 on Build GRCh38
rs956455811 Chr.X: 78747695-78747695 on Build GRCh38
rs12395284 Chr.X: 78747874-78747874 on Build GRCh38
rs1039769477 Chr.X: 78747665-78747665 on Build GRCh38
rs986889333 Chr.X: 78747676-78747676 on Build GRCh38
rs939259350 Chr.X: 78747907-78747907 on Build GRCh38
rs949873252 Chr.X: 78747916-78747916 on Build GRCh38
rs12011392 Chr.X: 78748078-78748078 on Build GRCh38
rs980864389 Chr.X: 78747850-78747850 on Build GRCh38
rs968772787 Chr.X: 78747795-78747795 on Build GRCh38
rs138318226 Chr.X: 78747791-78747791 on Build GRCh38
rs185664324 Chr.X: 78747810-78747810 on Build GRCh38
rs926521699 Chr.X: 78747892-78747892 on Build GRCh38
rs886979211 Chr.X: 78747936-78747936 on Build GRCh38
rs991440999 Chr.X: 78747783-78747783 on Build GRCh38
rs754790016 Chr.X: 78747831-78747831 on Build GRCh38
rs1031835628 Chr.X: 78747678-78747678 on Build GRCh38
rs180988284 Chr.X: 78747765-78747765 on Build GRCh38
rs891342507 Chr.X: 78748074-78748074 on Build GRCh38
rs1008474601 Chr.X: 78748082-78748082 on Build GRCh38
rs752904330 Chr.X: 78748099-78748099 on Build GRCh38
rs979947452 Chr.X: 78747927-78747927 on Build GRCh38
rs996833879 Chr.X: 78748002-78748002 on Build GRCh38
rs951258937 Chr.X: 78747697-78747697 on Build GRCh38
rs766972843 Chr.X: 78747718-78747718 on Build GRCh38

View Full Product Details