Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105413420-105413819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TEX22
Entrez Gene ID
647310
Gene Name
testis expressed 22
Gene Aliases
-
UniGene
Hs.729499
Gene Chromosome Location
Chr.14: 105398499-105413859 on Build GRCh38
Transcripts
NM_001195082.1 XM_006720234.3

SNP Detail

SNP ID Chromosome Location
rs75068446 Chr.14: 105413615-105413615 on Build GRCh38
rs906195136 Chr.14: 105413510-105413510 on Build GRCh38
rs587749406 Chr.14: 105413526-105413526 on Build GRCh38
rs781894098 Chr.14: 105413790-105413790 on Build GRCh38
rs754559170 Chr.14: 105413649-105413649 on Build GRCh38
rs944703397 Chr.14: 105413764-105413764 on Build GRCh38
rs953339946 Chr.14: 105413684-105413684 on Build GRCh38
rs1023398348 Chr.14: 105413502-105413502 on Build GRCh38
rs989819077 Chr.14: 105413632-105413632 on Build GRCh38
rs956673026 Chr.14: 105413598-105413598 on Build GRCh38
rs1049253424 Chr.14: 105413455-105413455 on Build GRCh38
rs1011970120 Chr.14: 105413459-105413459 on Build GRCh38
rs587700802 Chr.14: 105413662-105413662 on Build GRCh38
rs782442191 Chr.14: 105413511-105413511 on Build GRCh38
rs912018445 Chr.14: 105413749-105413749 on Build GRCh38
rs986138557 Chr.14: 105413742-105413742 on Build GRCh38
rs142310777 Chr.14: 105413567-105413567 on Build GRCh38

View Full Product Details