Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86565455-86566018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA4
Entrez Gene ID
22802
Gene Name
chloride channel accessory 4
Gene Aliases
CaCC, CaCC2
UniGene
Hs.567422
Gene Chromosome Location
Chr.1: 86547076-86580758 on Build GRCh38
Transcripts
NM_012128.3 NR_024602.1 XM_011541015.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3493072 c.83G>A p.G28E Chr.1: 86566010-86566010 on Build GRCh38
6127960 c.23C>T p.P8L Chr.1: 86565950-86565950 on Build GRCh38
6064616 c.784C>A p.L262I Chr.1: 86565850-86565850 on Build GRCh38
6305840 c.6G>T p.V2V Chr.1: 86565933-86565933 on Build GRCh38
5702991 c.739G>A p.V247I Chr.1: 86565805-86565805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2231590 Chr.1: 86565639-86565639 on Build GRCh38
rs776946663 Chr.1: 86565989-86565989 on Build GRCh38
rs2231592 Chr.1: 86565973-86565973 on Build GRCh38
rs552835839 Chr.1: 86565640-86565640 on Build GRCh38
rs1005024648 Chr.1: 86565524-86565524 on Build GRCh38
rs759191410 Chr.1: 86565645-86565645 on Build GRCh38
rs754154848 Chr.1: 86565866-86565866 on Build GRCh38
rs886713867 Chr.1: 86565720-86565720 on Build GRCh38
rs377545104 Chr.1: 86565904-86565904 on Build GRCh38
rs916090085 Chr.1: 86565636-86565636 on Build GRCh38
rs554716598 Chr.1: 86565707-86565707 on Build GRCh38
rs374848221 Chr.1: 86565487-86565487 on Build GRCh38
rs921892624 Chr.1: 86565715-86565715 on Build GRCh38
rs371206003 Chr.1: 86565665-86565665 on Build GRCh38
rs774204047 Chr.1: 86565964-86565964 on Build GRCh38
rs766688618 Chr.1: 86565864-86565864 on Build GRCh38
rs879136520 Chr.1: 86565995-86565995 on Build GRCh38
rs766008951 Chr.1: 86565489-86565489 on Build GRCh38
rs775266586 Chr.1: 86565928-86565928 on Build GRCh38
rs186153841 Chr.1: 86565886-86565886 on Build GRCh38
rs922057500 Chr.1: 86565897-86565897 on Build GRCh38
rs748724090 Chr.1: 86565769-86565769 on Build GRCh38
rs1018750206 Chr.1: 86565828-86565828 on Build GRCh38
rs1049325806 Chr.1: 86565504-86565504 on Build GRCh38
rs965913722 Chr.1: 86565861-86565861 on Build GRCh38
rs756797338 Chr.1: 86565923-86565923 on Build GRCh38
rs1010801696 Chr.1: 86565699-86565699 on Build GRCh38
rs757799842 Chr.1: 86565778-86565778 on Build GRCh38
rs776312652 Chr.1: 86565798-86565798 on Build GRCh38
rs775355339 Chr.1: 86565807-86565807 on Build GRCh38
rs746501996 Chr.1: 86565920-86565920 on Build GRCh38
rs781062541 Chr.1: 86565820-86565821 on Build GRCh38
rs538360706 Chr.1: 86565702-86565702 on Build GRCh38
rs753092233 Chr.1: 86565889-86565889 on Build GRCh38
rs2231589 Chr.1: 86565625-86565625 on Build GRCh38
rs754244572 Chr.1: 86565764-86565764 on Build GRCh38
rs561746865 Chr.1: 86565521-86565521 on Build GRCh38
rs899677302 Chr.1: 86565725-86565725 on Build GRCh38
rs758951749 Chr.1: 86565896-86565896 on Build GRCh38
rs982403358 Chr.1: 86565601-86565601 on Build GRCh38
rs940846574 Chr.1: 86565721-86565721 on Build GRCh38
rs1013911875 Chr.1: 86565564-86565564 on Build GRCh38
rs545676551 Chr.1: 86565805-86565805 on Build GRCh38
rs200731167 Chr.1: 86565766-86565766 on Build GRCh38
rs77380561 Chr.1: 86565895-86565895 on Build GRCh38
rs952121137 Chr.1: 86565925-86565925 on Build GRCh38
rs755357644 Chr.1: 86565867-86565867 on Build GRCh38
rs1054426340 Chr.1: 86565676-86565676 on Build GRCh38
rs893125132 Chr.1: 86565683-86565683 on Build GRCh38
rs930673419 Chr.1: 86565717-86565717 on Build GRCh38
rs375725423 Chr.1: 86565630-86565630 on Build GRCh38
rs760725513 Chr.1: 86565860-86565860 on Build GRCh38
rs992171986 Chr.1: 86565913-86565913 on Build GRCh38
rs998361971 Chr.1: 86565547-86565547 on Build GRCh38
rs998784514 Chr.1: 86565517-86565517 on Build GRCh38
rs200941009 Chr.1: 86565949-86565949 on Build GRCh38
rs79822589 Chr.1: 86565961-86565961 on Build GRCh38
rs756363123 Chr.1: 86565485-86565485 on Build GRCh38
rs540067219 Chr.1: 86565556-86565556 on Build GRCh38
rs962354275 Chr.1: 86565910-86565910 on Build GRCh38
rs745628367 Chr.1: 86565802-86565802 on Build GRCh38
rs772603 Chr.1: 86565632-86565632 on Build GRCh38
rs2231591 Chr.1: 86565719-86565719 on Build GRCh38
rs779532185 Chr.1: 86565768-86565768 on Build GRCh38
rs576255640 Chr.1: 86565753-86565753 on Build GRCh38
rs766598785 Chr.1: 86565988-86565988 on Build GRCh38
rs1014389278 Chr.1: 86565740-86565740 on Build GRCh38
rs1056590115 Chr.1: 86565733-86565733 on Build GRCh38
rs569459891 Chr.1: 86565663-86565663 on Build GRCh38
rs1010769191 Chr.1: 86565525-86565525 on Build GRCh38
rs951032721 Chr.1: 86565563-86565563 on Build GRCh38
rs963894305 Chr.1: 86565538-86565538 on Build GRCh38
rs749103949 Chr.1: 86565952-86565952 on Build GRCh38
rs1018553079 Chr.1: 86565528-86565528 on Build GRCh38
rs765721412 Chr.1: 86565882-86565882 on Build GRCh38
rs373050506 Chr.1: 86565842-86565842 on Build GRCh38
rs533976477 Chr.1: 86565745-86565745 on Build GRCh38
rs934665619 Chr.1: 86565670-86565670 on Build GRCh38
rs765220655 Chr.1: 86565827-86565827 on Build GRCh38
rs955974655 Chr.1: 86565801-86565801 on Build GRCh38
rs916137005 Chr.1: 86565916-86565916 on Build GRCh38
rs746564354 Chr.1: 86565792-86565792 on Build GRCh38
rs199899112 Chr.1: 86565819-86565819 on Build GRCh38
rs527247089 Chr.1: 86565531-86565531 on Build GRCh38
rs971981455 Chr.1: 86565929-86565929 on Build GRCh38
rs777359179 Chr.1: 86565788-86565788 on Build GRCh38
rs908124082 Chr.1: 86565941-86565941 on Build GRCh38
rs1023016243 Chr.1: 86565599-86565599 on Build GRCh38
rs896664083 Chr.1: 86565736-86565736 on Build GRCh38
rs770599952 Chr.1: 86565794-86565794 on Build GRCh38
rs780779596 Chr.1: 86565926-86565926 on Build GRCh38
rs559686334 Chr.1: 86565570-86565570 on Build GRCh38
rs944825343 Chr.1: 86565669-86565669 on Build GRCh38
rs748394443 Chr.1: 86565971-86565974 on Build GRCh38
rs951147671 Chr.1: 86565835-86565835 on Build GRCh38

View Full Product Details