Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 26982617-26983158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TM7SF3
Entrez Gene ID
51768
Gene Name
transmembrane 7 superfamily member 3
Gene Aliases
-
UniGene
Hs.438641
Gene Chromosome Location
Chr.12: 26966519-27014427 on Build GRCh38
Transcripts
NM_016551.2 XM_005253391.3 XM_011520690.2 XM_011520691.1 XM_017019463.1 XM_017019464.1 XM_017019465.1 XM_017019466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6284794 c.901T>G p.F301V Chr.12: 26982827-26982827 on Build GRCh38
4805532 c.922A>G p.I308V Chr.12: 26982806-26982806 on Build GRCh38
693544 c.930C>G p.F310L Chr.12: 26982798-26982798 on Build GRCh38
4943452 c.948G>A p.W316* Chr.12: 26982780-26982780 on Build GRCh38
3811736 c.935G>T p.G312V Chr.12: 26982793-26982793 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772768641 Chr.12: 26982753-26982753 on Build GRCh38
rs768589420 Chr.12: 26982735-26982735 on Build GRCh38
rs373940135 Chr.12: 26983123-26983123 on Build GRCh38
rs1027576694 Chr.12: 26983090-26983090 on Build GRCh38
rs1035942436 Chr.12: 26983105-26983105 on Build GRCh38
rs994797087 Chr.12: 26983065-26983065 on Build GRCh38
rs772288406 Chr.12: 26982716-26982716 on Build GRCh38
rs746150345 Chr.12: 26982807-26982807 on Build GRCh38
rs1053962382 Chr.12: 26982913-26982913 on Build GRCh38
rs765686884 Chr.12: 26982778-26982778 on Build GRCh38
rs778819221 Chr.12: 26982795-26982795 on Build GRCh38
rs753247477 Chr.12: 26982780-26982780 on Build GRCh38
rs778746786 Chr.12: 26982850-26982850 on Build GRCh38
rs544287344 Chr.12: 26983040-26983040 on Build GRCh38
rs757458529 Chr.12: 26982793-26982793 on Build GRCh38
rs932158351 Chr.12: 26982927-26982927 on Build GRCh38
rs765229979 Chr.12: 26982917-26982918 on Build GRCh38
rs188536801 Chr.12: 26982979-26982979 on Build GRCh38
rs975247516 Chr.12: 26982857-26982857 on Build GRCh38
rs760212102 Chr.12: 26982892-26982892 on Build GRCh38
rs921083234 Chr.12: 26982879-26982879 on Build GRCh38
rs1003069067 Chr.12: 26983101-26983101 on Build GRCh38
rs768305869 Chr.12: 26982836-26982836 on Build GRCh38
rs751692383 Chr.12: 26982791-26982791 on Build GRCh38
rs932501854 Chr.12: 26982893-26982893 on Build GRCh38
rs76200792 Chr.12: 26982985-26982985 on Build GRCh38
rs961976940 Chr.12: 26982652-26982652 on Build GRCh38
rs61923404 Chr.12: 26982657-26982657 on Build GRCh38
rs775606459 Chr.12: 26982928-26982928 on Build GRCh38
rs779226836 Chr.12: 26982667-26982667 on Build GRCh38
rs74332145 Chr.12: 26982736-26982736 on Build GRCh38
rs372752187 Chr.12: 26982924-26982924 on Build GRCh38
rs761779903 Chr.12: 26982745-26982745 on Build GRCh38
rs34653193 Chr.12: 26982861-26982862 on Build GRCh38
rs1036059996 Chr.12: 26983003-26983003 on Build GRCh38
rs577734942 Chr.12: 26983104-26983104 on Build GRCh38
rs74404575 Chr.12: 26982969-26982969 on Build GRCh38
rs939686148 Chr.12: 26982941-26982941 on Build GRCh38
rs199838819 Chr.12: 26982856-26982856 on Build GRCh38
rs754192829 Chr.12: 26982765-26982765 on Build GRCh38
rs760263534 Chr.12: 26982754-26982754 on Build GRCh38
rs772645766 Chr.12: 26982855-26982855 on Build GRCh38
rs956239089 Chr.12: 26983112-26983112 on Build GRCh38
rs771565787 Chr.12: 26982750-26982750 on Build GRCh38
rs772011376 Chr.12: 26982854-26982854 on Build GRCh38
rs748269155 Chr.12: 26982672-26982672 on Build GRCh38
rs368873949 Chr.12: 26982757-26982757 on Build GRCh38
rs777939618 Chr.12: 26982788-26982788 on Build GRCh38
rs868741287 Chr.12: 26983089-26983089 on Build GRCh38
rs376416647 Chr.12: 26982852-26982852 on Build GRCh38
rs746047391 Chr.12: 26982820-26982820 on Build GRCh38
rs373537314 Chr.12: 26982766-26982766 on Build GRCh38
rs898163135 Chr.12: 26982970-26982970 on Build GRCh38
rs192073722 Chr.12: 26983063-26983063 on Build GRCh38
rs1009047619 Chr.12: 26982990-26982990 on Build GRCh38
rs757034446 Chr.12: 26982729-26982729 on Build GRCh38
rs1050957585 Chr.12: 26982911-26982911 on Build GRCh38
rs906011581 Chr.12: 26983034-26983034 on Build GRCh38
rs988852287 Chr.12: 26982767-26982767 on Build GRCh38
rs759030108 Chr.12: 26982785-26982785 on Build GRCh38

View Full Product Details