Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115700948-115701496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASQ2
Entrez Gene ID
845
Gene Name
calsequestrin 2
Gene Aliases
PDIB2
UniGene
Hs.57975
Gene Chromosome Location
Chr.1: 115700003-115768805 on Build GRCh38
Transcripts
NM_001232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
366777 c.1123G>T p.D375Y Chr.1: 115701318-115701318 on Build GRCh38
3801390 c.1100C>G p.S367C Chr.1: 115701341-115701341 on Build GRCh38
4669404 c.1110A>C p.I370I Chr.1: 115701331-115701331 on Build GRCh38
6057417 c.1195G>T p.E399* Chr.1: 115701246-115701246 on Build GRCh38
1293151 c.1131A>T p.E377D Chr.1: 115701310-115701310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749303480 Chr.1: 115701223-115701256 on Build GRCh38
rs762324565 Chr.1: 115701302-115701310 on Build GRCh38
rs1036432828 Chr.1: 115701093-115701093 on Build GRCh38
rs575326580 Chr.1: 115701410-115701410 on Build GRCh38
rs569364415 Chr.1: 115701461-115701461 on Build GRCh38
rs765100974 Chr.1: 115701475-115701475 on Build GRCh38
rs939792810 Chr.1: 115701084-115701084 on Build GRCh38
rs72554070 Chr.1: 115701251-115701253 on Build GRCh38
rs541969897 Chr.1: 115701035-115701035 on Build GRCh38
rs771063769 Chr.1: 115701238-115701238 on Build GRCh38
rs768840860 Chr.1: 115701105-115701105 on Build GRCh38
rs921569197 Chr.1: 115701025-115701025 on Build GRCh38
rs764599645 Chr.1: 115701315-115701315 on Build GRCh38
rs397516639 Chr.1: 115701350-115701351 on Build GRCh38
rs760797081 Chr.1: 115701436-115701436 on Build GRCh38
rs894218334 Chr.1: 115701452-115701452 on Build GRCh38
rs369982880 Chr.1: 115701394-115701394 on Build GRCh38
rs776130201 Chr.1: 115701304-115701304 on Build GRCh38
rs1014029101 Chr.1: 115701465-115701465 on Build GRCh38
rs765914619 Chr.1: 115701308-115701310 on Build GRCh38
rs202241842 Chr.1: 115701311-115701313 on Build GRCh38
rs749043824 Chr.1: 115701269-115701269 on Build GRCh38
rs758060229 Chr.1: 115701391-115701391 on Build GRCh38
rs760094361 Chr.1: 115701251-115701256 on Build GRCh38
rs375286334 Chr.1: 115701245-115701245 on Build GRCh38
rs924774071 Chr.1: 115700967-115700967 on Build GRCh38
rs967682583 Chr.1: 115701172-115701172 on Build GRCh38
rs767803602 Chr.1: 115701345-115701345 on Build GRCh38
rs750480601 Chr.1: 115701258-115701258 on Build GRCh38
rs749065364 Chr.1: 115701190-115701190 on Build GRCh38
rs786205789 Chr.1: 115701256-115701256 on Build GRCh38
rs772969265 Chr.1: 115701298-115701299 on Build GRCh38
rs932876716 Chr.1: 115701044-115701044 on Build GRCh38
rs940257382 Chr.1: 115701305-115701305 on Build GRCh38
rs779986877 Chr.1: 115701264-115701264 on Build GRCh38
rs786205798 Chr.1: 115701423-115701424 on Build GRCh38
rs786205272 Chr.1: 115701256-115701258 on Build GRCh38
rs753832194 Chr.1: 115701462-115701462 on Build GRCh38
rs28730709 Chr.1: 115701098-115701098 on Build GRCh38
rs1051283799 Chr.1: 115700996-115700996 on Build GRCh38
rs756155045 Chr.1: 115701348-115701348 on Build GRCh38
rs760001239 Chr.1: 115701233-115701233 on Build GRCh38
rs886045159 Chr.1: 115701321-115701321 on Build GRCh38
rs564451231 Chr.1: 115701215-115701215 on Build GRCh38
rs199955994 Chr.1: 115701203-115701203 on Build GRCh38
rs894946581 Chr.1: 115701420-115701420 on Build GRCh38
rs771682039 Chr.1: 115701277-115701277 on Build GRCh38
rs886045158 Chr.1: 115701320-115701320 on Build GRCh38
rs786205796 Chr.1: 115701314-115701314 on Build GRCh38
rs372259035 Chr.1: 115701202-115701202 on Build GRCh38
rs762153545 Chr.1: 115701344-115701344 on Build GRCh38
rs200958192 Chr.1: 115701426-115701426 on Build GRCh38
rs563342580 Chr.1: 115701066-115701066 on Build GRCh38
rs1014069220 Chr.1: 115701147-115701147 on Build GRCh38
rs397516641 Chr.1: 115701254-115701256 on Build GRCh38
rs936149495 Chr.1: 115700976-115700976 on Build GRCh38
rs746635989 Chr.1: 115701400-115701400 on Build GRCh38
rs267597944 Chr.1: 115701390-115701390 on Build GRCh38
rs750485996 Chr.1: 115701346-115701346 on Build GRCh38
rs150486780 Chr.1: 115701307-115701307 on Build GRCh38
rs375320117 Chr.1: 115701429-115701429 on Build GRCh38
rs772619751 Chr.1: 115701291-115701291 on Build GRCh38
rs756032036 Chr.1: 115701259-115701259 on Build GRCh38
rs770530169 Chr.1: 115701403-115701403 on Build GRCh38
rs998822686 Chr.1: 115701196-115701196 on Build GRCh38
rs1024328511 Chr.1: 115701223-115701223 on Build GRCh38
rs7413162 Chr.1: 115701256-115701256 on Build GRCh38
rs768209948 Chr.1: 115701198-115701198 on Build GRCh38
rs765725948 Chr.1: 115701238-115701238 on Build GRCh38
rs1034059940 Chr.1: 115701100-115701100 on Build GRCh38
rs770204929 Chr.1: 115701296-115701296 on Build GRCh38
rs201706772 Chr.1: 115701311-115701311 on Build GRCh38
rs397516640 Chr.1: 115701293-115701293 on Build GRCh38
rs751885773 Chr.1: 115701253-115701253 on Build GRCh38
rs778009923 Chr.1: 115701206-115701216 on Build GRCh38
rs752492591 Chr.1: 115701477-115701477 on Build GRCh38
rs774882087 Chr.1: 115701415-115701415 on Build GRCh38
rs759500992 Chr.1: 115701469-115701469 on Build GRCh38
rs28730711 Chr.1: 115701247-115701247 on Build GRCh38
rs774681337 Chr.1: 115701244-115701245 on Build GRCh38
rs201492152 Chr.1: 115701163-115701163 on Build GRCh38
rs28730710 Chr.1: 115701103-115701103 on Build GRCh38
rs753872950 Chr.1: 115701367-115701367 on Build GRCh38
rs730880364 Chr.1: 115701291-115701292 on Build GRCh38
rs754836370 Chr.1: 115701271-115701271 on Build GRCh38
rs887343518 Chr.1: 115701037-115701037 on Build GRCh38
rs148824162 Chr.1: 115701310-115701310 on Build GRCh38
rs994513221 Chr.1: 115701099-115701099 on Build GRCh38
rs990715148 Chr.1: 115700990-115700990 on Build GRCh38
rs200530025 Chr.1: 115701275-115701275 on Build GRCh38
rs762214701 Chr.1: 115701419-115701419 on Build GRCh38
rs754889548 Chr.1: 115701382-115701382 on Build GRCh38
rs1050748719 Chr.1: 115701050-115701050 on Build GRCh38
rs786205792 Chr.1: 115701359-115701359 on Build GRCh38
rs375811174 Chr.1: 115701360-115701360 on Build GRCh38
rs200899037 Chr.1: 115701389-115701389 on Build GRCh38
rs72554069 Chr.1: 115701292-115701294 on Build GRCh38
rs786205793 Chr.1: 115701245-115701247 on Build GRCh38
rs786205267 Chr.1: 115701259-115701260 on Build GRCh38
rs371537492 Chr.1: 115701303-115701303 on Build GRCh38
rs780543114 Chr.1: 115701409-115701409 on Build GRCh38
rs764957093 Chr.1: 115701299-115701310 on Build GRCh38
rs747847223 Chr.1: 115701276-115701276 on Build GRCh38
rs372865933 Chr.1: 115701395-115701395 on Build GRCh38
rs754788819 Chr.1: 115701294-115701295 on Build GRCh38
rs368007942 Chr.1: 115701255-115701255 on Build GRCh38
rs775730256 Chr.1: 115701239-115701239 on Build GRCh38
rs529562535 Chr.1: 115701413-115701413 on Build GRCh38
rs774822570 Chr.1: 115701319-115701319 on Build GRCh38
rs1033929491 Chr.1: 115701118-115701118 on Build GRCh38

View Full Product Details