Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30491392-30491962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEC14L4
Entrez Gene ID
284904
Gene Name
SEC14 like lipid binding 4
Gene Aliases
TAP3
UniGene
Hs.517541
Gene Chromosome Location
Chr.22: 30488150-30505711 on Build GRCh38
Transcripts
NM_001161368.1 NM_174977.3 XM_006724235.2 XM_011530164.2 XM_017028787.1 XM_017028788.1 XM_017028789.1 XM_017028791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7153696 c.777G>T p.V259V Chr.22: 30491923-30491923 on Build GRCh38
7138247 c.954G>A p.M318I Chr.22: 30491655-30491655 on Build GRCh38
5907082 c.942T>G p.S314R Chr.22: 30491667-30491667 on Build GRCh38
2937150 c.984G>C p.Q328H Chr.22: 30491670-30491670 on Build GRCh38
3694013 c.1022G>A p.R341H Chr.22: 30491632-30491632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115178857 Chr.22: 30491681-30491681 on Build GRCh38
rs532519879 Chr.22: 30491433-30491433 on Build GRCh38
rs111263559 Chr.22: 30491427-30491427 on Build GRCh38
rs764667088 Chr.22: 30491753-30491753 on Build GRCh38
rs996413397 Chr.22: 30491418-30491418 on Build GRCh38
rs753082437 Chr.22: 30491643-30491643 on Build GRCh38
rs200834726 Chr.22: 30491770-30491770 on Build GRCh38
rs1049141683 Chr.22: 30491599-30491599 on Build GRCh38
rs867066229 Chr.22: 30491496-30491496 on Build GRCh38
rs746160644 Chr.22: 30491674-30491674 on Build GRCh38
rs922133126 Chr.22: 30491779-30491779 on Build GRCh38
rs201266658 Chr.22: 30491646-30491646 on Build GRCh38
rs563850172 Chr.22: 30491931-30491931 on Build GRCh38
rs777623894 Chr.22: 30491863-30491863 on Build GRCh38
rs770806581 Chr.22: 30491526-30491526 on Build GRCh38
rs531889100 Chr.22: 30491915-30491915 on Build GRCh38
rs370332280 Chr.22: 30491809-30491809 on Build GRCh38
rs781184596 Chr.22: 30491859-30491859 on Build GRCh38
rs745695548 Chr.22: 30491648-30491648 on Build GRCh38
rs532977655 Chr.22: 30491588-30491588 on Build GRCh38
rs375851923 Chr.22: 30491712-30491712 on Build GRCh38
rs761403546 Chr.22: 30491613-30491613 on Build GRCh38
rs370511113 Chr.22: 30491575-30491575 on Build GRCh38
rs969737491 Chr.22: 30491491-30491491 on Build GRCh38
rs753131637 Chr.22: 30491751-30491751 on Build GRCh38
rs76073588 Chr.22: 30491568-30491568 on Build GRCh38
rs367876819 Chr.22: 30491903-30491903 on Build GRCh38
rs370460762 Chr.22: 30491554-30491554 on Build GRCh38
rs183364479 Chr.22: 30491481-30491481 on Build GRCh38
rs376924335 Chr.22: 30491530-30491530 on Build GRCh38
rs767132550 Chr.22: 30491631-30491631 on Build GRCh38
rs558288860 Chr.22: 30491458-30491458 on Build GRCh38
rs1056003928 Chr.22: 30491675-30491675 on Build GRCh38
rs764612132 Chr.22: 30491828-30491828 on Build GRCh38
rs377213483 Chr.22: 30491911-30491911 on Build GRCh38
rs867694330 Chr.22: 30491742-30491742 on Build GRCh38
rs752180339 Chr.22: 30491754-30491754 on Build GRCh38
rs776570070 Chr.22: 30491541-30491541 on Build GRCh38
rs139429739 Chr.22: 30491940-30491940 on Build GRCh38
rs768045625 Chr.22: 30491896-30491896 on Build GRCh38
rs777954140 Chr.22: 30491649-30491649 on Build GRCh38
rs5753168 Chr.22: 30491514-30491514 on Build GRCh38
rs767664665 Chr.22: 30491834-30491834 on Build GRCh38
rs754216739 Chr.22: 30491562-30491562 on Build GRCh38
rs957321243 Chr.22: 30491498-30491498 on Build GRCh38
rs760384308 Chr.22: 30491732-30491732 on Build GRCh38
rs754072544 Chr.22: 30491849-30491849 on Build GRCh38
rs138793012 Chr.22: 30491576-30491576 on Build GRCh38
rs756196179 Chr.22: 30491846-30491846 on Build GRCh38
rs755193302 Chr.22: 30491907-30491907 on Build GRCh38
rs1029432930 Chr.22: 30491421-30491421 on Build GRCh38
rs780331813 Chr.22: 30491561-30491561 on Build GRCh38
rs534400617 Chr.22: 30491805-30491805 on Build GRCh38
rs201094843 Chr.22: 30491601-30491601 on Build GRCh38
rs759486934 Chr.22: 30491545-30491545 on Build GRCh38
rs760802294 Chr.22: 30491902-30491902 on Build GRCh38
rs749587670 Chr.22: 30491510-30491510 on Build GRCh38
rs368552886 Chr.22: 30491767-30491767 on Build GRCh38
rs74525490 Chr.22: 30491497-30491497 on Build GRCh38
rs374331336 Chr.22: 30491584-30491584 on Build GRCh38
rs1014973021 Chr.22: 30491445-30491445 on Build GRCh38
rs751164844 Chr.22: 30491559-30491559 on Build GRCh38
rs779287255 Chr.22: 30491567-30491567 on Build GRCh38
rs762415073 Chr.22: 30491832-30491832 on Build GRCh38
rs746499914 Chr.22: 30491864-30491864 on Build GRCh38
rs116307016 Chr.22: 30491440-30491440 on Build GRCh38
rs540038842 Chr.22: 30491934-30491934 on Build GRCh38
rs985310773 Chr.22: 30491801-30491801 on Build GRCh38
rs542989917 Chr.22: 30491869-30491869 on Build GRCh38
rs779664300 Chr.22: 30491766-30491766 on Build GRCh38
rs781435947 Chr.22: 30491758-30491758 on Build GRCh38
rs575158416 Chr.22: 30491914-30491914 on Build GRCh38
rs770597975 Chr.22: 30491787-30491787 on Build GRCh38
rs752745537 Chr.22: 30491548-30491548 on Build GRCh38
rs564283211 Chr.22: 30491577-30491577 on Build GRCh38
rs867321945 Chr.22: 30491603-30491603 on Build GRCh38
rs765687119 Chr.22: 30491640-30491640 on Build GRCh38
rs762501634 Chr.22: 30491733-30491733 on Build GRCh38
rs879164630 Chr.22: 30491468-30491468 on Build GRCh38
rs61741444 Chr.22: 30491932-30491932 on Build GRCh38
rs146280848 Chr.22: 30491667-30491667 on Build GRCh38
rs368386943 Chr.22: 30491632-30491632 on Build GRCh38
rs752055701 Chr.22: 30491830-30491830 on Build GRCh38
rs771303466 Chr.22: 30491578-30491578 on Build GRCh38
rs750544921 Chr.22: 30491760-30491760 on Build GRCh38
rs759276473 Chr.22: 30491882-30491882 on Build GRCh38
rs760199655 Chr.22: 30491774-30491774 on Build GRCh38
rs9608942 Chr.22: 30491459-30491459 on Build GRCh38
rs1011388034 Chr.22: 30491888-30491888 on Build GRCh38
rs958901588 Chr.22: 30491689-30491689 on Build GRCh38
rs192252618 Chr.22: 30491718-30491718 on Build GRCh38
rs757400230 Chr.22: 30491755-30491755 on Build GRCh38
rs571834829 Chr.22: 30491768-30491768 on Build GRCh38
rs760996476 Chr.22: 30491525-30491525 on Build GRCh38
rs773881704 Chr.22: 30491612-30491612 on Build GRCh38
rs748137933 Chr.22: 30491771-30491771 on Build GRCh38
rs750932309 Chr.22: 30491900-30491900 on Build GRCh38
rs1024220754 Chr.22: 30491450-30491450 on Build GRCh38
rs747065781 Chr.22: 30491652-30491652 on Build GRCh38
rs772722091 Chr.22: 30491722-30491722 on Build GRCh38
rs569191306 Chr.22: 30491721-30491721 on Build GRCh38
rs774887890 Chr.22: 30491889-30491889 on Build GRCh38
rs772023328 Chr.22: 30491856-30491856 on Build GRCh38
rs974941142 Chr.22: 30491776-30491776 on Build GRCh38
rs745949823 Chr.22: 30491883-30491884 on Build GRCh38
rs780290342 Chr.22: 30491848-30491848 on Build GRCh38
rs781074094 Chr.22: 30491474-30491474 on Build GRCh38
rs771508479 Chr.22: 30491773-30491773 on Build GRCh38
rs188527804 Chr.22: 30491591-30491591 on Build GRCh38
rs952239187 Chr.22: 30491795-30491795 on Build GRCh38
rs373569391 Chr.22: 30491875-30491875 on Build GRCh38
rs200372039 Chr.22: 30491887-30491887 on Build GRCh38
rs776334854 Chr.22: 30491878-30491878 on Build GRCh38
rs757455217 Chr.22: 30491653-30491653 on Build GRCh38
rs373622881 Chr.22: 30491560-30491560 on Build GRCh38
rs748041801 Chr.22: 30491854-30491854 on Build GRCh38
rs538317267 Chr.22: 30491752-30491752 on Build GRCh38
rs554458992 Chr.22: 30491821-30491821 on Build GRCh38
rs144996989 Chr.22: 30491941-30491941 on Build GRCh38
rs149720232 Chr.22: 30491838-30491838 on Build GRCh38
rs765482631 Chr.22: 30491739-30491739 on Build GRCh38
rs774319016 Chr.22: 30491714-30491714 on Build GRCh38
rs1055715342 Chr.22: 30491880-30491880 on Build GRCh38
rs920413955 Chr.22: 30491489-30491489 on Build GRCh38
rs758978373 Chr.22: 30491644-30491644 on Build GRCh38
rs372310044 Chr.22: 30491711-30491711 on Build GRCh38
rs762961704 Chr.22: 30491600-30491600 on Build GRCh38
rs775276603 Chr.22: 30491593-30491593 on Build GRCh38
rs141872549 Chr.22: 30491586-30491586 on Build GRCh38
rs375622265 Chr.22: 30491763-30491763 on Build GRCh38
rs989842374 Chr.22: 30491511-30491511 on Build GRCh38
rs578232257 Chr.22: 30491417-30491417 on Build GRCh38
rs752470201 Chr.22: 30491909-30491909 on Build GRCh38
rs764967084 Chr.22: 30491908-30491908 on Build GRCh38
rs967008401 Chr.22: 30491533-30491533 on Build GRCh38
rs199909190 Chr.22: 30491633-30491633 on Build GRCh38
rs1052066190 Chr.22: 30491539-30491539 on Build GRCh38
rs371858918 Chr.22: 30491895-30491895 on Build GRCh38
rs755431698 Chr.22: 30491566-30491566 on Build GRCh38

View Full Product Details