Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27138227-27138817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR30
Entrez Gene ID
339779
Gene Name
proline rich 30
Gene Aliases
C2orf53
UniGene
Hs.136555
Gene Chromosome Location
Chr.2: 27136847-27139464 on Build GRCh38
Transcripts
NM_178553.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4831104 c.62C>A p.T21N Chr.2: 27138268-27138268 on Build GRCh38
4532608 c.18G>A p.K6K Chr.2: 27138312-27138312 on Build GRCh38
5693594 c.50C>A p.P17H Chr.2: 27138280-27138280 on Build GRCh38
5695122 c.94C>T p.P32S Chr.2: 27138236-27138236 on Build GRCh38
5396789 c.36G>A p.Q12Q Chr.2: 27138294-27138294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143780871 Chr.2: 27138331-27138331 on Build GRCh38
rs915588033 Chr.2: 27138384-27138385 on Build GRCh38
rs771195790 Chr.2: 27138723-27138723 on Build GRCh38
rs374970677 Chr.2: 27138280-27138280 on Build GRCh38
rs905240671 Chr.2: 27138557-27138557 on Build GRCh38
rs920332655 Chr.2: 27138680-27138680 on Build GRCh38
rs890675452 Chr.2: 27138468-27138468 on Build GRCh38
rs892651785 Chr.2: 27138736-27138736 on Build GRCh38
rs760470470 Chr.2: 27138330-27138330 on Build GRCh38
rs567852649 Chr.2: 27138478-27138478 on Build GRCh38
rs778180516 Chr.2: 27138658-27138658 on Build GRCh38
rs878965385 Chr.2: 27138479-27138479 on Build GRCh38
rs753140818 Chr.2: 27138365-27138365 on Build GRCh38
rs536340924 Chr.2: 27138780-27138780 on Build GRCh38
rs779428124 Chr.2: 27138394-27138394 on Build GRCh38
rs770519932 Chr.2: 27138302-27138302 on Build GRCh38
rs755532316 Chr.2: 27138272-27138272 on Build GRCh38
rs34036706 Chr.2: 27138319-27138320 on Build GRCh38
rs946902742 Chr.2: 27138747-27138747 on Build GRCh38
rs62128708 Chr.2: 27138550-27138550 on Build GRCh38
rs775275641 Chr.2: 27138329-27138329 on Build GRCh38
rs756539150 Chr.2: 27138366-27138366 on Build GRCh38
rs752161876 Chr.2: 27138261-27138261 on Build GRCh38
rs73921459 Chr.2: 27138678-27138678 on Build GRCh38
rs1037386950 Chr.2: 27138423-27138423 on Build GRCh38
rs554145658 Chr.2: 27138726-27138726 on Build GRCh38
rs1050697836 Chr.2: 27138467-27138467 on Build GRCh38
rs940414402 Chr.2: 27138416-27138416 on Build GRCh38
rs777438697 Chr.2: 27138282-27138282 on Build GRCh38
rs2304680 Chr.2: 27138378-27138378 on Build GRCh38
rs750923648 Chr.2: 27138352-27138352 on Build GRCh38
rs1011437277 Chr.2: 27138544-27138544 on Build GRCh38
rs1010975157 Chr.2: 27138606-27138606 on Build GRCh38
rs376148989 Chr.2: 27138395-27138395 on Build GRCh38
rs1053627335 Chr.2: 27138649-27138671 on Build GRCh38
rs1015144218 Chr.2: 27138651-27138651 on Build GRCh38
rs747221538 Chr.2: 27138692-27138692 on Build GRCh38
rs992695135 Chr.2: 27138636-27138636 on Build GRCh38
rs549359564 Chr.2: 27138383-27138383 on Build GRCh38
rs1007437923 Chr.2: 27138656-27138656 on Build GRCh38
rs985753304 Chr.2: 27138696-27138696 on Build GRCh38
rs770218623 Chr.2: 27138588-27138588 on Build GRCh38
rs367907710 Chr.2: 27138296-27138296 on Build GRCh38
rs1034237559 Chr.2: 27138574-27138574 on Build GRCh38
rs765974242 Chr.2: 27138346-27138346 on Build GRCh38
rs13419636 Chr.2: 27138720-27138720 on Build GRCh38
rs892564695 Chr.2: 27138522-27138522 on Build GRCh38
rs565675953 Chr.2: 27138763-27138763 on Build GRCh38
rs953543598 Chr.2: 27138655-27138655 on Build GRCh38
rs2304679 Chr.2: 27138354-27138354 on Build GRCh38
rs571583226 Chr.2: 27138666-27138666 on Build GRCh38
rs920288583 Chr.2: 27138446-27138446 on Build GRCh38
rs961894228 Chr.2: 27138675-27138675 on Build GRCh38
rs374317460 Chr.2: 27138699-27138699 on Build GRCh38
rs969345653 Chr.2: 27138295-27138295 on Build GRCh38
rs915575794 Chr.2: 27138756-27138756 on Build GRCh38
rs543742544 Chr.2: 27138305-27138305 on Build GRCh38
rs751688433 Chr.2: 27138353-27138353 on Build GRCh38
rs145535878 Chr.2: 27138277-27138277 on Build GRCh38
rs550079995 Chr.2: 27138663-27138663 on Build GRCh38
rs931619629 Chr.2: 27138453-27138453 on Build GRCh38
rs771927604 Chr.2: 27138328-27138328 on Build GRCh38
rs906661599 Chr.2: 27138508-27138508 on Build GRCh38
rs951760901 Chr.2: 27138724-27138724 on Build GRCh38
rs956964990 Chr.2: 27138260-27138260 on Build GRCh38
rs200670608 Chr.2: 27138326-27138326 on Build GRCh38
rs753159897 Chr.2: 27138512-27138512 on Build GRCh38
rs1002153219 Chr.2: 27138567-27138567 on Build GRCh38
rs779346142 Chr.2: 27138316-27138316 on Build GRCh38
rs781734497 Chr.2: 27138275-27138275 on Build GRCh38
rs201696070 Chr.2: 27138358-27138358 on Build GRCh38
rs1003673679 Chr.2: 27138524-27138524 on Build GRCh38
rs948392802 Chr.2: 27138770-27138770 on Build GRCh38

View Full Product Details