Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 193618540-193619111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPA1
Entrez Gene ID
4976
Gene Name
OPA1, mitochondrial dynamin like GTPase
Gene Aliases
BERHS, MGM1, MTDPS14, NPG, NTG, largeG
UniGene
Hs.594504
Gene Chromosome Location
Chr.3: 193593144-193697811 on Build GRCh38
Transcripts
NM_015560.2 NM_130831.2 NM_130832.2 NM_130833.2 NM_130834.2 NM_130835.2 NM_130836.2 NM_130837.2 XM_017006500.1 NR_046634.1

Gene Symbol
OPA1-AS1
Entrez Gene ID
100873941
Gene Name
OPA1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.679364
Gene Chromosome Location
Chr.3: 193618599-193627332 on Build GRCh38
Transcripts
NM_015560.2 NM_130831.2 NM_130832.2 NM_130833.2 NM_130834.2 NM_130835.2 NM_130836.2 NM_130837.2 XM_017006500.1 NR_046634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4932711 c.650G>C p.G217A Chr.3: 193618908-193618908 on Build GRCh38
7352012 c.611-1G>A p.? Chr.3: 193618868-193618868 on Build GRCh38
3591497 c.653C>T p.S218F Chr.3: 193618911-193618911 on Build GRCh38
1193810 c.655G>A p.E219K Chr.3: 193618913-193618913 on Build GRCh38
7348247 c.646C>T p.R216C Chr.3: 193618904-193618904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749863596 Chr.3: 193618673-193618676 on Build GRCh38
rs972120723 Chr.3: 193618819-193618819 on Build GRCh38
rs200243596 Chr.3: 193618911-193618911 on Build GRCh38
rs780784567 Chr.3: 193618982-193618982 on Build GRCh38
rs902132310 Chr.3: 193618991-193618991 on Build GRCh38
rs747996365 Chr.3: 193618986-193618986 on Build GRCh38
rs766723697 Chr.3: 193618954-193618954 on Build GRCh38
rs371781817 Chr.3: 193618947-193618947 on Build GRCh38
rs796479349 Chr.3: 193618639-193618639 on Build GRCh38
rs895264492 Chr.3: 193618603-193618603 on Build GRCh38
rs745444482 Chr.3: 193618762-193618762 on Build GRCh38
rs576969867 Chr.3: 193618679-193618680 on Build GRCh38
rs761083696 Chr.3: 193618946-193618946 on Build GRCh38
rs556511087 Chr.3: 193618816-193618816 on Build GRCh38
rs749933498 Chr.3: 193618964-193618964 on Build GRCh38
rs772596317 Chr.3: 193618884-193618884 on Build GRCh38
rs1015950949 Chr.3: 193618796-193618796 on Build GRCh38
rs534089185 Chr.3: 193618699-193618699 on Build GRCh38
rs190453962 Chr.3: 193619070-193619070 on Build GRCh38
rs758025983 Chr.3: 193618965-193618965 on Build GRCh38
rs551744312 Chr.3: 193618644-193618644 on Build GRCh38
rs374767769 Chr.3: 193618981-193618981 on Build GRCh38
rs775620290 Chr.3: 193618876-193618876 on Build GRCh38
rs772090345 Chr.3: 193618875-193618875 on Build GRCh38
rs543112669 Chr.3: 193619064-193619064 on Build GRCh38
rs777475151 Chr.3: 193618840-193618840 on Build GRCh38
rs938620488 Chr.3: 193618567-193618567 on Build GRCh38
rs773472534 Chr.3: 193618885-193618885 on Build GRCh38
rs574576946 Chr.3: 193619063-193619063 on Build GRCh38
rs894086848 Chr.3: 193619023-193619023 on Build GRCh38
rs997729774 Chr.3: 193618680-193618680 on Build GRCh38
rs756926862 Chr.3: 193618960-193618960 on Build GRCh38
rs748949575 Chr.3: 193618849-193618849 on Build GRCh38
rs1055606090 Chr.3: 193618606-193618606 on Build GRCh38
rs759660552 Chr.3: 193618893-193618893 on Build GRCh38
rs145678823 Chr.3: 193618689-193618689 on Build GRCh38
rs367814426 Chr.3: 193618919-193618919 on Build GRCh38
rs3772393 Chr.3: 193618850-193618850 on Build GRCh38
rs997109202 Chr.3: 193618707-193618707 on Build GRCh38
rs1021097854 Chr.3: 193619060-193619060 on Build GRCh38
rs1011474945 Chr.3: 193619026-193619026 on Build GRCh38
rs563180004 Chr.3: 193618631-193618631 on Build GRCh38
rs774364044 Chr.3: 193618890-193618890 on Build GRCh38
rs561115644 Chr.3: 193618949-193618949 on Build GRCh38
rs751938009 Chr.3: 193618980-193618980 on Build GRCh38
rs745405591 Chr.3: 193618869-193618869 on Build GRCh38
rs1021445268 Chr.3: 193618795-193618795 on Build GRCh38
rs778734134 Chr.3: 193618961-193618961 on Build GRCh38
rs185914758 Chr.3: 193618826-193618826 on Build GRCh38
rs780137912 Chr.3: 193618968-193618968 on Build GRCh38
rs886884265 Chr.3: 193618664-193618664 on Build GRCh38
rs567398473 Chr.3: 193618798-193618798 on Build GRCh38
rs550849134 Chr.3: 193618721-193618722 on Build GRCh38
rs989698174 Chr.3: 193618785-193618785 on Build GRCh38
rs770938026 Chr.3: 193619008-193619008 on Build GRCh38
rs372157755 Chr.3: 193618860-193618860 on Build GRCh38
rs181430974 Chr.3: 193618750-193618750 on Build GRCh38
rs34307082 Chr.3: 193618887-193618887 on Build GRCh38
rs746979322 Chr.3: 193618977-193618977 on Build GRCh38
rs766593183 Chr.3: 193618888-193618888 on Build GRCh38
rs748510302 Chr.3: 193618854-193618854 on Build GRCh38
rs1019307224 Chr.3: 193618675-193618675 on Build GRCh38
rs756474187 Chr.3: 193618839-193618839 on Build GRCh38
rs112720126 Chr.3: 193618648-193618648 on Build GRCh38
rs571632534 Chr.3: 193618672-193618672 on Build GRCh38
rs900841367 Chr.3: 193618979-193618979 on Build GRCh38
rs984505069 Chr.3: 193618790-193618790 on Build GRCh38
rs536391293 Chr.3: 193618810-193618810 on Build GRCh38
rs1029169789 Chr.3: 193619009-193619009 on Build GRCh38
rs541206547 Chr.3: 193618905-193618905 on Build GRCh38
rs569675223 Chr.3: 193618877-193618877 on Build GRCh38
rs935376325 Chr.3: 193618980-193618980 on Build GRCh38
rs761489870 Chr.3: 193618944-193618953 on Build GRCh38
rs9832709 Chr.3: 193618636-193618636 on Build GRCh38
rs965588584 Chr.3: 193618703-193618703 on Build GRCh38
rs1003862595 Chr.3: 193618665-193618665 on Build GRCh38

View Full Product Details