Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98579254-98579785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPOX
Entrez Gene ID
1371
Gene Name
coproporphyrinogen oxidase
Gene Aliases
CPO, CPX, HCP
UniGene
Hs.476982
Gene Chromosome Location
Chr.3: 98568448-98593684 on Build GRCh38
Transcripts
NM_000097.5 XM_005247125.4

SNP Detail

SNP ID Chromosome Location
rs866149655 Chr.3: 98579306-98579306 on Build GRCh38
rs948833716 Chr.3: 98579559-98579559 on Build GRCh38
rs916170620 Chr.3: 98579534-98579534 on Build GRCh38
rs773271834 Chr.3: 98579434-98579434 on Build GRCh38
rs994798130 Chr.3: 98579361-98579361 on Build GRCh38
rs966117303 Chr.3: 98579404-98579404 on Build GRCh38
rs775384142 Chr.3: 98579607-98579607 on Build GRCh38
rs906291457 Chr.3: 98579285-98579285 on Build GRCh38
rs1023676235 Chr.3: 98579459-98579459 on Build GRCh38
rs889695429 Chr.3: 98579298-98579299 on Build GRCh38
rs1023195940 Chr.3: 98579757-98579757 on Build GRCh38
rs889044351 Chr.3: 98579687-98579687 on Build GRCh38
rs148251059 Chr.3: 98579611-98579611 on Build GRCh38
rs957850337 Chr.3: 98579468-98579468 on Build GRCh38
rs149544416 Chr.3: 98579369-98579369 on Build GRCh38
rs367770478 Chr.3: 98579579-98579579 on Build GRCh38
rs901922133 Chr.3: 98579711-98579711 on Build GRCh38
rs994635551 Chr.3: 98579371-98579371 on Build GRCh38
rs1032087466 Chr.3: 98579733-98579733 on Build GRCh38
rs977807792 Chr.3: 98579418-98579418 on Build GRCh38
rs570661497 Chr.3: 98579694-98579694 on Build GRCh38
rs763280780 Chr.3: 98579560-98579560 on Build GRCh38
rs1036050208 Chr.3: 98579321-98579321 on Build GRCh38
rs982386968 Chr.3: 98579509-98579509 on Build GRCh38
rs914328221 Chr.3: 98579564-98579564 on Build GRCh38
rs563018310 Chr.3: 98579443-98579443 on Build GRCh38
rs990456890 Chr.3: 98579486-98579486 on Build GRCh38
rs888533206 Chr.3: 98579724-98579724 on Build GRCh38
rs886058940 Chr.3: 98579509-98579509 on Build GRCh38
rs190427981 Chr.3: 98579277-98579277 on Build GRCh38
rs1027897143 Chr.3: 98579364-98579364 on Build GRCh38
rs1048744880 Chr.3: 98579721-98579721 on Build GRCh38
rs1023622463 Chr.3: 98579754-98579754 on Build GRCh38
rs886058941 Chr.3: 98579530-98579530 on Build GRCh38
rs539237301 Chr.3: 98579744-98579744 on Build GRCh38
rs897720741 Chr.3: 98579333-98579333 on Build GRCh38
rs1616167 Chr.3: 98579403-98579403 on Build GRCh38
rs116113441 Chr.3: 98579367-98579367 on Build GRCh38
rs939110503 Chr.3: 98579589-98579589 on Build GRCh38
rs547878913 Chr.3: 98579658-98579658 on Build GRCh38
rs896951176 Chr.3: 98579639-98579639 on Build GRCh38
rs148143224 Chr.3: 98579350-98579350 on Build GRCh38
rs947163287 Chr.3: 98579574-98579574 on Build GRCh38
rs763864217 Chr.3: 98579637-98579637 on Build GRCh38
rs73133922 Chr.3: 98579578-98579578 on Build GRCh38
rs548611832 Chr.3: 98579563-98579563 on Build GRCh38
rs35926732 Chr.3: 98579732-98579733 on Build GRCh38
rs531771288 Chr.3: 98579555-98579555 on Build GRCh38
rs1007323062 Chr.3: 98579697-98579697 on Build GRCh38
rs998984529 Chr.3: 98579722-98579722 on Build GRCh38
rs970955485 Chr.3: 98579506-98579506 on Build GRCh38
rs7103 Chr.3: 98579756-98579756 on Build GRCh38
rs1018757410 Chr.3: 98579699-98579699 on Build GRCh38
rs956909823 Chr.3: 98579551-98579551 on Build GRCh38
rs988934012 Chr.3: 98579554-98579554 on Build GRCh38
rs897527256 Chr.3: 98579608-98579608 on Build GRCh38

View Full Product Details