Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63985914-63986453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUB1
Entrez Gene ID
55611
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
Gene Aliases
HSPC263, OTB1, OTU1
UniGene
Hs.473788
Gene Chromosome Location
Chr.11: 63985853-63998420 on Build GRCh38
Transcripts
NM_017670.2 NR_003089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6995349 c.16-19_16-2del18 p.? Chr.11: 63986342-63986359 on Build GRCh38
5960254 c.31delG p.A12fs*20 Chr.11: 63986376-63986376 on Build GRCh38
4408119 c.64C>T p.R22W Chr.11: 63986409-63986409 on Build GRCh38
5223894 c.65G>C p.R22P Chr.11: 63986410-63986410 on Build GRCh38
6988150 c.16-1G>T p.? Chr.11: 63986360-63986360 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs949543638 Chr.11: 63986019-63986019 on Build GRCh38
rs1035074456 Chr.11: 63986113-63986113 on Build GRCh38
rs769102907 Chr.11: 63986398-63986398 on Build GRCh38
rs946533597 Chr.11: 63986405-63986405 on Build GRCh38
rs1014677033 Chr.11: 63986310-63986310 on Build GRCh38
rs1039276070 Chr.11: 63985972-63985972 on Build GRCh38
rs1052171301 Chr.11: 63985937-63985937 on Build GRCh38
rs972507939 Chr.11: 63986316-63986316 on Build GRCh38
rs573757280 Chr.11: 63986360-63986360 on Build GRCh38
rs570509482 Chr.11: 63985963-63985963 on Build GRCh38
rs749783159 Chr.11: 63986396-63986396 on Build GRCh38
rs961846060 Chr.11: 63986317-63986317 on Build GRCh38
rs929316459 Chr.11: 63985934-63985934 on Build GRCh38
rs757399684 Chr.11: 63986117-63986117 on Build GRCh38
rs909801020 Chr.11: 63986284-63986284 on Build GRCh38
rs962520704 Chr.11: 63986296-63986296 on Build GRCh38
rs919356763 Chr.11: 63986343-63986343 on Build GRCh38
rs1046577578 Chr.11: 63986020-63986020 on Build GRCh38
rs538532170 Chr.11: 63985948-63985948 on Build GRCh38
rs567318533 Chr.11: 63986074-63986074 on Build GRCh38
rs386754310 Chr.11: 63986089-63986090 on Build GRCh38
rs369689358 Chr.11: 63986408-63986408 on Build GRCh38
rs940408370 Chr.11: 63985940-63985940 on Build GRCh38
rs558456346 Chr.11: 63985974-63985974 on Build GRCh38
rs920321634 Chr.11: 63986325-63986325 on Build GRCh38
rs1028925686 Chr.11: 63986299-63986299 on Build GRCh38
rs1008251979 Chr.11: 63985969-63985969 on Build GRCh38
rs553751425 Chr.11: 63986234-63986234 on Build GRCh38
rs747998320 Chr.11: 63986258-63986259 on Build GRCh38
rs954606724 Chr.11: 63986303-63986303 on Build GRCh38
rs929360239 Chr.11: 63986361-63986361 on Build GRCh38
rs752548901 Chr.11: 63986083-63986083 on Build GRCh38
rs763932949 Chr.11: 63986094-63986094 on Build GRCh38
rs751374640 Chr.11: 63986103-63986103 on Build GRCh38
rs750456434 Chr.11: 63986186-63986186 on Build GRCh38
rs149677251 Chr.11: 63985966-63985967 on Build GRCh38
rs756221352 Chr.11: 63986293-63986293 on Build GRCh38
rs754932521 Chr.11: 63986118-63986118 on Build GRCh38
rs991171667 Chr.11: 63986273-63986273 on Build GRCh38
rs779333248 Chr.11: 63986411-63986411 on Build GRCh38
rs781402324 Chr.11: 63986180-63986180 on Build GRCh38
rs543122498 Chr.11: 63986384-63986384 on Build GRCh38
rs890733869 Chr.11: 63985962-63985962 on Build GRCh38
rs1020717372 Chr.11: 63986051-63986051 on Build GRCh38
rs34689071 Chr.11: 63986075-63986075 on Build GRCh38
rs562913442 Chr.11: 63986391-63986391 on Build GRCh38
rs973166950 Chr.11: 63986321-63986321 on Build GRCh38
rs56813604 Chr.11: 63985981-63985982 on Build GRCh38
rs916854953 Chr.11: 63986364-63986364 on Build GRCh38
rs996569313 Chr.11: 63986286-63986286 on Build GRCh38
rs577603695 Chr.11: 63985976-63985976 on Build GRCh38

View Full Product Details