Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 105110827-105111247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1033
Entrez Gene ID
23325
Gene Name
KIAA1033
Gene Aliases
MRT43, SWIP
UniGene
Hs.12144
Gene Chromosome Location
Chr.12: 105107714-105169134 on Build GRCh38
Transcripts
NM_001293640.1 NM_015275.2 XM_011538073.2 XM_011538074.2 XM_011538077.2 XM_017019082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3810847 c.88A>C p.N30H Chr.12: 105111151-105111151 on Build GRCh38
4186638 c.151G>T p.D51Y Chr.12: 105111214-105111214 on Build GRCh38
4838840 c.73G>A p.E25K Chr.12: 105111136-105111136 on Build GRCh38
7373552 c.112T>C p.Y38H Chr.12: 105111175-105111175 on Build GRCh38
5374022 c.62-1G>A p.? Chr.12: 105111124-105111124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974959002 Chr.12: 105110864-105110864 on Build GRCh38
rs200700259 Chr.12: 105111086-105111086 on Build GRCh38
rs536953726 Chr.12: 105111220-105111220 on Build GRCh38
rs563733109 Chr.12: 105110916-105110916 on Build GRCh38
rs907643031 Chr.12: 105110910-105110910 on Build GRCh38
rs774589525 Chr.12: 105111189-105111189 on Build GRCh38
rs751867701 Chr.12: 105111085-105111085 on Build GRCh38
rs759625987 Chr.12: 105111198-105111198 on Build GRCh38
rs577292869 Chr.12: 105111041-105111041 on Build GRCh38
rs376604258 Chr.12: 105111217-105111217 on Build GRCh38
rs372395687 Chr.12: 105111216-105111216 on Build GRCh38
rs367807155 Chr.12: 105111141-105111141 on Build GRCh38
rs532409419 Chr.12: 105111145-105111145 on Build GRCh38
rs955034316 Chr.12: 105111199-105111199 on Build GRCh38
rs1196839 Chr.12: 105110986-105110986 on Build GRCh38
rs373669216 Chr.12: 105111103-105111103 on Build GRCh38
rs776428689 Chr.12: 105111074-105111074 on Build GRCh38
rs767793363 Chr.12: 105111092-105111092 on Build GRCh38
rs745332143 Chr.12: 105111134-105111134 on Build GRCh38
rs35294333 Chr.12: 105111175-105111175 on Build GRCh38
rs1043056859 Chr.12: 105111159-105111159 on Build GRCh38
rs147988679 Chr.12: 105111180-105111180 on Build GRCh38
rs58670310 Chr.12: 105110868-105110868 on Build GRCh38
rs766343767 Chr.12: 105111082-105111082 on Build GRCh38
rs961746627 Chr.12: 105110922-105110922 on Build GRCh38
rs1042083862 Chr.12: 105111073-105111073 on Build GRCh38
rs559291934 Chr.12: 105111035-105111035 on Build GRCh38
rs954485995 Chr.12: 105110888-105110888 on Build GRCh38
rs147112620 Chr.12: 105110935-105110935 on Build GRCh38
rs767441928 Chr.12: 105110922-105110924 on Build GRCh38
rs777596923 Chr.12: 105111221-105111221 on Build GRCh38
rs968900508 Chr.12: 105110854-105110854 on Build GRCh38
rs754194473 Chr.12: 105111219-105111219 on Build GRCh38
rs746872657 Chr.12: 105111156-105111156 on Build GRCh38
rs779613567 Chr.12: 105111144-105111144 on Build GRCh38
rs866295026 Chr.12: 105111154-105111154 on Build GRCh38
rs573755173 Chr.12: 105111001-105111001 on Build GRCh38
rs374922043 Chr.12: 105111042-105111042 on Build GRCh38
rs753120367 Chr.12: 105111095-105111095 on Build GRCh38
rs778402246 Chr.12: 105111102-105111102 on Build GRCh38
rs771724807 Chr.12: 105111135-105111135 on Build GRCh38
rs768318313 Chr.12: 105111170-105111170 on Build GRCh38
rs927747954 Chr.12: 105111053-105111053 on Build GRCh38
rs1196838 Chr.12: 105111009-105111009 on Build GRCh38
rs987622804 Chr.12: 105110905-105110906 on Build GRCh38
rs763147384 Chr.12: 105111075-105111075 on Build GRCh38
rs756546800 Chr.12: 105111098-105111098 on Build GRCh38
rs373183026 Chr.12: 105110908-105110908 on Build GRCh38
rs947935254 Chr.12: 105111012-105111012 on Build GRCh38
rs368178226 Chr.12: 105111204-105111204 on Build GRCh38
rs754493327 Chr.12: 105111023-105111023 on Build GRCh38
rs973316672 Chr.12: 105110993-105110993 on Build GRCh38
rs984827522 Chr.12: 105110878-105110878 on Build GRCh38
rs775845456 Chr.12: 105111207-105111207 on Build GRCh38

View Full Product Details