Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9344360-9344953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASAP2
Entrez Gene ID
8853
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
Gene Aliases
AMAP2, CENTB3, DDEF2, PAG3, PAP, Pap-alpha, SHAG1
UniGene
Hs.555902
Gene Chromosome Location
Chr.2: 9206765-9405683 on Build GRCh38
Transcripts
NM_001135191.1 NM_003887.2 XM_006711898.1 XM_006711901.3 XM_006711902.1 XM_011510403.2 XM_011510404.2 XM_011510405.2 XM_011510406.2 XM_011510407.2 XM_011510408.2 XM_011510409.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5761582 c.901A>G p.N301D Chr.2: 9344583-9344583 on Build GRCh38
1023644 c.1001T>G p.F334C Chr.2: 9344778-9344778 on Build GRCh38
1023643 c.862C>T p.R288C Chr.2: 9344544-9344544 on Build GRCh38
7078435 c.901A>T p.N301Y Chr.2: 9344583-9344583 on Build GRCh38
5518274 c.951C>T p.D317D Chr.2: 9344633-9344633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745812154 Chr.2: 9344713-9344713 on Build GRCh38
rs540952400 Chr.2: 9344901-9344901 on Build GRCh38
rs769838169 Chr.2: 9344620-9344620 on Build GRCh38
rs760405024 Chr.2: 9344644-9344644 on Build GRCh38
rs897514775 Chr.2: 9344461-9344461 on Build GRCh38
rs763602369 Chr.2: 9344645-9344645 on Build GRCh38
rs776492234 Chr.2: 9344737-9344737 on Build GRCh38
rs761935035 Chr.2: 9344610-9344610 on Build GRCh38
rs143591778 Chr.2: 9344800-9344800 on Build GRCh38
rs953091283 Chr.2: 9344427-9344427 on Build GRCh38
rs893657327 Chr.2: 9344917-9344917 on Build GRCh38
rs751021676 Chr.2: 9344654-9344654 on Build GRCh38
rs567533726 Chr.2: 9344446-9344446 on Build GRCh38
rs905087721 Chr.2: 9344561-9344561 on Build GRCh38
rs375691674 Chr.2: 9344858-9344858 on Build GRCh38
rs942200201 Chr.2: 9344680-9344680 on Build GRCh38
rs760314853 Chr.2: 9344836-9344836 on Build GRCh38
rs369467344 Chr.2: 9344671-9344671 on Build GRCh38
rs767296380 Chr.2: 9344523-9344523 on Build GRCh38
rs777343307 Chr.2: 9344541-9344541 on Build GRCh38
rs866576370 Chr.2: 9344864-9344864 on Build GRCh38
rs779611839 Chr.2: 9344569-9344569 on Build GRCh38
rs375874873 Chr.2: 9344866-9344866 on Build GRCh38
rs576089706 Chr.2: 9344633-9344633 on Build GRCh38
rs755665966 Chr.2: 9344540-9344540 on Build GRCh38
rs758969973 Chr.2: 9344807-9344807 on Build GRCh38
rs762982469 Chr.2: 9344507-9344507 on Build GRCh38
rs905113318 Chr.2: 9344513-9344513 on Build GRCh38
rs769920271 Chr.2: 9344674-9344674 on Build GRCh38
rs1016942470 Chr.2: 9344705-9344705 on Build GRCh38
rs755369265 Chr.2: 9344841-9344841 on Build GRCh38
rs748368782 Chr.2: 9344673-9344673 on Build GRCh38
rs1018621322 Chr.2: 9344868-9344868 on Build GRCh38
rs761540224 Chr.2: 9344787-9344787 on Build GRCh38
rs371588307 Chr.2: 9344648-9344648 on Build GRCh38
rs747290099 Chr.2: 9344670-9344670 on Build GRCh38
rs559656538 Chr.2: 9344487-9344487 on Build GRCh38
rs111761084 Chr.2: 9344893-9344893 on Build GRCh38
rs200401085 Chr.2: 9344441-9344441 on Build GRCh38
rs201120621 Chr.2: 9344514-9344514 on Build GRCh38
rs35424556 Chr.2: 9344462-9344463 on Build GRCh38
rs766847798 Chr.2: 9344812-9344812 on Build GRCh38
rs774339512 Chr.2: 9344621-9344621 on Build GRCh38
rs184080718 Chr.2: 9344400-9344400 on Build GRCh38
rs774456784 Chr.2: 9344640-9344640 on Build GRCh38
rs771965273 Chr.2: 9344707-9344707 on Build GRCh38
rs147850846 Chr.2: 9344436-9344436 on Build GRCh38
rs772540793 Chr.2: 9344494-9344494 on Build GRCh38
rs772628333 Chr.2: 9344593-9344593 on Build GRCh38
rs776797223 Chr.2: 9344609-9344609 on Build GRCh38
rs758915186 Chr.2: 9344656-9344656 on Build GRCh38
rs768723108 Chr.2: 9344605-9344605 on Build GRCh38
rs751479586 Chr.2: 9344515-9344515 on Build GRCh38
rs1056380934 Chr.2: 9344597-9344597 on Build GRCh38
rs759370018 Chr.2: 9344516-9344516 on Build GRCh38
rs79044067 Chr.2: 9344433-9344436 on Build GRCh38
rs753245751 Chr.2: 9344545-9344545 on Build GRCh38
rs764850926 Chr.2: 9344799-9344799 on Build GRCh38
rs777760631 Chr.2: 9344801-9344802 on Build GRCh38
rs780267210 Chr.2: 9344600-9344600 on Build GRCh38
rs776513072 Chr.2: 9344505-9344505 on Build GRCh38
rs760434449 Chr.2: 9344723-9344723 on Build GRCh38
rs373425778 Chr.2: 9344442-9344442 on Build GRCh38
rs753508196 Chr.2: 9344589-9344589 on Build GRCh38
rs753907080 Chr.2: 9344842-9344843 on Build GRCh38
rs757858736 Chr.2: 9344559-9344559 on Build GRCh38
rs772723813 Chr.2: 9344793-9344793 on Build GRCh38
rs753506040 Chr.2: 9344647-9344647 on Build GRCh38
rs771866390 Chr.2: 9344715-9344715 on Build GRCh38
rs747266776 Chr.2: 9344604-9344604 on Build GRCh38
rs763918419 Chr.2: 9344731-9344731 on Build GRCh38
rs1020353303 Chr.2: 9344535-9344535 on Build GRCh38
rs746342296 Chr.2: 9344574-9344574 on Build GRCh38
rs1032601093 Chr.2: 9344520-9344520 on Build GRCh38
rs773219130 Chr.2: 9344690-9344690 on Build GRCh38
rs551957892 Chr.2: 9344396-9344396 on Build GRCh38
rs768834729 Chr.2: 9344500-9344500 on Build GRCh38
rs536801863 Chr.2: 9344495-9344495 on Build GRCh38
rs141948077 Chr.2: 9344434-9344434 on Build GRCh38
rs781608259 Chr.2: 9344843-9344843 on Build GRCh38
rs397756780 Chr.2: 9344439-9344442 on Build GRCh38
rs374692643 Chr.2: 9344498-9344498 on Build GRCh38
rs147574074 Chr.2: 9344435-9344435 on Build GRCh38
rs1045006619 Chr.2: 9344555-9344555 on Build GRCh38
rs949075993 Chr.2: 9344828-9344828 on Build GRCh38
rs898682130 Chr.2: 9344719-9344719 on Build GRCh38
rs987899909 Chr.2: 9344898-9344898 on Build GRCh38
rs6724103 Chr.2: 9344413-9344413 on Build GRCh38
rs768678377 Chr.2: 9344534-9344534 on Build GRCh38
rs775376099 Chr.2: 9344717-9344717 on Build GRCh38
rs374869948 Chr.2: 9344668-9344668 on Build GRCh38

View Full Product Details