Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27499011-27499523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCKR
Entrez Gene ID
2646
Gene Name
glucokinase (hexokinase 4) regulator
Gene Aliases
FGQTL5, GKRP
UniGene
Hs.89771
Gene Chromosome Location
Chr.2: 27496839-27523689 on Build GRCh38
Transcripts
NM_001486.3 XM_011532763.1 XM_017003796.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
387008 c.431C>A p.S144Y Chr.2: 27499144-27499144 on Build GRCh38
1019728 c.448G>A p.E150K Chr.2: 27499161-27499161 on Build GRCh38
384372 c.513G>T p.K171N Chr.2: 27499414-27499414 on Build GRCh38
6670014 c.524T>G p.V175G Chr.2: 27499425-27499425 on Build GRCh38
1019730 c.548C>A p.S183Y Chr.2: 27499449-27499449 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024264338 Chr.2: 27499091-27499091 on Build GRCh38
rs763708498 Chr.2: 27499185-27499185 on Build GRCh38
rs377307319 Chr.2: 27499488-27499488 on Build GRCh38
rs770731800 Chr.2: 27499356-27499356 on Build GRCh38
rs752753879 Chr.2: 27499441-27499441 on Build GRCh38
rs534572958 Chr.2: 27499319-27499319 on Build GRCh38
rs757507770 Chr.2: 27499454-27499454 on Build GRCh38
rs900675420 Chr.2: 27499041-27499041 on Build GRCh38
rs370591964 Chr.2: 27499273-27499273 on Build GRCh38
rs922938036 Chr.2: 27499237-27499237 on Build GRCh38
rs776352577 Chr.2: 27499485-27499488 on Build GRCh38
rs200836461 Chr.2: 27499202-27499202 on Build GRCh38
rs373221901 Chr.2: 27499468-27499468 on Build GRCh38
rs1003714969 Chr.2: 27499181-27499181 on Build GRCh38
rs780121754 Chr.2: 27499202-27499204 on Build GRCh38
rs748234481 Chr.2: 27499231-27499231 on Build GRCh38
rs813592 Chr.2: 27499104-27499104 on Build GRCh38
rs943619050 Chr.2: 27499213-27499213 on Build GRCh38
rs762671458 Chr.2: 27499111-27499111 on Build GRCh38
rs745769517 Chr.2: 27499373-27499373 on Build GRCh38
rs768151448 Chr.2: 27499433-27499433 on Build GRCh38
rs879044830 Chr.2: 27499442-27499442 on Build GRCh38
rs976290555 Chr.2: 27499216-27499216 on Build GRCh38
rs139834228 Chr.2: 27499122-27499122 on Build GRCh38
rs777863397 Chr.2: 27499349-27499349 on Build GRCh38
rs759000345 Chr.2: 27499137-27499137 on Build GRCh38
rs374682058 Chr.2: 27499339-27499339 on Build GRCh38
rs766995710 Chr.2: 27499160-27499160 on Build GRCh38
rs776255061 Chr.2: 27499497-27499497 on Build GRCh38
rs151082324 Chr.2: 27499449-27499449 on Build GRCh38
rs763737817 Chr.2: 27499397-27499397 on Build GRCh38
rs996386237 Chr.2: 27499058-27499058 on Build GRCh38
rs976565174 Chr.2: 27499326-27499326 on Build GRCh38
rs766951214 Chr.2: 27499196-27499196 on Build GRCh38
rs760367846 Chr.2: 27499177-27499177 on Build GRCh38
rs557115243 Chr.2: 27499493-27499493 on Build GRCh38
rs1033463558 Chr.2: 27499080-27499080 on Build GRCh38
rs971389143 Chr.2: 27499099-27499099 on Build GRCh38
rs766161729 Chr.2: 27499078-27499078 on Build GRCh38
rs774972223 Chr.2: 27499381-27499381 on Build GRCh38
rs758692756 Chr.2: 27499188-27499188 on Build GRCh38
rs756177740 Chr.2: 27499443-27499443 on Build GRCh38
rs771943495 Chr.2: 27499463-27499463 on Build GRCh38
rs919388814 Chr.2: 27499133-27499133 on Build GRCh38
rs781389240 Chr.2: 27499228-27499228 on Build GRCh38
rs1042442805 Chr.2: 27499249-27499249 on Build GRCh38
rs983391888 Chr.2: 27499318-27499318 on Build GRCh38
rs746587914 Chr.2: 27499224-27499224 on Build GRCh38
rs745724467 Chr.2: 27499459-27499459 on Build GRCh38
rs1024883004 Chr.2: 27499362-27499362 on Build GRCh38
rs777999797 Chr.2: 27499444-27499444 on Build GRCh38
rs763329011 Chr.2: 27499406-27499406 on Build GRCh38
rs778670984 Chr.2: 27499456-27499456 on Build GRCh38
rs755590397 Chr.2: 27499209-27499209 on Build GRCh38
rs558646703 Chr.2: 27499268-27499268 on Build GRCh38
rs537136458 Chr.2: 27499259-27499259 on Build GRCh38
rs574660039 Chr.2: 27499390-27499390 on Build GRCh38
rs774306506 Chr.2: 27499134-27499134 on Build GRCh38
rs139153177 Chr.2: 27499411-27499411 on Build GRCh38
rs768048286 Chr.2: 27499445-27499446 on Build GRCh38
rs945093972 Chr.2: 27499239-27499239 on Build GRCh38
rs1010548025 Chr.2: 27499102-27499102 on Build GRCh38
rs903590942 Chr.2: 27499064-27499064 on Build GRCh38
rs202099374 Chr.2: 27499354-27499354 on Build GRCh38
rs755528355 Chr.2: 27499425-27499425 on Build GRCh38
rs769977399 Chr.2: 27499108-27499108 on Build GRCh38
rs903633232 Chr.2: 27499277-27499277 on Build GRCh38
rs185430509 Chr.2: 27499313-27499313 on Build GRCh38
rs952175724 Chr.2: 27499462-27499462 on Build GRCh38
rs951975316 Chr.2: 27499312-27499312 on Build GRCh38
rs8179208 Chr.2: 27499320-27499320 on Build GRCh38
rs1013433945 Chr.2: 27499090-27499090 on Build GRCh38
rs772068432 Chr.2: 27499378-27499378 on Build GRCh38
rs773047068 Chr.2: 27499109-27499109 on Build GRCh38
rs748380298 Chr.2: 27499105-27499105 on Build GRCh38
rs752249072 Chr.2: 27499173-27499173 on Build GRCh38
rs1042101999 Chr.2: 27499039-27499039 on Build GRCh38
rs547476053 Chr.2: 27499086-27499086 on Build GRCh38
rs8179207 Chr.2: 27499187-27499187 on Build GRCh38
rs189485742 Chr.2: 27499405-27499405 on Build GRCh38
rs752011709 Chr.2: 27499424-27499424 on Build GRCh38
rs774177724 Chr.2: 27499370-27499370 on Build GRCh38
rs936391849 Chr.2: 27499301-27499301 on Build GRCh38

View Full Product Details