Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 33054919-33055472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KL
Entrez Gene ID
9365
Gene Name
klotho
Gene Aliases
-
UniGene
Hs.524953
Gene Chromosome Location
Chr.13: 33016063-33066145 on Build GRCh38
Transcripts
NM_004795.3 XM_006719895.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3468450 c.1388G>A p.G463D Chr.13: 33055104-33055104 on Build GRCh38
6198110 c.616C>A p.Q206K Chr.13: 33055253-33055253 on Build GRCh38
185679 c.1392C>T p.F464F Chr.13: 33055108-33055108 on Build GRCh38
6074219 c.1444C>T p.L482L Chr.13: 33055160-33055160 on Build GRCh38
3468454 c.1576G>A p.G526R Chr.13: 33055292-33055292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770054152 Chr.13: 33055005-33055005 on Build GRCh38
rs267603803 Chr.13: 33055274-33055274 on Build GRCh38
rs372711699 Chr.13: 33055150-33055150 on Build GRCh38
rs373674206 Chr.13: 33055331-33055331 on Build GRCh38
rs761748531 Chr.13: 33055243-33055243 on Build GRCh38
rs371650258 Chr.13: 33055310-33055310 on Build GRCh38
rs924120761 Chr.13: 33055000-33055000 on Build GRCh38
rs761695593 Chr.13: 33055399-33055399 on Build GRCh38
rs776038884 Chr.13: 33055080-33055080 on Build GRCh38
rs752846304 Chr.13: 33055308-33055308 on Build GRCh38
rs767340507 Chr.13: 33055432-33055432 on Build GRCh38
rs762838530 Chr.13: 33055090-33055090 on Build GRCh38
rs771960861 Chr.13: 33055402-33055402 on Build GRCh38
rs544984451 Chr.13: 33055007-33055007 on Build GRCh38
rs367577680 Chr.13: 33055344-33055344 on Build GRCh38
rs145255620 Chr.13: 33055180-33055180 on Build GRCh38
rs138916294 Chr.13: 33055123-33055123 on Build GRCh38
rs779716626 Chr.13: 33055364-33055364 on Build GRCh38
rs143526946 Chr.13: 33055085-33055085 on Build GRCh38
rs536288517 Chr.13: 33055430-33055430 on Build GRCh38
rs758701481 Chr.13: 33055136-33055136 on Build GRCh38
rs769417229 Chr.13: 33055151-33055151 on Build GRCh38
rs772960733 Chr.13: 33055406-33055406 on Build GRCh38
rs62637617 Chr.13: 33055309-33055309 on Build GRCh38
rs751485574 Chr.13: 33055108-33055108 on Build GRCh38
rs916797636 Chr.13: 33055405-33055405 on Build GRCh38
rs77557976 Chr.13: 33055417-33055417 on Build GRCh38
rs778812497 Chr.13: 33055322-33055322 on Build GRCh38
rs773691984 Chr.13: 33055054-33055054 on Build GRCh38
rs973052241 Chr.13: 33055345-33055345 on Build GRCh38
rs757683841 Chr.13: 33055327-33055327 on Build GRCh38
rs769129498 Chr.13: 33055350-33055350 on Build GRCh38
rs747524710 Chr.13: 33055069-33055069 on Build GRCh38
rs3752472 Chr.13: 33055256-33055256 on Build GRCh38
rs144181834 Chr.13: 33055135-33055135 on Build GRCh38
rs766430740 Chr.13: 33055119-33055119 on Build GRCh38
rs868138955 Chr.13: 33055226-33055226 on Build GRCh38
rs778998621 Chr.13: 33055134-33055134 on Build GRCh38
rs1031858876 Chr.13: 33055269-33055269 on Build GRCh38
rs369262610 Chr.13: 33055109-33055109 on Build GRCh38
rs751901530 Chr.13: 33055324-33055324 on Build GRCh38
rs376035130 Chr.13: 33055205-33055205 on Build GRCh38
rs978233222 Chr.13: 33054999-33054999 on Build GRCh38
rs886050119 Chr.13: 33055329-33055329 on Build GRCh38
rs987516871 Chr.13: 33055031-33055031 on Build GRCh38
rs749483987 Chr.13: 33055020-33055020 on Build GRCh38
rs377180541 Chr.13: 33055015-33055015 on Build GRCh38
rs765369493 Chr.13: 33055305-33055305 on Build GRCh38
rs751614510 Chr.13: 33055013-33055013 on Build GRCh38
rs747471621 Chr.13: 33055137-33055137 on Build GRCh38
rs199758453 Chr.13: 33055008-33055008 on Build GRCh38
rs150268586 Chr.13: 33055087-33055087 on Build GRCh38
rs906413250 Chr.13: 33055084-33055084 on Build GRCh38
rs1035707821 Chr.13: 33054949-33054952 on Build GRCh38
rs765044587 Chr.13: 33055006-33055006 on Build GRCh38
rs1057436484 Chr.13: 33055097-33055097 on Build GRCh38
rs768399054 Chr.13: 33055386-33055386 on Build GRCh38
rs765187533 Chr.13: 33055412-33055412 on Build GRCh38
rs920320354 Chr.13: 33055346-33055346 on Build GRCh38
rs559442255 Chr.13: 33054959-33054959 on Build GRCh38
rs201245224 Chr.13: 33055050-33055050 on Build GRCh38
rs372540823 Chr.13: 33055075-33055075 on Build GRCh38
rs764096679 Chr.13: 33055012-33055012 on Build GRCh38
rs930001829 Chr.13: 33055445-33055445 on Build GRCh38
rs764039600 Chr.13: 33055098-33055098 on Build GRCh38
rs759516810 Chr.13: 33055292-33055292 on Build GRCh38
rs756494516 Chr.13: 33054961-33054961 on Build GRCh38
rs755125157 Chr.13: 33055127-33055127 on Build GRCh38
rs948043681 Chr.13: 33055021-33055021 on Build GRCh38
rs750440983 Chr.13: 33055273-33055273 on Build GRCh38
rs967122817 Chr.13: 33054971-33054971 on Build GRCh38
rs958221621 Chr.13: 33055335-33055335 on Build GRCh38
rs750225113 Chr.13: 33055105-33055105 on Build GRCh38
rs774186371 Chr.13: 33055397-33055397 on Build GRCh38
rs771453117 Chr.13: 33055141-33055141 on Build GRCh38
rs762709016 Chr.13: 33055218-33055218 on Build GRCh38
rs577818913 Chr.13: 33054998-33054998 on Build GRCh38
rs777919048 Chr.13: 33055176-33055178 on Build GRCh38
rs774307767 Chr.13: 33055222-33055222 on Build GRCh38
rs768500981 Chr.13: 33055219-33055219 on Build GRCh38
rs779243484 Chr.13: 33055043-33055043 on Build GRCh38
rs142012227 Chr.13: 33055038-33055038 on Build GRCh38
rs748430461 Chr.13: 33055044-33055044 on Build GRCh38
rs748911384 Chr.13: 33055372-33055372 on Build GRCh38
rs760662427 Chr.13: 33055411-33055411 on Build GRCh38
rs922313744 Chr.13: 33054970-33054970 on Build GRCh38
rs370182085 Chr.13: 33055016-33055016 on Build GRCh38
rs1013023907 Chr.13: 33054989-33054989 on Build GRCh38
rs929101956 Chr.13: 33055076-33055076 on Build GRCh38
rs1046383482 Chr.13: 33055029-33055029 on Build GRCh38
rs538764483 Chr.13: 33055149-33055149 on Build GRCh38
rs781749095 Chr.13: 33055144-33055144 on Build GRCh38

View Full Product Details