Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50943361-50943910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLK5
Entrez Gene ID
25818
Gene Name
kallikrein related peptidase 5
Gene Aliases
KLK-L2, KLKL2, SCTE
UniGene
Hs.50915
Gene Chromosome Location
Chr.19: 50943299-50953088 on Build GRCh38
Transcripts
NM_001077491.1 NM_001077492.1 NM_012427.4 XM_011526702.1 XM_011526703.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3389255 c.758G>C p.G253A Chr.19: 50943755-50943755 on Build GRCh38
4999847 c.785G>A p.G262E Chr.19: 50943728-50943728 on Build GRCh38
1740470 c.727G>T p.G243C Chr.19: 50943786-50943786 on Build GRCh38
3414094 c.881G>C p.*294S Chr.19: 50943632-50943632 on Build GRCh38
192333 c.800C>T p.A267V Chr.19: 50943713-50943713 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139575737 Chr.19: 50943672-50943672 on Build GRCh38
rs781648641 Chr.19: 50943772-50943772 on Build GRCh38
rs762297287 Chr.19: 50943711-50943711 on Build GRCh38
rs550878265 Chr.19: 50943729-50943729 on Build GRCh38
rs62115182 Chr.19: 50943441-50943441 on Build GRCh38
rs771697729 Chr.19: 50943794-50943794 on Build GRCh38
rs759297010 Chr.19: 50943613-50943613 on Build GRCh38
rs960616461 Chr.19: 50943575-50943575 on Build GRCh38
rs767233157 Chr.19: 50943617-50943617 on Build GRCh38
rs376478781 Chr.19: 50943747-50943747 on Build GRCh38
rs778966720 Chr.19: 50943786-50943786 on Build GRCh38
rs574761210 Chr.19: 50943473-50943473 on Build GRCh38
rs369872276 Chr.19: 50943638-50943638 on Build GRCh38
rs200795065 Chr.19: 50943752-50943752 on Build GRCh38
rs1051692387 Chr.19: 50943822-50943822 on Build GRCh38
rs528292923 Chr.19: 50943486-50943486 on Build GRCh38
rs759975234 Chr.19: 50943834-50943834 on Build GRCh38
rs142508860 Chr.19: 50943483-50943483 on Build GRCh38
rs773015791 Chr.19: 50943596-50943596 on Build GRCh38
rs773560373 Chr.19: 50943737-50943737 on Build GRCh38
rs933538540 Chr.19: 50943479-50943479 on Build GRCh38
rs778500379 Chr.19: 50943801-50943801 on Build GRCh38
rs2659092 Chr.19: 50943404-50943404 on Build GRCh38
rs1030805781 Chr.19: 50943565-50943565 on Build GRCh38
rs771493844 Chr.19: 50943810-50943810 on Build GRCh38
rs759248677 Chr.19: 50943437-50943437 on Build GRCh38
rs182328761 Chr.19: 50943477-50943477 on Build GRCh38
rs766848629 Chr.19: 50943739-50943739 on Build GRCh38
rs748039264 Chr.19: 50943677-50943677 on Build GRCh38
rs752873684 Chr.19: 50943767-50943767 on Build GRCh38
rs757316544 Chr.19: 50943784-50943784 on Build GRCh38
rs374348589 Chr.19: 50943646-50943646 on Build GRCh38
rs745352150 Chr.19: 50943700-50943700 on Build GRCh38
rs560825538 Chr.19: 50943386-50943386 on Build GRCh38
rs753623477 Chr.19: 50943777-50943777 on Build GRCh38
rs532213513 Chr.19: 50943649-50943649 on Build GRCh38
rs371210292 Chr.19: 50943591-50943591 on Build GRCh38
rs990986894 Chr.19: 50943771-50943771 on Build GRCh38
rs973437559 Chr.19: 50943474-50943475 on Build GRCh38
rs138001786 Chr.19: 50943827-50943827 on Build GRCh38
rs187240680 Chr.19: 50943517-50943517 on Build GRCh38
rs371401715 Chr.19: 50943606-50943606 on Build GRCh38
rs765533748 Chr.19: 50943725-50943725 on Build GRCh38
rs1021488818 Chr.19: 50943409-50943409 on Build GRCh38
rs777456488 Chr.19: 50943772-50943772 on Build GRCh38
rs147723778 Chr.19: 50943468-50943468 on Build GRCh38
rs1042726041 Chr.19: 50943594-50943594 on Build GRCh38
rs985253579 Chr.19: 50943449-50943449 on Build GRCh38
rs750881474 Chr.19: 50943640-50943640 on Build GRCh38
rs568810143 Chr.19: 50943686-50943686 on Build GRCh38
rs889425085 Chr.19: 50943487-50943487 on Build GRCh38
rs940839398 Chr.19: 50943456-50943456 on Build GRCh38
rs894112500 Chr.19: 50943581-50943581 on Build GRCh38
rs1701949 Chr.19: 50943809-50943809 on Build GRCh38
rs769808746 Chr.19: 50943586-50943586 on Build GRCh38
rs745440032 Chr.19: 50943792-50943792 on Build GRCh38
rs369452061 Chr.19: 50943699-50943699 on Build GRCh38
rs754521981 Chr.19: 50943798-50943798 on Build GRCh38
rs550428618 Chr.19: 50943665-50943665 on Build GRCh38
rs375497016 Chr.19: 50943608-50943608 on Build GRCh38
rs2232539 Chr.19: 50943607-50943607 on Build GRCh38
rs372740238 Chr.19: 50943805-50943805 on Build GRCh38
rs545837890 Chr.19: 50943450-50943450 on Build GRCh38
rs999075586 Chr.19: 50943414-50943414 on Build GRCh38
rs529687425 Chr.19: 50943693-50943693 on Build GRCh38
rs779559144 Chr.19: 50943803-50943803 on Build GRCh38
rs377276035 Chr.19: 50943697-50943697 on Build GRCh38
rs756289545 Chr.19: 50943768-50943768 on Build GRCh38
rs777187698 Chr.19: 50943623-50943623 on Build GRCh38
rs999759118 Chr.19: 50943531-50943531 on Build GRCh38
rs373660204 Chr.19: 50943738-50943738 on Build GRCh38
rs146723372 Chr.19: 50943710-50943710 on Build GRCh38
rs1013486895 Chr.19: 50943590-50943590 on Build GRCh38
rs569112989 Chr.19: 50943756-50943756 on Build GRCh38
rs751590358 Chr.19: 50943654-50943654 on Build GRCh38
rs753107705 Chr.19: 50943518-50943518 on Build GRCh38
rs903708378 Chr.19: 50943528-50943528 on Build GRCh38
rs532474224 Chr.19: 50943577-50943577 on Build GRCh38
rs768305278 Chr.19: 50943806-50943806 on Build GRCh38
rs558620755 Chr.19: 50943389-50943389 on Build GRCh38
rs759781166 Chr.19: 50943771-50943772 on Build GRCh38
rs910515032 Chr.19: 50943462-50943462 on Build GRCh38
rs369743279 Chr.19: 50943837-50943837 on Build GRCh38
rs61729816 Chr.19: 50943714-50943714 on Build GRCh38
rs1017473606 Chr.19: 50943853-50943853 on Build GRCh38
rs757672704 Chr.19: 50943591-50943592 on Build GRCh38
rs199972562 Chr.19: 50943685-50943685 on Build GRCh38
rs1007392133 Chr.19: 50943839-50943839 on Build GRCh38
rs771894142 Chr.19: 50943703-50943703 on Build GRCh38
rs570725805 Chr.19: 50943871-50943871 on Build GRCh38
rs982677643 Chr.19: 50943510-50943510 on Build GRCh38
rs762054760 Chr.19: 50943627-50943627 on Build GRCh38
rs149057523 Chr.19: 50943883-50943883 on Build GRCh38
rs938407455 Chr.19: 50943526-50943526 on Build GRCh38
rs767782509 Chr.19: 50943757-50943757 on Build GRCh38

View Full Product Details