Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 64917500-64917897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF273
Entrez Gene ID
10793
Gene Name
zinc finger protein 273
Gene Aliases
HZF9
UniGene
Hs.520889
Gene Chromosome Location
Chr.7: 64882493-64937316 on Build GRCh38
Transcripts
NM_021148.2 NR_003099.1 XM_011515746.2 XM_017011688.1 XM_017011689.1 XM_017011690.1 XM_017011691.1 XM_017011692.1 XM_017011693.1 XM_017011694.1 XM_017011695.1 XM_017011696.1 XM_017011697.1 XM_017011698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4153058 c.130A>C p.I44L Chr.7: 64917608-64917608 on Build GRCh38
3085293 c.188G>C p.R63T Chr.7: 64917666-64917666 on Build GRCh38
5976450 c.206A>G p.N69S Chr.7: 64917684-64917684 on Build GRCh38
5014692 c.107C>A p.P36Q Chr.7: 64917585-64917585 on Build GRCh38
1233791 c.121G>T p.D41Y Chr.7: 64917599-64917599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17850314 Chr.7: 64917611-64917611 on Build GRCh38
rs769085143 Chr.7: 64917638-64917638 on Build GRCh38
rs934639580 Chr.7: 64917710-64917710 on Build GRCh38
rs202202845 Chr.7: 64917552-64917552 on Build GRCh38
rs748932461 Chr.7: 64917668-64917668 on Build GRCh38
rs766023603 Chr.7: 64917747-64917749 on Build GRCh38
rs1005100268 Chr.7: 64917737-64917737 on Build GRCh38
rs767460744 Chr.7: 64917564-64917565 on Build GRCh38
rs760624942 Chr.7: 64917758-64917758 on Build GRCh38
rs373816678 Chr.7: 64917620-64917620 on Build GRCh38
rs770531951 Chr.7: 64917702-64917702 on Build GRCh38
rs749951489 Chr.7: 64917608-64917608 on Build GRCh38
rs760173208 Chr.7: 64917704-64917705 on Build GRCh38
rs773893508 Chr.7: 64917727-64917727 on Build GRCh38
rs1028686587 Chr.7: 64917801-64917801 on Build GRCh38
rs924630292 Chr.7: 64917704-64917704 on Build GRCh38
rs765864989 Chr.7: 64917560-64917560 on Build GRCh38
rs775344795 Chr.7: 64917752-64917752 on Build GRCh38
rs758958915 Chr.7: 64917602-64917602 on Build GRCh38
rs759489441 Chr.7: 64917566-64917566 on Build GRCh38
rs531384434 Chr.7: 64917581-64917581 on Build GRCh38
rs997613184 Chr.7: 64917771-64917771 on Build GRCh38
rs200674814 Chr.7: 64917722-64917722 on Build GRCh38
rs185063511 Chr.7: 64917548-64917548 on Build GRCh38
rs759108313 Chr.7: 64917736-64917736 on Build GRCh38
rs780773605 Chr.7: 64917612-64917612 on Build GRCh38
rs562295144 Chr.7: 64917855-64917855 on Build GRCh38
rs571556570 Chr.7: 64917562-64917562 on Build GRCh38
rs774368599 Chr.7: 64917561-64917562 on Build GRCh38
rs767441493 Chr.7: 64917745-64917745 on Build GRCh38
rs755836426 Chr.7: 64917570-64917570 on Build GRCh38
rs769566150 Chr.7: 64917544-64917544 on Build GRCh38
rs986814274 Chr.7: 64917608-64917608 on Build GRCh38
rs948975761 Chr.7: 64917861-64917861 on Build GRCh38
rs893419352 Chr.7: 64917842-64917842 on Build GRCh38
rs747523476 Chr.7: 64917635-64917635 on Build GRCh38
rs776882785 Chr.7: 64917646-64917646 on Build GRCh38
rs368719165 Chr.7: 64917592-64917592 on Build GRCh38
rs1049298887 Chr.7: 64917616-64917616 on Build GRCh38
rs761615627 Chr.7: 64917560-64917561 on Build GRCh38
rs745823249 Chr.7: 64917526-64917526 on Build GRCh38
rs201727030 Chr.7: 64917574-64917574 on Build GRCh38
rs752480236 Chr.7: 64917568-64917568 on Build GRCh38
rs1015603949 Chr.7: 64917746-64917746 on Build GRCh38
rs35440582 Chr.7: 64917748-64917749 on Build GRCh38
rs527911325 Chr.7: 64917856-64917856 on Build GRCh38
rs200608022 Chr.7: 64917600-64917600 on Build GRCh38

View Full Product Details