Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 24463761-24464285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM7
Entrez Gene ID
8756
Gene Name
ADAM metallopeptidase domain 7
Gene Aliases
ADAM 7, ADAM-7, EAPI, GP-83, GP83
UniGene
Hs.116147
Gene Chromosome Location
Chr.8: 24440996-24509565 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Gene Symbol
LOC101929294
Entrez Gene ID
101929294
Gene Name
uncharacterized LOC101929294
Gene Aliases
-
UniGene
Hs.637673
Gene Chromosome Location
Chr.8: 24295814-24548618 on Build GRCh38
Transcripts
NM_003817.3 XM_017013943.1 NR_125808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6013096 c.234-5T>G p.? Chr.8: 24463877-24463877 on Build GRCh38
3899058 c.297G>A p.R99R Chr.8: 24463945-24463945 on Build GRCh38
6875506 c.253T>A p.Y85N Chr.8: 24463901-24463901 on Build GRCh38
6875507 c.312G>A p.M104I Chr.8: 24463960-24463960 on Build GRCh38
360416 c.312G>T p.M104I Chr.8: 24463960-24463960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77483512 Chr.8: 24464053-24464053 on Build GRCh38
rs776799762 Chr.8: 24463918-24463918 on Build GRCh38
rs765041281 Chr.8: 24463962-24463962 on Build GRCh38
rs181609484 Chr.8: 24463864-24463864 on Build GRCh38
rs961932827 Chr.8: 24464218-24464218 on Build GRCh38
rs576717055 Chr.8: 24463845-24463845 on Build GRCh38
rs771522387 Chr.8: 24464000-24464000 on Build GRCh38
rs117555143 Chr.8: 24463866-24463866 on Build GRCh38
rs199523762 Chr.8: 24464007-24464007 on Build GRCh38
rs748222279 Chr.8: 24463924-24463924 on Build GRCh38
rs139771485 Chr.8: 24463896-24463896 on Build GRCh38
rs186791873 Chr.8: 24463872-24463872 on Build GRCh38
rs758302865 Chr.8: 24463982-24463982 on Build GRCh38
rs756518198 Chr.8: 24463979-24463980 on Build GRCh38
rs371059047 Chr.8: 24463856-24463856 on Build GRCh38
rs754742522 Chr.8: 24463984-24463984 on Build GRCh38
rs760886626 Chr.8: 24463846-24463846 on Build GRCh38
rs137964591 Chr.8: 24463983-24463983 on Build GRCh38
rs145275948 Chr.8: 24463935-24463935 on Build GRCh38
rs778765479 Chr.8: 24463885-24463885 on Build GRCh38
rs1016566759 Chr.8: 24464217-24464217 on Build GRCh38
rs374550095 Chr.8: 24464120-24464120 on Build GRCh38
rs762407602 Chr.8: 24463861-24463861 on Build GRCh38
rs922734388 Chr.8: 24464050-24464050 on Build GRCh38
rs763449473 Chr.8: 24463936-24463936 on Build GRCh38
rs750336900 Chr.8: 24463975-24463975 on Build GRCh38
rs983787103 Chr.8: 24463979-24463979 on Build GRCh38
rs537426755 Chr.8: 24464177-24464177 on Build GRCh38
rs538314922 Chr.8: 24464228-24464228 on Build GRCh38
rs536854278 Chr.8: 24463996-24463996 on Build GRCh38
rs750886049 Chr.8: 24463978-24463979 on Build GRCh38
rs774806198 Chr.8: 24463958-24463958 on Build GRCh38
rs1018719720 Chr.8: 24464024-24464024 on Build GRCh38
rs776636404 Chr.8: 24463858-24463858 on Build GRCh38
rs936022723 Chr.8: 24464061-24464061 on Build GRCh38
rs150324699 Chr.8: 24463867-24463867 on Build GRCh38
rs934157749 Chr.8: 24464051-24464051 on Build GRCh38
rs200343668 Chr.8: 24463871-24463871 on Build GRCh38
rs973319476 Chr.8: 24464035-24464035 on Build GRCh38
rs779761218 Chr.8: 24463910-24463910 on Build GRCh38
rs376376733 Chr.8: 24463831-24463831 on Build GRCh38
rs372747188 Chr.8: 24463999-24463999 on Build GRCh38
rs781449211 Chr.8: 24463985-24463985 on Build GRCh38
rs913406006 Chr.8: 24463874-24463874 on Build GRCh38
rs552737525 Chr.8: 24464196-24464196 on Build GRCh38
rs764800800 Chr.8: 24463970-24463970 on Build GRCh38
rs781280552 Chr.8: 24464065-24464065 on Build GRCh38
rs911293277 Chr.8: 24464060-24464060 on Build GRCh38
rs766863348 Chr.8: 24463943-24463943 on Build GRCh38
rs966011077 Chr.8: 24464032-24464032 on Build GRCh38
rs546375253 Chr.8: 24463817-24463817 on Build GRCh38
rs200862896 Chr.8: 24463884-24463884 on Build GRCh38
rs779522382 Chr.8: 24463869-24463869 on Build GRCh38
rs1003366716 Chr.8: 24464154-24464154 on Build GRCh38
rs906400524 Chr.8: 24464125-24464125 on Build GRCh38
rs149466364 Chr.8: 24464010-24464010 on Build GRCh38
rs1009830915 Chr.8: 24463916-24463916 on Build GRCh38
rs1042244685 Chr.8: 24464097-24464097 on Build GRCh38
rs756824540 Chr.8: 24463955-24463955 on Build GRCh38
rs758198198 Chr.8: 24463902-24463902 on Build GRCh38
rs745692107 Chr.8: 24463840-24463840 on Build GRCh38
rs577655614 Chr.8: 24464214-24464214 on Build GRCh38
rs753438964 Chr.8: 24463927-24463927 on Build GRCh38
rs747248068 Chr.8: 24463911-24463911 on Build GRCh38
rs767278717 Chr.8: 24463830-24463830 on Build GRCh38
rs748394619 Chr.8: 24463994-24463994 on Build GRCh38
rs959131234 Chr.8: 24464213-24464213 on Build GRCh38
rs763880379 Chr.8: 24463965-24463965 on Build GRCh38
rs768820063 Chr.8: 24463913-24463913 on Build GRCh38
rs937661026 Chr.8: 24463796-24463796 on Build GRCh38
rs1027456632 Chr.8: 24463821-24463821 on Build GRCh38
rs957563443 Chr.8: 24464086-24464086 on Build GRCh38
rs755768463 Chr.8: 24463870-24463870 on Build GRCh38
rs376626403 Chr.8: 24463969-24463969 on Build GRCh38
rs371722057 Chr.8: 24463968-24463968 on Build GRCh38
rs569474979 Chr.8: 24463995-24463995 on Build GRCh38
rs981633096 Chr.8: 24464232-24464232 on Build GRCh38

View Full Product Details