Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123082449-123082963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D31
Entrez Gene ID
93594
Gene Name
TBC1 domain family member 31
Gene Aliases
Gm85, WDR67
UniGene
Hs.492716
Gene Chromosome Location
Chr.8: 123041961-123165169 on Build GRCh38
Transcripts
NM_001145088.1 NM_145647.3 XM_005251102.1 XM_005251103.1 XM_005251104.1 XM_011517378.2 XM_011517379.2 XM_017013983.1 XM_017013984.1 XM_017013985.1 XM_017013986.1 XM_017013987.1 XM_017013988.1 XM_017013989.1 XM_017013990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6549716 c.277C>T p.L93F Chr.8: 123082754-123082754 on Build GRCh38
6549717 c.290C>T p.S97F Chr.8: 123082767-123082767 on Build GRCh38
6842835 c.252A>G p.Q84Q Chr.8: 123082729-123082729 on Build GRCh38
223306 c.245C>T p.T82I Chr.8: 123082722-123082722 on Build GRCh38
4626119 c.338C>T p.T113I Chr.8: 123082815-123082815 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769229128 Chr.8: 123082732-123082732 on Build GRCh38
rs930065458 Chr.8: 123082940-123082940 on Build GRCh38
rs369654384 Chr.8: 123082789-123082789 on Build GRCh38
rs746079162 Chr.8: 123082724-123082724 on Build GRCh38
rs915839723 Chr.8: 123082893-123082893 on Build GRCh38
rs765073590 Chr.8: 123082680-123082680 on Build GRCh38
rs752364951 Chr.8: 123082681-123082681 on Build GRCh38
rs117552523 Chr.8: 123082758-123082758 on Build GRCh38
rs760309449 Chr.8: 123082742-123082742 on Build GRCh38
rs990063707 Chr.8: 123082623-123082623 on Build GRCh38
rs770551081 Chr.8: 123082747-123082747 on Build GRCh38
rs1027145095 Chr.8: 123082550-123082550 on Build GRCh38
rs752361165 Chr.8: 123082766-123082766 on Build GRCh38
rs764683859 Chr.8: 123082677-123082678 on Build GRCh38
rs890826522 Chr.8: 123082475-123082475 on Build GRCh38
rs762313129 Chr.8: 123082506-123082506 on Build GRCh38
rs535319416 Chr.8: 123082617-123082617 on Build GRCh38
rs989928984 Chr.8: 123082537-123082537 on Build GRCh38
rs897417270 Chr.8: 123082488-123082488 on Build GRCh38
rs139956818 Chr.8: 123082607-123082607 on Build GRCh38
rs749558093 Chr.8: 123082731-123082731 on Build GRCh38
rs770125183 Chr.8: 123082729-123082729 on Build GRCh38
rs930055690 Chr.8: 123082676-123082676 on Build GRCh38
rs372744245 Chr.8: 123082706-123082706 on Build GRCh38
rs759335555 Chr.8: 123082485-123082485 on Build GRCh38
rs964025510 Chr.8: 123082503-123082503 on Build GRCh38
rs777101511 Chr.8: 123082477-123082477 on Build GRCh38
rs116501345 Chr.8: 123082908-123082908 on Build GRCh38
rs767857977 Chr.8: 123082689-123082689 on Build GRCh38
rs111584092 Chr.8: 123082632-123082632 on Build GRCh38
rs762612207 Chr.8: 123082767-123082767 on Build GRCh38
rs774967786 Chr.8: 123082740-123082740 on Build GRCh38
rs572624777 Chr.8: 123082609-123082609 on Build GRCh38
rs367584320 Chr.8: 123082734-123082734 on Build GRCh38
rs562263232 Chr.8: 123082828-123082828 on Build GRCh38
rs977290096 Chr.8: 123082590-123082590 on Build GRCh38
rs113739088 Chr.8: 123082593-123082593 on Build GRCh38
rs764069592 Chr.8: 123082911-123082911 on Build GRCh38
rs777519565 Chr.8: 123082691-123082691 on Build GRCh38
rs375672841 Chr.8: 123082719-123082719 on Build GRCh38
rs532093457 Chr.8: 123082592-123082592 on Build GRCh38
rs941957002 Chr.8: 123082899-123082899 on Build GRCh38
rs375186187 Chr.8: 123082757-123082757 on Build GRCh38
rs560194025 Chr.8: 123082881-123082881 on Build GRCh38
rs555328637 Chr.8: 123082624-123082624 on Build GRCh38
rs527519230 Chr.8: 123082779-123082779 on Build GRCh38
rs112412659 Chr.8: 123082683-123082683 on Build GRCh38
rs757300043 Chr.8: 123082707-123082707 on Build GRCh38
rs564730874 Chr.8: 123082581-123082581 on Build GRCh38
rs990096612 Chr.8: 123082927-123082927 on Build GRCh38
rs977132998 Chr.8: 123082793-123082793 on Build GRCh38
rs1012846311 Chr.8: 123082844-123082844 on Build GRCh38
rs991327634 Chr.8: 123082887-123082888 on Build GRCh38
rs62520599 Chr.8: 123082907-123082907 on Build GRCh38
rs574883476 Chr.8: 123082631-123082631 on Build GRCh38
rs150696820 Chr.8: 123082600-123082600 on Build GRCh38
rs995755286 Chr.8: 123082754-123082754 on Build GRCh38
rs751037663 Chr.8: 123082587-123082587 on Build GRCh38
rs959530882 Chr.8: 123082564-123082564 on Build GRCh38
rs991774657 Chr.8: 123082599-123082599 on Build GRCh38
rs751479788 Chr.8: 123082797-123082797 on Build GRCh38
rs971227822 Chr.8: 123082896-123082896 on Build GRCh38
rs558769277 Chr.8: 123082577-123082577 on Build GRCh38
rs780472552 Chr.8: 123082730-123082730 on Build GRCh38
rs1021412767 Chr.8: 123082551-123082551 on Build GRCh38
rs371808668 Chr.8: 123082750-123082750 on Build GRCh38
rs139223176 Chr.8: 123082811-123082811 on Build GRCh38
rs780090721 Chr.8: 123082825-123082825 on Build GRCh38
rs1025100290 Chr.8: 123082874-123082874 on Build GRCh38
rs376046753 Chr.8: 123082679-123082679 on Build GRCh38
rs914506534 Chr.8: 123082942-123082942 on Build GRCh38
rs757389232 Chr.8: 123082806-123082806 on Build GRCh38
rs1008322012 Chr.8: 123082476-123082476 on Build GRCh38
rs756373633 Chr.8: 123082821-123082821 on Build GRCh38
rs184476506 Chr.8: 123082860-123082860 on Build GRCh38

View Full Product Details