Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 118009014-118009411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB11FIP2
Entrez Gene ID
22841
Gene Name
RAB11 family interacting protein 2
Gene Aliases
Rab11-FIP2, nRip11
UniGene
Hs.173656
Gene Chromosome Location
Chr.10: 118004916-118046921 on Build GRCh38
Transcripts
NM_014904.2 XM_005269629.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3434581 c.1401G>C p.G467G Chr.10: 118009196-118009196 on Build GRCh38
6761187 c.1555C>T p.P519S Chr.10: 118009042-118009042 on Build GRCh38
6761192 c.1493T>C p.V498A Chr.10: 118009044-118009044 on Build GRCh38
5679367 c.1336G>T p.A446S Chr.10: 118009201-118009201 on Build GRCh38
6761186 c.1495C>T p.P499S Chr.10: 118009042-118009042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748788392 Chr.10: 118009172-118009172 on Build GRCh38
rs778497765 Chr.10: 118009225-118009225 on Build GRCh38
rs894388473 Chr.10: 118009384-118009384 on Build GRCh38
rs373912288 Chr.10: 118009166-118009166 on Build GRCh38
rs529280892 Chr.10: 118009118-118009118 on Build GRCh38
rs777104709 Chr.10: 118009266-118009266 on Build GRCh38
rs1031516904 Chr.10: 118009181-118009181 on Build GRCh38
rs922678293 Chr.10: 118009085-118009085 on Build GRCh38
rs146055595 Chr.10: 118009160-118009160 on Build GRCh38
rs894887775 Chr.10: 118009299-118009299 on Build GRCh38
rs908668651 Chr.10: 118009036-118009036 on Build GRCh38
rs114502427 Chr.10: 118009372-118009372 on Build GRCh38
rs773589855 Chr.10: 118009257-118009257 on Build GRCh38
rs532633716 Chr.10: 118009304-118009304 on Build GRCh38
rs757062448 Chr.10: 118009135-118009135 on Build GRCh38
rs953076487 Chr.10: 118009335-118009335 on Build GRCh38
rs999594305 Chr.10: 118009179-118009179 on Build GRCh38
rs1005766582 Chr.10: 118009353-118009353 on Build GRCh38
rs756141948 Chr.10: 118009159-118009159 on Build GRCh38
rs576182820 Chr.10: 118009037-118009037 on Build GRCh38
rs529193174 Chr.10: 118009309-118009309 on Build GRCh38
rs35208509 Chr.10: 118009122-118009123 on Build GRCh38
rs1018480573 Chr.10: 118009207-118009207 on Build GRCh38
rs909659459 Chr.10: 118009361-118009361 on Build GRCh38
rs1012251684 Chr.10: 118009104-118009104 on Build GRCh38
rs760990794 Chr.10: 118009259-118009259 on Build GRCh38
rs111700598 Chr.10: 118009173-118009173 on Build GRCh38
rs531523752 Chr.10: 118009363-118009363 on Build GRCh38
rs7923321 Chr.10: 118009040-118009040 on Build GRCh38
rs542858569 Chr.10: 118009314-118009314 on Build GRCh38
rs371077404 Chr.10: 118009144-118009144 on Build GRCh38
rs143170621 Chr.10: 118009327-118009327 on Build GRCh38
rs1042622044 Chr.10: 118009114-118009114 on Build GRCh38
rs560060091 Chr.10: 118009278-118009278 on Build GRCh38
rs946616110 Chr.10: 118009105-118009105 on Build GRCh38
rs77914264 Chr.10: 118009097-118009097 on Build GRCh38
rs138030602 Chr.10: 118009088-118009088 on Build GRCh38
rs149075550 Chr.10: 118009094-118009094 on Build GRCh38
rs774812190 Chr.10: 118009228-118009229 on Build GRCh38
rs200821985 Chr.10: 118009242-118009242 on Build GRCh38
rs1003331373 Chr.10: 118009250-118009250 on Build GRCh38
rs770420785 Chr.10: 118009302-118009302 on Build GRCh38
rs977418501 Chr.10: 118009234-118009234 on Build GRCh38
rs150361845 Chr.10: 118009061-118009061 on Build GRCh38
rs780046089 Chr.10: 118009033-118009033 on Build GRCh38
rs1049728944 Chr.10: 118009096-118009096 on Build GRCh38
rs771471767 Chr.10: 118009260-118009260 on Build GRCh38
rs958187622 Chr.10: 118009191-118009191 on Build GRCh38
rs761528205 Chr.10: 118009192-118009192 on Build GRCh38
rs745406723 Chr.10: 118009226-118009227 on Build GRCh38
rs774057787 Chr.10: 118009142-118009142 on Build GRCh38
rs761185223 Chr.10: 118009075-118009075 on Build GRCh38
rs929647800 Chr.10: 118009334-118009334 on Build GRCh38
rs138292683 Chr.10: 118009106-118009106 on Build GRCh38
rs781135777 Chr.10: 118009139-118009139 on Build GRCh38
rs747842214 Chr.10: 118009246-118009246 on Build GRCh38
rs771657357 Chr.10: 118009255-118009255 on Build GRCh38
rs139031897 Chr.10: 118009196-118009196 on Build GRCh38
rs142779822 Chr.10: 118009113-118009113 on Build GRCh38
rs759588453 Chr.10: 118009269-118009269 on Build GRCh38
rs1056166606 Chr.10: 118009272-118009272 on Build GRCh38
rs765351244 Chr.10: 118009273-118009273 on Build GRCh38

View Full Product Details