Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5643021-5643572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM34
Entrez Gene ID
53840
Gene Name
tripartite motif containing 34
Gene Aliases
IFP1, RNF21
UniGene
Hs.729048
Gene Chromosome Location
Chr.11: 5619944-5644398 on Build GRCh38
Transcripts
NM_001003827.1 NM_021616.5 NM_001003819.3

Gene Symbol
TRIM6-TRIM34
Entrez Gene ID
445372
Gene Name
TRIM6-TRIM34 readthrough
Gene Aliases
IFP1, RNF21, TRIM34
UniGene
Hs.729048
Gene Chromosome Location
Chr.11: 5596635-5644398 on Build GRCh38
Transcripts
NM_001003827.1 NM_021616.5 NM_001003819.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5947169 c.903G>A p.V301V Chr.11: 5643145-5643145 on Build GRCh38
3869525 c.2303C>T p.S768F Chr.11: 5643483-5643483 on Build GRCh38
4736189 c.2381A>G p.E794G Chr.11: 5643561-5643561 on Build GRCh38
300450 c.1045C>T p.H349Y Chr.11: 5643287-5643287 on Build GRCh38
928493 c.1159C>A p.L387I Chr.11: 5643401-5643401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759688780 Chr.11: 5643304-5643304 on Build GRCh38
rs560919939 Chr.11: 5643537-5643537 on Build GRCh38
rs773891933 Chr.11: 5643446-5643446 on Build GRCh38
rs770783100 Chr.11: 5643368-5643368 on Build GRCh38
rs768521194 Chr.11: 5643142-5643142 on Build GRCh38
rs59472811 Chr.11: 5643126-5643127 on Build GRCh38
rs759672818 Chr.11: 5643377-5643377 on Build GRCh38
rs746670724 Chr.11: 5643141-5643141 on Build GRCh38
rs191638141 Chr.11: 5643497-5643497 on Build GRCh38
rs765329235 Chr.11: 5643151-5643151 on Build GRCh38
rs878963986 Chr.11: 5643126-5643126 on Build GRCh38
rs371198821 Chr.11: 5643210-5643210 on Build GRCh38
rs747919367 Chr.11: 5643349-5643349 on Build GRCh38
rs113128572 Chr.11: 5643114-5643115 on Build GRCh38
rs200678141 Chr.11: 5643185-5643185 on Build GRCh38
rs144441856 Chr.11: 5643515-5643515 on Build GRCh38
rs781037595 Chr.11: 5643339-5643339 on Build GRCh38
rs774414692 Chr.11: 5643374-5643374 on Build GRCh38
rs906052422 Chr.11: 5643424-5643424 on Build GRCh38
rs199610670 Chr.11: 5643420-5643420 on Build GRCh38
rs753048481 Chr.11: 5643311-5643311 on Build GRCh38
rs937477168 Chr.11: 5643429-5643429 on Build GRCh38
rs770632519 Chr.11: 5643126-5643126 on Build GRCh38
rs757717918 Chr.11: 5643333-5643333 on Build GRCh38
rs990617386 Chr.11: 5643256-5643256 on Build GRCh38
rs1045895496 Chr.11: 5643104-5643104 on Build GRCh38
rs138080438 Chr.11: 5643392-5643392 on Build GRCh38
rs766931097 Chr.11: 5643116-5643117 on Build GRCh38
rs148270580 Chr.11: 5643384-5643384 on Build GRCh38
rs746601799 Chr.11: 5643236-5643236 on Build GRCh38
rs142937651 Chr.11: 5643499-5643499 on Build GRCh38
rs371170379 Chr.11: 5643148-5643148 on Build GRCh38
rs764396132 Chr.11: 5643322-5643322 on Build GRCh38
rs751178489 Chr.11: 5643335-5643335 on Build GRCh38
rs200448609 Chr.11: 5643129-5643129 on Build GRCh38
rs961702285 Chr.11: 5643126-5643127 on Build GRCh38
rs143652980 Chr.11: 5643442-5643442 on Build GRCh38
rs374386649 Chr.11: 5643128-5643128 on Build GRCh38
rs761765281 Chr.11: 5643468-5643468 on Build GRCh38
rs773148877 Chr.11: 5643158-5643158 on Build GRCh38
rs201442240 Chr.11: 5643131-5643131 on Build GRCh38
rs956838203 Chr.11: 5643353-5643353 on Build GRCh38
rs745849246 Chr.11: 5643126-5643128 on Build GRCh38
rs760759282 Chr.11: 5643536-5643536 on Build GRCh38
rs745336901 Chr.11: 5643229-5643229 on Build GRCh38
rs765639607 Chr.11: 5643136-5643137 on Build GRCh38
rs755379162 Chr.11: 5643112-5643112 on Build GRCh38
rs766482956 Chr.11: 5643166-5643166 on Build GRCh38
rs754321193 Chr.11: 5643329-5643329 on Build GRCh38
rs751851026 Chr.11: 5643110-5643110 on Build GRCh38
rs953910519 Chr.11: 5643524-5643524 on Build GRCh38
rs751768911 Chr.11: 5643181-5643181 on Build GRCh38
rs201419504 Chr.11: 5643124-5643124 on Build GRCh38
rs776559981 Chr.11: 5643252-5643252 on Build GRCh38
rs753065007 Chr.11: 5643202-5643202 on Build GRCh38
rs998366704 Chr.11: 5643065-5643065 on Build GRCh38
rs780882235 Chr.11: 5643417-5643417 on Build GRCh38
rs143229169 Chr.11: 5643230-5643230 on Build GRCh38
rs954278150 Chr.11: 5643091-5643091 on Build GRCh38
rs767811505 Chr.11: 5643196-5643196 on Build GRCh38
rs972737789 Chr.11: 5643205-5643205 on Build GRCh38
rs779451503 Chr.11: 5643232-5643232 on Build GRCh38
rs533217366 Chr.11: 5643053-5643053 on Build GRCh38
rs866726342 Chr.11: 5643123-5643123 on Build GRCh38
rs754443193 Chr.11: 5643123-5643124 on Build GRCh38
rs777452380 Chr.11: 5643357-5643357 on Build GRCh38
rs140596118 Chr.11: 5643459-5643459 on Build GRCh38
rs774312326 Chr.11: 5643296-5643296 on Build GRCh38
rs1014874334 Chr.11: 5643094-5643094 on Build GRCh38
rs1015355628 Chr.11: 5643178-5643178 on Build GRCh38
rs777273270 Chr.11: 5643127-5643128 on Build GRCh38
rs936004267 Chr.11: 5643370-5643370 on Build GRCh38
rs141466735 Chr.11: 5643331-5643331 on Build GRCh38
rs756609869 Chr.11: 5643213-5643213 on Build GRCh38
rs768423512 Chr.11: 5643237-5643237 on Build GRCh38
rs754661462 Chr.11: 5643338-5643338 on Build GRCh38
rs148404455 Chr.11: 5643516-5643516 on Build GRCh38
rs749108201 Chr.11: 5643437-5643437 on Build GRCh38
rs759947660 Chr.11: 5643124-5643125 on Build GRCh38
rs778404270 Chr.11: 5643218-5643218 on Build GRCh38
rs566884877 Chr.11: 5643125-5643125 on Build GRCh38
rs777040814 Chr.11: 5643469-5643469 on Build GRCh38
rs763578937 Chr.11: 5643505-5643505 on Build GRCh38
rs867745841 Chr.11: 5643533-5643533 on Build GRCh38
rs755735525 Chr.11: 5643423-5643423 on Build GRCh38
rs772880134 Chr.11: 5643123-5643123 on Build GRCh38
rs779549131 Chr.11: 5643334-5643334 on Build GRCh38
rs765720504 Chr.11: 5643401-5643401 on Build GRCh38
rs767079968 Chr.11: 5643514-5643514 on Build GRCh38
rs202235864 Chr.11: 5643127-5643127 on Build GRCh38
rs1028460435 Chr.11: 5643086-5643086 on Build GRCh38
rs761078882 Chr.11: 5643312-5643312 on Build GRCh38
rs768760349 Chr.11: 5643444-5643444 on Build GRCh38
rs753118424 Chr.11: 5643441-5643442 on Build GRCh38
rs201334864 Chr.11: 5643273-5643273 on Build GRCh38
rs773243395 Chr.11: 5643276-5643276 on Build GRCh38
rs775246778 Chr.11: 5643137-5643137 on Build GRCh38
rs375812190 Chr.11: 5643383-5643383 on Build GRCh38
rs148309490 Chr.11: 5643242-5643242 on Build GRCh38
rs377017683 Chr.11: 5643165-5643165 on Build GRCh38
rs751090808 Chr.11: 5643407-5643407 on Build GRCh38
rs549036249 Chr.11: 5643285-5643285 on Build GRCh38
rs977019471 Chr.11: 5643264-5643264 on Build GRCh38
rs754573804 Chr.11: 5643415-5643415 on Build GRCh38
rs776580619 Chr.11: 5643143-5643143 on Build GRCh38
rs748011819 Chr.11: 5643268-5643268 on Build GRCh38
rs75158171 Chr.11: 5643393-5643393 on Build GRCh38
rs770998237 Chr.11: 5643286-5643286 on Build GRCh38
rs973154393 Chr.11: 5643457-5643457 on Build GRCh38
rs376536851 Chr.11: 5643114-5643114 on Build GRCh38
rs755219648 Chr.11: 5643192-5643192 on Build GRCh38
rs551729741 Chr.11: 5643080-5643080 on Build GRCh38
rs771612989 Chr.11: 5643136-5643136 on Build GRCh38
rs764457321 Chr.11: 5643388-5643388 on Build GRCh38
rs751693225 Chr.11: 5643125-5643126 on Build GRCh38
rs757045852 Chr.11: 5643540-5643540 on Build GRCh38
rs767794973 Chr.11: 5643305-5643305 on Build GRCh38
rs769593188 Chr.11: 5643356-5643356 on Build GRCh38
rs16933844 Chr.11: 5643454-5643454 on Build GRCh38
rs767710216 Chr.11: 5643500-5643500 on Build GRCh38
rs777557214 Chr.11: 5643436-5643436 on Build GRCh38
rs778699709 Chr.11: 5643542-5643542 on Build GRCh38
rs770892020 Chr.11: 5643441-5643441 on Build GRCh38
rs749192409 Chr.11: 5643363-5643363 on Build GRCh38

View Full Product Details