Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85980120-85980692 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAT4C
Entrez Gene ID
25834
Gene Name
MGAT4 family member C
Gene Aliases
GNTIVH, HGNT-IV-H
UniGene
Hs.589093
Gene Chromosome Location
Chr.12: 85972281-86838999 on Build GRCh38
Transcripts
NM_013244.3 XM_005268776.4 XM_005268779.3 XM_011538150.2 XM_011538151.2 XM_011538156.2 XM_011538157.2 XM_017019143.1 XM_017019144.1 XM_017019145.1 XM_017019146.1 XM_017019147.1 XM_017019148.1 XM_017019149.1 XM_017019150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7294704 c.445C>A p.R149S Chr.12: 85980281-85980281 on Build GRCh38
1994615 c.310G>C p.G104R Chr.12: 85980416-85980416 on Build GRCh38
5890714 c.494C>T p.S165L Chr.12: 85980319-85980319 on Build GRCh38
1364663 c.329G>A p.R110Q Chr.12: 85980397-85980397 on Build GRCh38
4783931 c.416G>A p.R139Q Chr.12: 85980397-85980397 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139777686 Chr.12: 85980398-85980398 on Build GRCh38
rs777587242 Chr.12: 85980402-85980402 on Build GRCh38
rs757539324 Chr.12: 85980240-85980240 on Build GRCh38
rs763067361 Chr.12: 85980372-85980372 on Build GRCh38
rs566043053 Chr.12: 85980223-85980223 on Build GRCh38
rs868789272 Chr.12: 85980319-85980319 on Build GRCh38
rs1021871008 Chr.12: 85980648-85980648 on Build GRCh38
rs765429825 Chr.12: 85980243-85980243 on Build GRCh38
rs747318851 Chr.12: 85980476-85980476 on Build GRCh38
rs1051224249 Chr.12: 85980536-85980536 on Build GRCh38
rs1031482564 Chr.12: 85980335-85980335 on Build GRCh38
rs769004377 Chr.12: 85980482-85980482 on Build GRCh38
rs561866218 Chr.12: 85980513-85980513 on Build GRCh38
rs755975066 Chr.12: 85980271-85980271 on Build GRCh38
rs150545721 Chr.12: 85980166-85980166 on Build GRCh38
rs753826277 Chr.12: 85980222-85980222 on Build GRCh38
rs142839367 Chr.12: 85980270-85980270 on Build GRCh38
rs759994326 Chr.12: 85980152-85980152 on Build GRCh38
rs147418986 Chr.12: 85980397-85980397 on Build GRCh38
rs777409272 Chr.12: 85980274-85980274 on Build GRCh38
rs141672136 Chr.12: 85980157-85980157 on Build GRCh38
rs765264406 Chr.12: 85980368-85980368 on Build GRCh38
rs373313230 Chr.12: 85980433-85980433 on Build GRCh38
rs748169936 Chr.12: 85980260-85980260 on Build GRCh38
rs933091433 Chr.12: 85980510-85980510 on Build GRCh38
rs771588801 Chr.12: 85980452-85980452 on Build GRCh38
rs1007149128 Chr.12: 85980245-85980245 on Build GRCh38
rs761892291 Chr.12: 85980363-85980363 on Build GRCh38
rs745564493 Chr.12: 85980450-85980450 on Build GRCh38
rs185118551 Chr.12: 85980339-85980339 on Build GRCh38
rs748796788 Chr.12: 85980280-85980280 on Build GRCh38
rs767389242 Chr.12: 85980376-85980376 on Build GRCh38
rs940530815 Chr.12: 85980394-85980394 on Build GRCh38
rs761281614 Chr.12: 85980332-85980332 on Build GRCh38
rs750669561 Chr.12: 85980370-85980370 on Build GRCh38
rs754801625 Chr.12: 85980252-85980252 on Build GRCh38
rs776093722 Chr.12: 85980159-85980159 on Build GRCh38
rs774149822 Chr.12: 85980300-85980300 on Build GRCh38
rs980691500 Chr.12: 85980387-85980387 on Build GRCh38
rs1002920526 Chr.12: 85980551-85980551 on Build GRCh38
rs190762233 Chr.12: 85980410-85980410 on Build GRCh38
rs1036192252 Chr.12: 85980552-85980552 on Build GRCh38
rs566418071 Chr.12: 85980281-85980281 on Build GRCh38
rs113387514 Chr.12: 85980216-85980216 on Build GRCh38
rs1010843205 Chr.12: 85980638-85980638 on Build GRCh38
rs1007139183 Chr.12: 85980601-85980601 on Build GRCh38
rs918972844 Chr.12: 85980458-85980458 on Build GRCh38
rs756857554 Chr.12: 85980426-85980426 on Build GRCh38
rs764279389 Chr.12: 85980198-85980198 on Build GRCh38
rs140499591 Chr.12: 85980244-85980244 on Build GRCh38
rs539968414 Chr.12: 85980305-85980305 on Build GRCh38
rs780529746 Chr.12: 85980453-85980453 on Build GRCh38
rs752431991 Chr.12: 85980395-85980395 on Build GRCh38
rs891337139 Chr.12: 85980561-85980561 on Build GRCh38
rs995456136 Chr.12: 85980434-85980434 on Build GRCh38
rs956205255 Chr.12: 85980637-85980637 on Build GRCh38
rs780805071 Chr.12: 85980255-85980255 on Build GRCh38
rs776746849 Chr.12: 85980359-85980359 on Build GRCh38
rs1018284129 Chr.12: 85980273-85980273 on Build GRCh38
rs991920430 Chr.12: 85980340-85980340 on Build GRCh38
rs969275266 Chr.12: 85980381-85980381 on Build GRCh38
rs536944591 Chr.12: 85980308-85980308 on Build GRCh38
rs775693691 Chr.12: 85980330-85980330 on Build GRCh38

View Full Product Details