Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 99850678-99851265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGL
Entrez Gene ID
178
Gene Name
amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase
Gene Aliases
GDE
UniGene
Hs.904
Gene Chromosome Location
Chr.1: 99850077-99924023 on Build GRCh38
Transcripts
NM_000028.2 NM_000642.2 NM_000643.2 NM_000644.2 NM_000646.2 XM_005270557.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1952904 c.13A>G p.K5E Chr.1: 99851055-99851055 on Build GRCh38
6120371 c.50A>T p.K17I Chr.1: 99851092-99851092 on Build GRCh38
3470266 c.76G>A p.E26K Chr.1: 99851118-99851118 on Build GRCh38
892885 c.18G>T p.Q6H Chr.1: 99851060-99851060 on Build GRCh38
6596254 c.50A>G p.K17R Chr.1: 99851092-99851092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1049763653 Chr.1: 99850788-99850788 on Build GRCh38
rs765098686 Chr.1: 99851128-99851128 on Build GRCh38
rs909738818 Chr.1: 99850705-99850705 on Build GRCh38
rs749411087 Chr.1: 99851082-99851082 on Build GRCh38
rs770080270 Chr.1: 99851236-99851236 on Build GRCh38
rs765881303 Chr.1: 99851116-99851116 on Build GRCh38
rs191023853 Chr.1: 99850973-99850973 on Build GRCh38
rs113994127 Chr.1: 99851059-99851060 on Build GRCh38
rs568255289 Chr.1: 99850805-99850805 on Build GRCh38
rs1054599524 Chr.1: 99850885-99850885 on Build GRCh38
rs749324859 Chr.1: 99851171-99851171 on Build GRCh38
rs925817436 Chr.1: 99850897-99850897 on Build GRCh38
rs758178826 Chr.1: 99851133-99851133 on Build GRCh38
rs775012924 Chr.1: 99851103-99851103 on Build GRCh38
rs142331734 Chr.1: 99850856-99850857 on Build GRCh38
rs781592803 Chr.1: 99851118-99851118 on Build GRCh38
rs528149531 Chr.1: 99850808-99850810 on Build GRCh38
rs778848880 Chr.1: 99851047-99851047 on Build GRCh38
rs780347417 Chr.1: 99851150-99851150 on Build GRCh38
rs762292526 Chr.1: 99850995-99850995 on Build GRCh38
rs769898930 Chr.1: 99851080-99851080 on Build GRCh38
rs764738143 Chr.1: 99850991-99850992 on Build GRCh38
rs1057517057 Chr.1: 99851106-99851106 on Build GRCh38
rs1037108652 Chr.1: 99850758-99850758 on Build GRCh38
rs113994126 Chr.1: 99851058-99851058 on Build GRCh38
rs199701996 Chr.1: 99850997-99850997 on Build GRCh38
rs753243528 Chr.1: 99851117-99851117 on Build GRCh38
rs774816587 Chr.1: 99850988-99850988 on Build GRCh38
rs940072388 Chr.1: 99850737-99850737 on Build GRCh38
rs557247196 Chr.1: 99850840-99850840 on Build GRCh38
rs777480976 Chr.1: 99851142-99851142 on Build GRCh38
rs373822240 Chr.1: 99851018-99851018 on Build GRCh38
rs768864238 Chr.1: 99851173-99851173 on Build GRCh38
rs933996683 Chr.1: 99850785-99850785 on Build GRCh38
rs663848 Chr.1: 99851209-99851209 on Build GRCh38
rs973010478 Chr.1: 99851197-99851197 on Build GRCh38
rs752437446 Chr.1: 99851050-99851050 on Build GRCh38
rs915774407 Chr.1: 99850959-99850959 on Build GRCh38
rs746137555 Chr.1: 99851072-99851072 on Build GRCh38
rs998227893 Chr.1: 99850786-99850786 on Build GRCh38
rs991460303 Chr.1: 99850948-99850948 on Build GRCh38
rs374515839 Chr.1: 99850780-99850780 on Build GRCh38
rs756251291 Chr.1: 99851149-99851149 on Build GRCh38
rs997881890 Chr.1: 99851093-99851093 on Build GRCh38
rs762204351 Chr.1: 99851106-99851106 on Build GRCh38
rs2307130 Chr.1: 99851033-99851033 on Build GRCh38
rs890001672 Chr.1: 99850803-99850803 on Build GRCh38
rs891956210 Chr.1: 99850921-99850921 on Build GRCh38
rs758199444 Chr.1: 99851052-99851052 on Build GRCh38
rs957194478 Chr.1: 99850898-99850898 on Build GRCh38
rs758360729 Chr.1: 99850730-99850731 on Build GRCh38
rs961230782 Chr.1: 99851158-99851158 on Build GRCh38
rs138203039 Chr.1: 99851081-99851081 on Build GRCh38
rs1057516870 Chr.1: 99851064-99851064 on Build GRCh38
rs1049732612 Chr.1: 99851221-99851221 on Build GRCh38
rs762545015 Chr.1: 99851077-99851078 on Build GRCh38
rs750631960 Chr.1: 99851148-99851148 on Build GRCh38
rs143110759 Chr.1: 99851120-99851120 on Build GRCh38
rs764523804 Chr.1: 99850987-99850987 on Build GRCh38
rs377176234 Chr.1: 99850756-99850756 on Build GRCh38
rs548120281 Chr.1: 99850706-99850706 on Build GRCh38
rs754944003 Chr.1: 99851038-99851038 on Build GRCh38
rs750047257 Chr.1: 99851000-99851000 on Build GRCh38
rs553053029 Chr.1: 99851013-99851013 on Build GRCh38
rs967078774 Chr.1: 99850853-99850853 on Build GRCh38
rs794727255 Chr.1: 99851060-99851061 on Build GRCh38
rs772905291 Chr.1: 99851113-99851113 on Build GRCh38
rs537393462 Chr.1: 99850806-99850806 on Build GRCh38
rs934070517 Chr.1: 99851215-99851215 on Build GRCh38
rs769238334 Chr.1: 99851102-99851102 on Build GRCh38
rs752633408 Chr.1: 99851132-99851132 on Build GRCh38

View Full Product Details