Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160289925-160290353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPA
Entrez Gene ID
1314
Gene Name
coatomer protein complex subunit alpha
Gene Aliases
AILJK, HEP-COP
UniGene
Hs.162121
Gene Chromosome Location
Chr.1: 160288587-160343564 on Build GRCh38
Transcripts
NM_001098398.1 NM_004371.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5911275 c.3671G>T p.R1224L Chr.1: 160290161-160290161 on Build GRCh38
4995305 c.3617T>A p.V1206E Chr.1: 160290215-160290215 on Build GRCh38
1583654 c.3697C>T p.R1233C Chr.1: 160290162-160290162 on Build GRCh38
6122274 c.3651G>C p.E1217D Chr.1: 160290208-160290208 on Build GRCh38
898257 c.3670C>T p.R1224C Chr.1: 160290162-160290162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748610112 Chr.1: 160290319-160290319 on Build GRCh38
rs774816658 Chr.1: 160290022-160290022 on Build GRCh38
rs964022038 Chr.1: 160289993-160289993 on Build GRCh38
rs113237409 Chr.1: 160290260-160290260 on Build GRCh38
rs1056362134 Chr.1: 160290292-160290292 on Build GRCh38
rs1031925181 Chr.1: 160290320-160290320 on Build GRCh38
rs558169833 Chr.1: 160290328-160290328 on Build GRCh38
rs762917412 Chr.1: 160290157-160290157 on Build GRCh38
rs547416247 Chr.1: 160290000-160290000 on Build GRCh38
rs878890759 Chr.1: 160289952-160289952 on Build GRCh38
rs745581082 Chr.1: 160290124-160290124 on Build GRCh38
rs534690817 Chr.1: 160290314-160290314 on Build GRCh38
rs894575845 Chr.1: 160290230-160290232 on Build GRCh38
rs774710055 Chr.1: 160290162-160290162 on Build GRCh38
rs766622969 Chr.1: 160290161-160290161 on Build GRCh38
rs531161196 Chr.1: 160290050-160290050 on Build GRCh38
rs759764245 Chr.1: 160290167-160290167 on Build GRCh38
rs535699604 Chr.1: 160290136-160290136 on Build GRCh38
rs773805961 Chr.1: 160290154-160290154 on Build GRCh38
rs199663993 Chr.1: 160290227-160290227 on Build GRCh38
rs756710986 Chr.1: 160290252-160290252 on Build GRCh38
rs894734736 Chr.1: 160290322-160290322 on Build GRCh38
rs533080502 Chr.1: 160290093-160290093 on Build GRCh38
rs772437650 Chr.1: 160290233-160290236 on Build GRCh38
rs769007832 Chr.1: 160290125-160290125 on Build GRCh38
rs778464744 Chr.1: 160290259-160290259 on Build GRCh38
rs748562621 Chr.1: 160290140-160290140 on Build GRCh38
rs778517512 Chr.1: 160290120-160290120 on Build GRCh38
rs71485853 Chr.1: 160290243-160290244 on Build GRCh38
rs752491205 Chr.1: 160290186-160290186 on Build GRCh38
rs200071788 Chr.1: 160290119-160290119 on Build GRCh38
rs767865430 Chr.1: 160290171-160290171 on Build GRCh38
rs931248114 Chr.1: 160290049-160290049 on Build GRCh38
rs548159687 Chr.1: 160290102-160290102 on Build GRCh38
rs192179731 Chr.1: 160290114-160290114 on Build GRCh38
rs945140372 Chr.1: 160290163-160290163 on Build GRCh38
rs566002988 Chr.1: 160290309-160290309 on Build GRCh38
rs746304508 Chr.1: 160290225-160290228 on Build GRCh38
rs369317260 Chr.1: 160290236-160290236 on Build GRCh38
rs774868292 Chr.1: 160290222-160290223 on Build GRCh38
rs79708106 Chr.1: 160290224-160290224 on Build GRCh38
rs770155815 Chr.1: 160290151-160290151 on Build GRCh38
rs34264388 Chr.1: 160290197-160290198 on Build GRCh38
rs1028274644 Chr.1: 160290043-160290043 on Build GRCh38
rs753864677 Chr.1: 160290245-160290245 on Build GRCh38

View Full Product Details