Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150487083-150487552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TARS2
Entrez Gene ID
80222
Gene Name
threonyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD21, TARSL1, thrRS
UniGene
Hs.288974
Gene Chromosome Location
Chr.1: 150487364-150507609 on Build GRCh38
Transcripts
NM_001271895.1 NM_001271896.1 NM_025150.4 NR_073513.1 NR_073514.1 XM_006711555.1 XM_006711556.1 XM_017002394.1 XM_017002395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3740636 c.48G>T p.Q16H Chr.1: 150487498-150487498 on Build GRCh38
4805369 c.8T>A p.L3Q Chr.1: 150487458-150487458 on Build GRCh38
4788562 c.7C>T p.L3L Chr.1: 150487457-150487457 on Build GRCh38
4805370 c.8T>A p.L3Q Chr.1: 150487458-150487458 on Build GRCh38
4788561 c.7C>T p.L3L Chr.1: 150487457-150487457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76472322 Chr.1: 150487217-150487217 on Build GRCh38
rs116587387 Chr.1: 150487461-150487461 on Build GRCh38
rs184819264 Chr.1: 150487270-150487270 on Build GRCh38
rs908416259 Chr.1: 150487222-150487222 on Build GRCh38
rs747247447 Chr.1: 150487414-150487414 on Build GRCh38
rs1009609928 Chr.1: 150487367-150487367 on Build GRCh38
rs756811374 Chr.1: 150487454-150487454 on Build GRCh38
rs755138526 Chr.1: 150487429-150487430 on Build GRCh38
rs896260962 Chr.1: 150487363-150487363 on Build GRCh38
rs766358360 Chr.1: 150487212-150487212 on Build GRCh38
rs771904063 Chr.1: 150487151-150487151 on Build GRCh38
rs760387549 Chr.1: 150487206-150487206 on Build GRCh38
rs964120341 Chr.1: 150487197-150487197 on Build GRCh38
rs779884817 Chr.1: 150487413-150487413 on Build GRCh38
rs771827606 Chr.1: 150487412-150487412 on Build GRCh38
rs139414061 Chr.1: 150487516-150487516 on Build GRCh38
rs922584437 Chr.1: 150487207-150487207 on Build GRCh38
rs745679347 Chr.1: 150487225-150487225 on Build GRCh38
rs960145964 Chr.1: 150487484-150487484 on Build GRCh38
rs748959322 Chr.1: 150487511-150487511 on Build GRCh38
rs1047074983 Chr.1: 150487213-150487213 on Build GRCh38
rs201336268 Chr.1: 150487429-150487429 on Build GRCh38
rs181816509 Chr.1: 150487449-150487449 on Build GRCh38
rs746757094 Chr.1: 150487470-150487470 on Build GRCh38
rs897319573 Chr.1: 150487372-150487372 on Build GRCh38
rs1056114756 Chr.1: 150487345-150487345 on Build GRCh38
rs983180955 Chr.1: 150487467-150487467 on Build GRCh38
rs370477849 Chr.1: 150487427-150487427 on Build GRCh38
rs773331553 Chr.1: 150487507-150487507 on Build GRCh38
rs748347195 Chr.1: 150487489-150487489 on Build GRCh38
rs3840448 Chr.1: 150487418-150487421 on Build GRCh38
rs773635549 Chr.1: 150487428-150487428 on Build GRCh38
rs113460508 Chr.1: 150487378-150487378 on Build GRCh38
rs28372874 Chr.1: 150487420-150487423 on Build GRCh38
rs1001897742 Chr.1: 150487315-150487315 on Build GRCh38
rs955969795 Chr.1: 150487236-150487236 on Build GRCh38
rs980530644 Chr.1: 150487472-150487472 on Build GRCh38
rs587600628 Chr.1: 150487425-150487425 on Build GRCh38
rs780002234 Chr.1: 150487421-150487421 on Build GRCh38
rs587611228 Chr.1: 150487340-150487340 on Build GRCh38
rs773122392 Chr.1: 150487109-150487109 on Build GRCh38
rs955443982 Chr.1: 150487458-150487458 on Build GRCh38
rs370926339 Chr.1: 150487439-150487439 on Build GRCh38
rs200939116 Chr.1: 150487524-150487524 on Build GRCh38
rs1031491107 Chr.1: 150487226-150487226 on Build GRCh38
rs968024094 Chr.1: 150487384-150487384 on Build GRCh38
rs751897620 Chr.1: 150487432-150487432 on Build GRCh38
rs185148560 Chr.1: 150487522-150487522 on Build GRCh38
rs926173720 Chr.1: 150487194-150487194 on Build GRCh38
rs965757298 Chr.1: 150487216-150487216 on Build GRCh38
rs760324863 Chr.1: 150487437-150487437 on Build GRCh38
rs397844287 Chr.1: 150487218-150487218 on Build GRCh38
rs34001546 Chr.1: 150487239-150487239 on Build GRCh38
rs3840449 Chr.1: 150487217-150487217 on Build GRCh38
rs761932758 Chr.1: 150487426-150487426 on Build GRCh38
rs587694786 Chr.1: 150487515-150487515 on Build GRCh38
rs750384060 Chr.1: 150487455-150487455 on Build GRCh38
rs975591341 Chr.1: 150487443-150487443 on Build GRCh38
rs980094189 Chr.1: 150487275-150487275 on Build GRCh38
rs914619073 Chr.1: 150487249-150487249 on Build GRCh38
rs587714212 Chr.1: 150487322-150487322 on Build GRCh38
rs367864616 Chr.1: 150487499-150487499 on Build GRCh38
rs587725000 Chr.1: 150487423-150487423 on Build GRCh38
rs189677981 Chr.1: 150487406-150487406 on Build GRCh38
rs905059090 Chr.1: 150487297-150487297 on Build GRCh38
rs867315552 Chr.1: 150487473-150487473 on Build GRCh38
rs769017823 Chr.1: 150487474-150487474 on Build GRCh38
rs375885077 Chr.1: 150487410-150487410 on Build GRCh38
rs781611900 Chr.1: 150487488-150487488 on Build GRCh38
rs375480908 Chr.1: 150487465-150487465 on Build GRCh38
rs1020624669 Chr.1: 150487373-150487373 on Build GRCh38
rs587639300 Chr.1: 150487341-150487341 on Build GRCh38

View Full Product Details