Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 197913700-197914179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCG
Entrez Gene ID
84223
Gene Name
IQ motif containing G
Gene Aliases
CFAP122, DRC9
UniGene
Hs.591675
Gene Chromosome Location
Chr.3: 197889075-197960019 on Build GRCh38
Transcripts
NM_001134435.2 NM_001323027.1 NM_001323028.1 NM_001323029.1 NM_001323030.1 NM_032263.4 XM_017007316.1 XM_017007317.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7085211 c.880G>A p.E294K Chr.3: 197913889-197913889 on Build GRCh38
7118295 c.862G>C p.E288Q Chr.3: 197913907-197913907 on Build GRCh38
3380513 c.817A>G p.T273A Chr.3: 197913952-197913952 on Build GRCh38
4149817 c.865G>A p.E289K Chr.3: 197913904-197913904 on Build GRCh38
2948170 c.727-2A>T p.? Chr.3: 197914044-197914044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751524262 Chr.3: 197914143-197914143 on Build GRCh38
rs775989374 Chr.3: 197913828-197913828 on Build GRCh38
rs748706015 Chr.3: 197914011-197914011 on Build GRCh38
rs752719799 Chr.3: 197913922-197913922 on Build GRCh38
rs761843235 Chr.3: 197913850-197913850 on Build GRCh38
rs765344011 Chr.3: 197913854-197913854 on Build GRCh38
rs762951987 Chr.3: 197914031-197914031 on Build GRCh38
rs542122886 Chr.3: 197913818-197913818 on Build GRCh38
rs766569781 Chr.3: 197914041-197914041 on Build GRCh38
rs899192989 Chr.3: 197913839-197913839 on Build GRCh38
rs1024905046 Chr.3: 197914009-197914009 on Build GRCh38
rs1041650243 Chr.3: 197914117-197914117 on Build GRCh38
rs777534947 Chr.3: 197913805-197913805 on Build GRCh38
rs894568803 Chr.3: 197913936-197913936 on Build GRCh38
rs755713586 Chr.3: 197913801-197913801 on Build GRCh38
rs192649324 Chr.3: 197914049-197914049 on Build GRCh38
rs763297231 Chr.3: 197913886-197913886 on Build GRCh38
rs747339114 Chr.3: 197913831-197913831 on Build GRCh38
rs911785660 Chr.3: 197913790-197913791 on Build GRCh38
rs755662222 Chr.3: 197913933-197913933 on Build GRCh38
rs967035782 Chr.3: 197913727-197913727 on Build GRCh38
rs932075160 Chr.3: 197914062-197914062 on Build GRCh38
rs952275152 Chr.3: 197913874-197913874 on Build GRCh38
rs770454236 Chr.3: 197913918-197913918 on Build GRCh38
rs773287974 Chr.3: 197913864-197913864 on Build GRCh38
rs141338847 Chr.3: 197913969-197913969 on Build GRCh38
rs756741443 Chr.3: 197914077-197914077 on Build GRCh38
rs769042651 Chr.3: 197913988-197913988 on Build GRCh38
rs766087970 Chr.3: 197913898-197913898 on Build GRCh38
rs777399122 Chr.3: 197913940-197913940 on Build GRCh38
rs748888425 Chr.3: 197913947-197913947 on Build GRCh38
rs752666899 Chr.3: 197914057-197914057 on Build GRCh38
rs958448098 Chr.3: 197913773-197913773 on Build GRCh38
rs1055847515 Chr.3: 197913866-197913866 on Build GRCh38
rs778444674 Chr.3: 197913966-197913966 on Build GRCh38
rs1002575922 Chr.3: 197913746-197913746 on Build GRCh38
rs755315958 Chr.3: 197914091-197914091 on Build GRCh38
rs777145867 Chr.3: 197913846-197913846 on Build GRCh38
rs1040359452 Chr.3: 197913996-197913996 on Build GRCh38
rs760761004 Chr.3: 197914059-197914059 on Build GRCh38
rs201604300 Chr.3: 197914065-197914065 on Build GRCh38
rs762586334 Chr.3: 197913799-197913799 on Build GRCh38
rs767466541 Chr.3: 197914054-197914054 on Build GRCh38
rs774465798 Chr.3: 197914048-197914048 on Build GRCh38
rs138861904 Chr.3: 197914026-197914026 on Build GRCh38
rs773411538 Chr.3: 197914030-197914030 on Build GRCh38
rs200623949 Chr.3: 197913904-197913904 on Build GRCh38
rs112247427 Chr.3: 197913802-197913804 on Build GRCh38
rs769093997 Chr.3: 197913843-197913843 on Build GRCh38
rs751306501 Chr.3: 197913914-197913914 on Build GRCh38
rs766030244 Chr.3: 197913918-197913918 on Build GRCh38
rs748921663 Chr.3: 197913916-197913917 on Build GRCh38
rs199537552 Chr.3: 197914071-197914071 on Build GRCh38
rs990980863 Chr.3: 197913791-197913791 on Build GRCh38
rs953122482 Chr.3: 197914106-197914106 on Build GRCh38
rs1035405833 Chr.3: 197913770-197913770 on Build GRCh38
rs753332003 Chr.3: 197914072-197914072 on Build GRCh38
rs778590168 Chr.3: 197914078-197914078 on Build GRCh38
rs116790412 Chr.3: 197914103-197914103 on Build GRCh38
rs756867292 Chr.3: 197913964-197913964 on Build GRCh38
rs145134478 Chr.3: 197913998-197913998 on Build GRCh38
rs571593809 Chr.3: 197914139-197914139 on Build GRCh38
rs777094497 Chr.3: 197913990-197913990 on Build GRCh38
rs115346682 Chr.3: 197914070-197914070 on Build GRCh38
rs903110535 Chr.3: 197914129-197914129 on Build GRCh38
rs750118809 Chr.3: 197914080-197914080 on Build GRCh38

View Full Product Details