Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 96593439-96593975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDC2
Entrez Gene ID
6383
Gene Name
syndecan 2
Gene Aliases
CD362, HSPG, HSPG1, SYND2
UniGene
Hs.1501
Gene Chromosome Location
Chr.8: 96493654-96611809 on Build GRCh38
Transcripts
NM_002998.3 XM_011517212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5624710 c.82A>G p.K28E Chr.8: 96593501-96593501 on Build GRCh38
3835189 c.89T>C p.M30T Chr.8: 96593508-96593508 on Build GRCh38
1700388 c.170C>G p.S57W Chr.8: 96593589-96593589 on Build GRCh38
2792179 c.110T>C p.I37T Chr.8: 96593529-96593529 on Build GRCh38
5711056 c.119C>T p.A40V Chr.8: 96593538-96593538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762887653 Chr.8: 96593501-96593501 on Build GRCh38
rs928905302 Chr.8: 96593861-96593861 on Build GRCh38
rs752182500 Chr.8: 96593617-96593617 on Build GRCh38
rs377005291 Chr.8: 96593580-96593580 on Build GRCh38
rs905877769 Chr.8: 96593823-96593823 on Build GRCh38
rs763472339 Chr.8: 96593597-96593597 on Build GRCh38
rs2651476 Chr.8: 96593607-96593607 on Build GRCh38
rs141268636 Chr.8: 96593507-96593507 on Build GRCh38
rs950653725 Chr.8: 96593927-96593927 on Build GRCh38
rs755287762 Chr.8: 96593619-96593619 on Build GRCh38
rs183495161 Chr.8: 96593660-96593660 on Build GRCh38
rs1008247992 Chr.8: 96593703-96593703 on Build GRCh38
rs1051571797 Chr.8: 96593940-96593940 on Build GRCh38
rs749346319 Chr.8: 96593640-96593640 on Build GRCh38
rs1052691430 Chr.8: 96593583-96593583 on Build GRCh38
rs751788343 Chr.8: 96593662-96593662 on Build GRCh38
rs938913198 Chr.8: 96593891-96593891 on Build GRCh38
rs767413524 Chr.8: 96593590-96593590 on Build GRCh38
rs577445068 Chr.8: 96593908-96593908 on Build GRCh38
rs773116949 Chr.8: 96593494-96593494 on Build GRCh38
rs369529741 Chr.8: 96593757-96593757 on Build GRCh38
rs567673125 Chr.8: 96593629-96593629 on Build GRCh38
rs557616469 Chr.8: 96593838-96593838 on Build GRCh38
rs1014513928 Chr.8: 96593784-96593784 on Build GRCh38
rs753422501 Chr.8: 96593603-96593603 on Build GRCh38
rs150343338 Chr.8: 96593527-96593527 on Build GRCh38
rs755413425 Chr.8: 96593555-96593555 on Build GRCh38
rs755013779 Chr.8: 96593674-96593674 on Build GRCh38
rs755574295 Chr.8: 96593520-96593520 on Build GRCh38
rs766093366 Chr.8: 96593503-96593503 on Build GRCh38
rs759007672 Chr.8: 96593529-96593529 on Build GRCh38
rs1126681 Chr.8: 96593572-96593572 on Build GRCh38
rs970635205 Chr.8: 96593837-96593837 on Build GRCh38
rs760380434 Chr.8: 96593595-96593595 on Build GRCh38
rs900215114 Chr.8: 96593932-96593932 on Build GRCh38
rs1000759583 Chr.8: 96593756-96593756 on Build GRCh38
rs911560607 Chr.8: 96593654-96593654 on Build GRCh38
rs947195930 Chr.8: 96593901-96593901 on Build GRCh38
rs781627573 Chr.8: 96593624-96593624 on Build GRCh38
rs1026183576 Chr.8: 96593904-96593904 on Build GRCh38
rs1051373370 Chr.8: 96593649-96593649 on Build GRCh38
rs961514668 Chr.8: 96593726-96593726 on Build GRCh38
rs892040780 Chr.8: 96593776-96593776 on Build GRCh38
rs752434605 Chr.8: 96593512-96593512 on Build GRCh38
rs567190918 Chr.8: 96593681-96593681 on Build GRCh38
rs1027195053 Chr.8: 96593782-96593782 on Build GRCh38
rs1015582970 Chr.8: 96593704-96593704 on Build GRCh38
rs767400718 Chr.8: 96593548-96593548 on Build GRCh38
rs149448156 Chr.8: 96593933-96593933 on Build GRCh38
rs757217327 Chr.8: 96593942-96593942 on Build GRCh38
rs766963672 Chr.8: 96593610-96593610 on Build GRCh38
rs781271319 Chr.8: 96593559-96593559 on Build GRCh38
rs200211197 Chr.8: 96593566-96593566 on Build GRCh38
rs200340275 Chr.8: 96593481-96593481 on Build GRCh38
rs188138471 Chr.8: 96593946-96593946 on Build GRCh38
rs748248477 Chr.8: 96593628-96593628 on Build GRCh38
rs370731563 Chr.8: 96593508-96593508 on Build GRCh38
rs373803623 Chr.8: 96593564-96593564 on Build GRCh38
rs962854740 Chr.8: 96593864-96593864 on Build GRCh38
rs201901168 Chr.8: 96593538-96593538 on Build GRCh38
rs759218423 Chr.8: 96593589-96593589 on Build GRCh38
rs763674327 Chr.8: 96593522-96593522 on Build GRCh38
rs747359872 Chr.8: 96593549-96593549 on Build GRCh38
rs867762137 Chr.8: 96593744-96593744 on Build GRCh38
rs137986371 Chr.8: 96593563-96593563 on Build GRCh38
rs1024279653 Chr.8: 96593801-96593801 on Build GRCh38
rs912881541 Chr.8: 96593899-96593899 on Build GRCh38
rs116458570 Chr.8: 96593581-96593581 on Build GRCh38
rs988460239 Chr.8: 96593894-96593894 on Build GRCh38
rs748150403 Chr.8: 96593475-96593475 on Build GRCh38
rs192906471 Chr.8: 96593648-96593648 on Build GRCh38
rs947111222 Chr.8: 96593626-96593626 on Build GRCh38
rs565469070 Chr.8: 96593505-96593505 on Build GRCh38
rs756879826 Chr.8: 96593609-96593609 on Build GRCh38
rs763620841 Chr.8: 96593490-96593490 on Build GRCh38

View Full Product Details