Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154234480-154234829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBAP2L
Entrez Gene ID
9898
Gene Name
ubiquitin associated protein 2 like
Gene Aliases
NICE-4, NICE4
UniGene
Hs.490551
Gene Chromosome Location
Chr.1: 154220172-154271510 on Build GRCh38
Transcripts
NM_001127320.2 NM_001287815.1 NM_001287816.1 NM_014847.3 XM_005245658.3 XM_005245667.3 XM_005245668.3 XM_005245669.3 XM_005245670.3 XM_005245672.3 XM_005245673.3 XM_005245674.1 XM_011510215.2 XM_011510216.1 XM_017002972.1 XM_017002973.1 XM_017002974.1 XM_017002975.1 XM_017002976.1 XM_017002977.1 XM_017002978.1 XM_017002979.1 XM_017002980.1 XM_017002981.1 XM_017002982.1 XM_017002983.1 XM_017002984.1 XM_017002985.1 XM_017002986.1 XM_017002987.1 XM_017002988.1 XM_017002989.1 XM_017002990.1 XM_017002991.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4546545 c.401G>A p.R134H Chr.1: 154234712-154234712 on Build GRCh38
6459354 c.391A>G p.S131G Chr.1: 154234702-154234702 on Build GRCh38
6324746 c.371G>A p.R124Q Chr.1: 154234682-154234682 on Build GRCh38
3474621 c.352G>A p.E118K Chr.1: 154234663-154234663 on Build GRCh38
6324745 c.371G>A p.R124Q Chr.1: 154234682-154234682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376988539 Chr.1: 154234511-154234514 on Build GRCh38
rs543673230 Chr.1: 154234526-154234526 on Build GRCh38
rs750639357 Chr.1: 154234652-154234652 on Build GRCh38
rs939725092 Chr.1: 154234760-154234760 on Build GRCh38
rs374548186 Chr.1: 154234661-154234661 on Build GRCh38
rs766970460 Chr.1: 154234591-154234591 on Build GRCh38
rs201979564 Chr.1: 154234508-154234511 on Build GRCh38
rs74613943 Chr.1: 154234509-154234509 on Build GRCh38
rs768852366 Chr.1: 154234682-154234682 on Build GRCh38
rs763570821 Chr.1: 154234759-154234759 on Build GRCh38
rs773925753 Chr.1: 154234558-154234558 on Build GRCh38
rs777246805 Chr.1: 154234714-154234714 on Build GRCh38
rs775115634 Chr.1: 154234730-154234730 on Build GRCh38
rs78827015 Chr.1: 154234507-154234507 on Build GRCh38
rs978580818 Chr.1: 154234518-154234518 on Build GRCh38
rs770656735 Chr.1: 154234705-154234705 on Build GRCh38
rs771471058 Chr.1: 154234627-154234627 on Build GRCh38
rs761187218 Chr.1: 154234609-154234609 on Build GRCh38
rs779286424 Chr.1: 154234641-154234641 on Build GRCh38
rs774158418 Chr.1: 154234706-154234706 on Build GRCh38
rs759182413 Chr.1: 154234580-154234580 on Build GRCh38
rs1047962240 Chr.1: 154234585-154234585 on Build GRCh38
rs762909916 Chr.1: 154234551-154234551 on Build GRCh38
rs780010109 Chr.1: 154234664-154234664 on Build GRCh38
rs201442714 Chr.1: 154234562-154234562 on Build GRCh38
rs373213611 Chr.1: 154234555-154234555 on Build GRCh38
rs77358033 Chr.1: 154234512-154234512 on Build GRCh38
rs749218284 Chr.1: 154234691-154234691 on Build GRCh38
rs764648216 Chr.1: 154234611-154234611 on Build GRCh38
rs913817975 Chr.1: 154234702-154234702 on Build GRCh38
rs759119651 Chr.1: 154234719-154234719 on Build GRCh38
rs754308425 Chr.1: 154234624-154234624 on Build GRCh38
rs370921553 Chr.1: 154234608-154234608 on Build GRCh38
rs199924270 Chr.1: 154234687-154234687 on Build GRCh38
rs192836388 Chr.1: 154234553-154234553 on Build GRCh38
rs80096808 Chr.1: 154234513-154234513 on Build GRCh38
rs376260300 Chr.1: 154234587-154234587 on Build GRCh38
rs747145277 Chr.1: 154234665-154234665 on Build GRCh38
rs760214252 Chr.1: 154234748-154234748 on Build GRCh38
rs1035138872 Chr.1: 154234537-154234537 on Build GRCh38
rs994125901 Chr.1: 154234579-154234579 on Build GRCh38
rs771701488 Chr.1: 154234729-154234729 on Build GRCh38
rs985042195 Chr.1: 154234710-154234710 on Build GRCh38

View Full Product Details