Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63887344-63887925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPD52L2
Entrez Gene ID
7165
Gene Name
tumor protein D52 like 2
Gene Aliases
D54, TPD54
UniGene
Hs.473296
Gene Chromosome Location
Chr.20: 63865228-63891545 on Build GRCh38
Transcripts
NM_001243891.1 NM_001243892.1 NM_001243894.1 NM_001243895.1 NM_003288.3 NM_199359.2 NM_199360.2 NM_199361.2 NM_199362.2 NM_199363.2 NR_045090.1 XM_011529034.2 XM_011529035.2 XM_017028038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5807413 c.544A>T p.R182W Chr.20: 63887604-63887604 on Build GRCh38
1029197 c.538G>A p.A180T Chr.20: 63887598-63887598 on Build GRCh38
7269404 c.504-7C>T p.? Chr.20: 63887557-63887557 on Build GRCh38
6317732 c.534G>T p.M178I Chr.20: 63887594-63887594 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748533786 Chr.20: 63887547-63887547 on Build GRCh38
rs755072962 Chr.20: 63887629-63887629 on Build GRCh38
rs954668658 Chr.20: 63887444-63887444 on Build GRCh38
rs374716558 Chr.20: 63887520-63887520 on Build GRCh38
rs539407455 Chr.20: 63887848-63887848 on Build GRCh38
rs759155053 Chr.20: 63887596-63887596 on Build GRCh38
rs1003707525 Chr.20: 63887667-63887667 on Build GRCh38
rs921950202 Chr.20: 63887443-63887443 on Build GRCh38
rs572992381 Chr.20: 63887420-63887420 on Build GRCh38
rs745777364 Chr.20: 63887785-63887785 on Build GRCh38
rs112156967 Chr.20: 63887424-63887424 on Build GRCh38
rs376727122 Chr.20: 63887595-63887595 on Build GRCh38
rs372281047 Chr.20: 63887557-63887557 on Build GRCh38
rs546603083 Chr.20: 63887654-63887654 on Build GRCh38
rs949134149 Chr.20: 63887761-63887761 on Build GRCh38
rs781720725 Chr.20: 63887633-63887633 on Build GRCh38
rs776382756 Chr.20: 63887653-63887653 on Build GRCh38
rs912434413 Chr.20: 63887495-63887495 on Build GRCh38
rs79761489 Chr.20: 63887422-63887422 on Build GRCh38
rs1023797277 Chr.20: 63887402-63887402 on Build GRCh38
rs973012311 Chr.20: 63887512-63887512 on Build GRCh38
rs775344081 Chr.20: 63887610-63887610 on Build GRCh38
rs200097940 Chr.20: 63887527-63887527 on Build GRCh38
rs753079981 Chr.20: 63887639-63887639 on Build GRCh38
rs919771548 Chr.20: 63887693-63887693 on Build GRCh38
rs781318568 Chr.20: 63887779-63887779 on Build GRCh38
rs139901679 Chr.20: 63887553-63887553 on Build GRCh38
rs79604234 Chr.20: 63887575-63887575 on Build GRCh38
rs770674052 Chr.20: 63887644-63887644 on Build GRCh38
rs986727988 Chr.20: 63887471-63887471 on Build GRCh38
rs990535919 Chr.20: 63887752-63887752 on Build GRCh38
rs553611898 Chr.20: 63887753-63887753 on Build GRCh38
rs745354522 Chr.20: 63887647-63887647 on Build GRCh38
rs6011205 Chr.20: 63887534-63887534 on Build GRCh38
rs777333412 Chr.20: 63887660-63887660 on Build GRCh38
rs115196082 Chr.20: 63887688-63887688 on Build GRCh38
rs768673509 Chr.20: 63887652-63887652 on Build GRCh38
rs771727023 Chr.20: 63887653-63887654 on Build GRCh38
rs182111220 Chr.20: 63887627-63887627 on Build GRCh38
rs1054019869 Chr.20: 63887782-63887782 on Build GRCh38
rs879160409 Chr.20: 63887663-63887663 on Build GRCh38
rs767438764 Chr.20: 63887657-63887657 on Build GRCh38
rs1015361941 Chr.20: 63887892-63887892 on Build GRCh38
rs566610070 Chr.20: 63887672-63887672 on Build GRCh38
rs747037411 Chr.20: 63887776-63887776 on Build GRCh38
rs1013811499 Chr.20: 63887403-63887403 on Build GRCh38
rs1014924360 Chr.20: 63887674-63887674 on Build GRCh38
rs6011204 Chr.20: 63887410-63887410 on Build GRCh38
rs781600971 Chr.20: 63887537-63887537 on Build GRCh38
rs376139579 Chr.20: 63887651-63887651 on Build GRCh38
rs751848039 Chr.20: 63887655-63887655 on Build GRCh38
rs1012446291 Chr.20: 63887385-63887385 on Build GRCh38
rs778317923 Chr.20: 63887650-63887651 on Build GRCh38
rs554805538 Chr.20: 63887695-63887695 on Build GRCh38
rs186832187 Chr.20: 63887717-63887717 on Build GRCh38
rs754381629 Chr.20: 63887578-63887578 on Build GRCh38
rs578214901 Chr.20: 63887796-63887796 on Build GRCh38
rs550559293 Chr.20: 63887620-63887620 on Build GRCh38
rs369464684 Chr.20: 63887621-63887621 on Build GRCh38
rs369118030 Chr.20: 63887640-63887640 on Build GRCh38
rs961619311 Chr.20: 63887675-63887675 on Build GRCh38
rs1000065092 Chr.20: 63887418-63887418 on Build GRCh38
rs371717398 Chr.20: 63887519-63887519 on Build GRCh38
rs1046519061 Chr.20: 63887763-63887763 on Build GRCh38
rs558124314 Chr.20: 63887777-63887777 on Build GRCh38
rs533743559 Chr.20: 63887525-63887525 on Build GRCh38
rs745331955 Chr.20: 63887787-63887787 on Build GRCh38
rs367831892 Chr.20: 63887530-63887530 on Build GRCh38
rs778754519 Chr.20: 63887646-63887646 on Build GRCh38
rs761335673 Chr.20: 63887577-63887577 on Build GRCh38
rs966913321 Chr.20: 63887417-63887417 on Build GRCh38
rs939872258 Chr.20: 63887510-63887510 on Build GRCh38
rs751619067 Chr.20: 63887628-63887628 on Build GRCh38
rs576724977 Chr.20: 63887371-63887371 on Build GRCh38
rs376765531 Chr.20: 63887623-63887623 on Build GRCh38
rs773957956 Chr.20: 63887591-63887591 on Build GRCh38
rs954739240 Chr.20: 63887819-63887819 on Build GRCh38
rs931118496 Chr.20: 63887719-63887719 on Build GRCh38
rs3795156 Chr.20: 63887376-63887376 on Build GRCh38
rs767464842 Chr.20: 63887602-63887602 on Build GRCh38
rs200073648 Chr.20: 63887641-63887641 on Build GRCh38
rs765480249 Chr.20: 63887517-63887517 on Build GRCh38
rs372436734 Chr.20: 63887622-63887622 on Build GRCh38
rs747592166 Chr.20: 63887652-63887653 on Build GRCh38
rs190011010 Chr.20: 63887901-63887901 on Build GRCh38
rs1008970005 Chr.20: 63887869-63887869 on Build GRCh38
rs760511000 Chr.20: 63887615-63887615 on Build GRCh38
rs1054780577 Chr.20: 63887559-63887559 on Build GRCh38
rs1029085480 Chr.20: 63887829-63887829 on Build GRCh38
rs3795157 Chr.20: 63887413-63887413 on Build GRCh38

View Full Product Details