Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 37114732-37115041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2AK2
Entrez Gene ID
5610
Gene Name
eukaryotic translation initiation factor 2 alpha kinase 2
Gene Aliases
EIF2AK1, PKR, PPP1R83, PRKR
UniGene
Hs.131431
Gene Chromosome Location
Chr.2: 37099210-37157065 on Build GRCh38
Transcripts
NM_001135651.2 NM_001135652.2 NM_002759.3 XM_011532987.2 XM_017004503.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1641910 c.1343G>A p.S448N Chr.2: 37114765-37114765 on Build GRCh38
21215 c.1315C>G p.L439V Chr.2: 37114793-37114793 on Build GRCh38
4680531 c.1334G>A p.R445Q Chr.2: 37114774-37114774 on Build GRCh38
1724517 c.1249-6delT p.? Chr.2: 37114865-37114865 on Build GRCh38
5018941 c.1280A>C p.Q427P Chr.2: 37114828-37114828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768191204 Chr.2: 37114830-37114830 on Build GRCh38
rs372171549 Chr.2: 37114779-37114779 on Build GRCh38
rs753148148 Chr.2: 37114777-37114777 on Build GRCh38
rs757269448 Chr.2: 37114791-37114791 on Build GRCh38
rs767957226 Chr.2: 37114775-37114775 on Build GRCh38
rs575082112 Chr.2: 37115004-37115004 on Build GRCh38
rs907681710 Chr.2: 37114913-37114913 on Build GRCh38
rs56219559 Chr.2: 37114825-37114825 on Build GRCh38
rs200748103 Chr.2: 37114827-37114827 on Build GRCh38
rs1047529663 Chr.2: 37114882-37114882 on Build GRCh38
rs933079162 Chr.2: 37114859-37114859 on Build GRCh38
rs778985840 Chr.2: 37114798-37114798 on Build GRCh38
rs752996471 Chr.2: 37114887-37114887 on Build GRCh38
rs768194494 Chr.2: 37114885-37114885 on Build GRCh38
rs776281435 Chr.2: 37114834-37114834 on Build GRCh38
rs760092424 Chr.2: 37114876-37114876 on Build GRCh38
rs761575150 Chr.2: 37114863-37114863 on Build GRCh38
rs754007115 Chr.2: 37114786-37114786 on Build GRCh38
rs374584159 Chr.2: 37114845-37114845 on Build GRCh38
rs780387957 Chr.2: 37114864-37114865 on Build GRCh38
rs577436972 Chr.2: 37114774-37114774 on Build GRCh38
rs372081509 Chr.2: 37115007-37115007 on Build GRCh38
rs1002366902 Chr.2: 37115019-37115019 on Build GRCh38
rs745763401 Chr.2: 37114803-37114803 on Build GRCh38
rs769383677 Chr.2: 37114864-37114864 on Build GRCh38
rs746733915 Chr.2: 37114776-37114776 on Build GRCh38
rs975022412 Chr.2: 37114783-37114783 on Build GRCh38
rs768511306 Chr.2: 37114865-37114865 on Build GRCh38
rs772883798 Chr.2: 37114875-37114875 on Build GRCh38
rs776392243 Chr.2: 37114977-37114977 on Build GRCh38
rs779693187 Chr.2: 37114826-37114826 on Build GRCh38
rs752059539 Chr.2: 37114767-37114767 on Build GRCh38
rs764844778 Chr.2: 37114765-37114765 on Build GRCh38
rs772035707 Chr.2: 37114818-37114818 on Build GRCh38
rs907508502 Chr.2: 37114927-37114927 on Build GRCh38
rs920285723 Chr.2: 37114962-37114962 on Build GRCh38
rs778120333 Chr.2: 37114780-37114780 on Build GRCh38
rs376714065 Chr.2: 37115010-37115010 on Build GRCh38
rs572363125 Chr.2: 37114914-37114915 on Build GRCh38
rs1037606819 Chr.2: 37114916-37114916 on Build GRCh38
rs201097593 Chr.2: 37114901-37114901 on Build GRCh38
rs75744939 Chr.2: 37114865-37114865 on Build GRCh38
rs556845762 Chr.2: 37114937-37114937 on Build GRCh38
rs1038575972 Chr.2: 37114970-37114970 on Build GRCh38
rs958720606 Chr.2: 37114824-37114824 on Build GRCh38

View Full Product Details