Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127502469-127502931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IWS1
Entrez Gene ID
55677
Gene Name
IWS1, SUPT6H interacting protein
Gene Aliases
-
UniGene
Hs.469879
Gene Chromosome Location
Chr.2: 127480807-127526886 on Build GRCh38
Transcripts
NM_017969.2 XM_005263707.3 XM_006712626.3 XM_011511450.1 XM_011511452.1 XM_017004467.1 XM_017004468.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5553494 c.1459G>T p.E487* Chr.2: 127502823-127502823 on Build GRCh38
3042354 c.1410A>G p.E470E Chr.2: 127502872-127502872 on Build GRCh38
441083 c.1447G>A p.E483K Chr.2: 127502835-127502835 on Build GRCh38
3713373 c.1435G>C p.E479Q Chr.2: 127502847-127502847 on Build GRCh38
1727926 c.1415T>G p.L472R Chr.2: 127502867-127502867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962412901 Chr.2: 127502754-127502754 on Build GRCh38
rs776362277 Chr.2: 127502808-127502808 on Build GRCh38
rs918720808 Chr.2: 127502594-127502594 on Build GRCh38
rs958545149 Chr.2: 127502725-127502725 on Build GRCh38
rs1000567399 Chr.2: 127502655-127502655 on Build GRCh38
rs1041767632 Chr.2: 127502712-127502712 on Build GRCh38
rs765434753 Chr.2: 127502830-127502830 on Build GRCh38
rs761527694 Chr.2: 127502815-127502815 on Build GRCh38
rs771547564 Chr.2: 127502794-127502794 on Build GRCh38
rs930046297 Chr.2: 127502601-127502601 on Build GRCh38
rs750675141 Chr.2: 127502844-127502844 on Build GRCh38
rs746445411 Chr.2: 127502708-127502708 on Build GRCh38
rs963263876 Chr.2: 127502736-127502736 on Build GRCh38
rs1033107103 Chr.2: 127502668-127502668 on Build GRCh38
rs950265596 Chr.2: 127502606-127502606 on Build GRCh38
rs11883835 Chr.2: 127502677-127502677 on Build GRCh38
rs1015253145 Chr.2: 127502740-127502740 on Build GRCh38
rs150498536 Chr.2: 127502732-127502732 on Build GRCh38
rs745308184 Chr.2: 127502791-127502791 on Build GRCh38
rs77979885 Chr.2: 127502560-127502560 on Build GRCh38
rs568791400 Chr.2: 127502685-127502685 on Build GRCh38
rs950317309 Chr.2: 127502790-127502790 on Build GRCh38
rs77970345 Chr.2: 127502651-127502651 on Build GRCh38
rs770660316 Chr.2: 127502578-127502581 on Build GRCh38
rs764050684 Chr.2: 127502807-127502807 on Build GRCh38
rs779490599 Chr.2: 127502768-127502769 on Build GRCh38
rs748551434 Chr.2: 127502776-127502776 on Build GRCh38
rs143420139 Chr.2: 127502720-127502720 on Build GRCh38
rs1039985563 Chr.2: 127502699-127502699 on Build GRCh38
rs766415194 Chr.2: 127502872-127502872 on Build GRCh38
rs1039891872 Chr.2: 127502889-127502889 on Build GRCh38
rs376127727 Chr.2: 127502899-127502899 on Build GRCh38
rs755505938 Chr.2: 127502894-127502894 on Build GRCh38
rs546318732 Chr.2: 127502579-127502579 on Build GRCh38
rs776605491 Chr.2: 127502805-127502805 on Build GRCh38
rs561364104 Chr.2: 127502559-127502559 on Build GRCh38
rs11890544 Chr.2: 127502585-127502585 on Build GRCh38
rs752077949 Chr.2: 127502886-127502886 on Build GRCh38
rs757933480 Chr.2: 127502769-127502769 on Build GRCh38
rs865945619 Chr.2: 127502868-127502868 on Build GRCh38
rs796333917 Chr.2: 127502528-127502528 on Build GRCh38
rs1041771400 Chr.2: 127502621-127502621 on Build GRCh38
rs770565055 Chr.2: 127502777-127502777 on Build GRCh38
rs138376919 Chr.2: 127502759-127502759 on Build GRCh38
rs755504687 Chr.2: 127502821-127502823 on Build GRCh38
rs773885269 Chr.2: 127502784-127502784 on Build GRCh38
rs755376131 Chr.2: 127502764-127502764 on Build GRCh38
rs893351768 Chr.2: 127502704-127502704 on Build GRCh38
rs376103491 Chr.2: 127502806-127502806 on Build GRCh38
rs533950933 Chr.2: 127502870-127502870 on Build GRCh38
rs781721297 Chr.2: 127502769-127502769 on Build GRCh38
rs1051302700 Chr.2: 127502654-127502654 on Build GRCh38
rs1035421330 Chr.2: 127502722-127502722 on Build GRCh38
rs1056343715 Chr.2: 127502586-127502586 on Build GRCh38
rs941374805 Chr.2: 127502567-127502567 on Build GRCh38
rs987300478 Chr.2: 127502780-127502780 on Build GRCh38
rs752073534 Chr.2: 127502785-127502787 on Build GRCh38
rs902960838 Chr.2: 127502714-127502714 on Build GRCh38
rs1012670525 Chr.2: 127502730-127502730 on Build GRCh38

View Full Product Details