Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31189534-31190093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUS
Entrez Gene ID
2521
Gene Name
FUS RNA binding protein
Gene Aliases
ALS6, ETM4, FUS1, HNRNPP2, POMP75, TLS
UniGene
Hs.46894
Gene Chromosome Location
Chr.16: 31179444-31194871 on Build GRCh38
Transcripts
NM_001170634.1 NM_001170937.1 NM_004960.3 NR_028388.2 XM_005255233.4 XM_011545781.1 XM_011545782.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7173707 c.1051A>G p.I351V Chr.16: 31189779-31189779 on Build GRCh38
6212401 c.944A>G p.K315R Chr.16: 31189672-31189672 on Build GRCh38
5715566 c.1019C>T p.S340F Chr.16: 31189747-31189747 on Build GRCh38
4421092 c.1111C>T p.R371C Chr.16: 31190084-31190084 on Build GRCh38
4458485 c.1087C>T p.P363S Chr.16: 31190060-31190060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779527584 Chr.16: 31189809-31189809 on Build GRCh38
rs749604252 Chr.16: 31189667-31189667 on Build GRCh38
rs778616596 Chr.16: 31189907-31189907 on Build GRCh38
rs141404934 Chr.16: 31189965-31189965 on Build GRCh38
rs368342528 Chr.16: 31189636-31189636 on Build GRCh38
rs748632640 Chr.16: 31189640-31189640 on Build GRCh38
rs769791495 Chr.16: 31190065-31190065 on Build GRCh38
rs765049641 Chr.16: 31189806-31189806 on Build GRCh38
rs771737335 Chr.16: 31189987-31189987 on Build GRCh38
rs770393324 Chr.16: 31189645-31189645 on Build GRCh38
rs765952158 Chr.16: 31190009-31190009 on Build GRCh38
rs753723545 Chr.16: 31189766-31189766 on Build GRCh38
rs1045011834 Chr.16: 31189600-31189600 on Build GRCh38
rs917771084 Chr.16: 31189595-31189595 on Build GRCh38
rs778289704 Chr.16: 31189815-31189815 on Build GRCh38
rs865858018 Chr.16: 31189956-31189956 on Build GRCh38
rs940106398 Chr.16: 31189882-31189882 on Build GRCh38
rs759863892 Chr.16: 31189688-31189688 on Build GRCh38
rs556693665 Chr.16: 31189973-31189973 on Build GRCh38
rs748104616 Chr.16: 31190062-31190062 on Build GRCh38
rs756855832 Chr.16: 31189622-31189622 on Build GRCh38
rs935935656 Chr.16: 31189840-31189840 on Build GRCh38
rs928676757 Chr.16: 31189852-31189852 on Build GRCh38
rs777366298 Chr.16: 31189995-31189995 on Build GRCh38
rs754943057 Chr.16: 31189888-31189888 on Build GRCh38
rs753722607 Chr.16: 31189621-31189621 on Build GRCh38
rs200498893 Chr.16: 31189816-31189816 on Build GRCh38
rs772245489 Chr.16: 31189700-31189700 on Build GRCh38
rs184847010 Chr.16: 31189925-31189925 on Build GRCh38
rs780041820 Chr.16: 31189607-31189607 on Build GRCh38
rs1038385142 Chr.16: 31189628-31189628 on Build GRCh38
rs997071883 Chr.16: 31189641-31189641 on Build GRCh38
rs369370003 Chr.16: 31189811-31189811 on Build GRCh38
rs200156095 Chr.16: 31189837-31189837 on Build GRCh38
rs202036311 Chr.16: 31190016-31190016 on Build GRCh38
rs78590909 Chr.16: 31189856-31189856 on Build GRCh38
rs72550865 Chr.16: 31189988-31189988 on Build GRCh38
rs887212547 Chr.16: 31189875-31189875 on Build GRCh38
rs185933137 Chr.16: 31190014-31190014 on Build GRCh38
rs949313993 Chr.16: 31189598-31189598 on Build GRCh38
rs541962931 Chr.16: 31189916-31189916 on Build GRCh38
rs111515661 Chr.16: 31189625-31189625 on Build GRCh38
rs538248567 Chr.16: 31189972-31189972 on Build GRCh38
rs200960093 Chr.16: 31189831-31189831 on Build GRCh38
rs1038353976 Chr.16: 31189898-31189898 on Build GRCh38
rs757705019 Chr.16: 31189660-31189660 on Build GRCh38
rs528001562 Chr.16: 31189892-31189892 on Build GRCh38
rs775757526 Chr.16: 31189707-31189707 on Build GRCh38
rs1018594566 Chr.16: 31189955-31189955 on Build GRCh38
rs145232220 Chr.16: 31190059-31190059 on Build GRCh38
rs771216742 Chr.16: 31189679-31189679 on Build GRCh38
rs189774909 Chr.16: 31189970-31189970 on Build GRCh38
rs773049680 Chr.16: 31189998-31189998 on Build GRCh38
rs917724283 Chr.16: 31189865-31189865 on Build GRCh38
rs778470554 Chr.16: 31189626-31189626 on Build GRCh38
rs567135495 Chr.16: 31189891-31189891 on Build GRCh38
rs964288416 Chr.16: 31189980-31189980 on Build GRCh38
rs369794114 Chr.16: 31189845-31189845 on Build GRCh38
rs1045727628 Chr.16: 31189873-31189873 on Build GRCh38
rs370262636 Chr.16: 31189629-31189629 on Build GRCh38
rs747218305 Chr.16: 31190036-31190036 on Build GRCh38
rs181287335 Chr.16: 31190004-31190004 on Build GRCh38
rs374933969 Chr.16: 31189742-31189742 on Build GRCh38
rs190724342 Chr.16: 31190053-31190053 on Build GRCh38
rs774663066 Chr.16: 31189682-31189682 on Build GRCh38
rs761610757 Chr.16: 31189778-31189778 on Build GRCh38
rs754452897 Chr.16: 31189813-31189813 on Build GRCh38
rs779490965 Chr.16: 31189671-31189673 on Build GRCh38
rs955597548 Chr.16: 31189659-31189659 on Build GRCh38
rs887190183 Chr.16: 31189581-31189581 on Build GRCh38
rs779456444 Chr.16: 31189822-31189822 on Build GRCh38
rs775706022 Chr.16: 31189833-31189833 on Build GRCh38
rs536436219 Chr.16: 31189993-31189993 on Build GRCh38
rs779404384 Chr.16: 31190033-31190033 on Build GRCh38
rs760516023 Chr.16: 31189724-31189724 on Build GRCh38
rs1009019707 Chr.16: 31189943-31189943 on Build GRCh38
rs757864142 Chr.16: 31189631-31189631 on Build GRCh38
rs750742445 Chr.16: 31190018-31190020 on Build GRCh38
rs150914517 Chr.16: 31189996-31189996 on Build GRCh38
rs34752278 Chr.16: 31189699-31189700 on Build GRCh38
rs915866777 Chr.16: 31190000-31190000 on Build GRCh38
rs898640137 Chr.16: 31189902-31189902 on Build GRCh38
rs920098950 Chr.16: 31190052-31190052 on Build GRCh38
rs754335560 Chr.16: 31190027-31190027 on Build GRCh38
rs144170641 Chr.16: 31189921-31189922 on Build GRCh38
rs950126347 Chr.16: 31189871-31189871 on Build GRCh38
rs367842783 Chr.16: 31189808-31189808 on Build GRCh38
rs879237367 Chr.16: 31189830-31189830 on Build GRCh38
rs768630899 Chr.16: 31189838-31189838 on Build GRCh38
rs77813265 Chr.16: 31190034-31190034 on Build GRCh38
rs1045375982 Chr.16: 31189877-31189880 on Build GRCh38
rs766964324 Chr.16: 31190024-31190024 on Build GRCh38
rs1025790649 Chr.16: 31189658-31189658 on Build GRCh38
rs199705472 Chr.16: 31189655-31189655 on Build GRCh38
rs891370741 Chr.16: 31189935-31189935 on Build GRCh38
rs758121125 Chr.16: 31190023-31190023 on Build GRCh38
rs941927129 Chr.16: 31189896-31189897 on Build GRCh38
rs372038288 Chr.16: 31189646-31189646 on Build GRCh38
rs543862509 Chr.16: 31189818-31189818 on Build GRCh38
rs780265649 Chr.16: 31190035-31190035 on Build GRCh38

View Full Product Details