Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47951668-47952233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR15L
Entrez Gene ID
79170
Gene Name
proline rich 15 like
Gene Aliases
ATAD4
UniGene
Hs.368260
Gene Chromosome Location
Chr.17: 47951967-47957905 on Build GRCh38
Transcripts
NM_024320.3 XM_005257663.4 XM_005257664.2 XM_005257665.1 XM_005257666.4

SNP Detail

SNP ID Chromosome Location
rs190847382 Chr.17: 47952059-47952059 on Build GRCh38
rs966675157 Chr.17: 47952211-47952211 on Build GRCh38
rs552855412 Chr.17: 47951762-47951762 on Build GRCh38
rs576202760 Chr.17: 47952051-47952051 on Build GRCh38
rs980075979 Chr.17: 47951816-47951816 on Build GRCh38
rs115159645 Chr.17: 47952000-47952000 on Build GRCh38
rs11651753 Chr.17: 47951723-47951723 on Build GRCh38
rs529724738 Chr.17: 47952195-47952195 on Build GRCh38
rs1039312225 Chr.17: 47951927-47951927 on Build GRCh38
rs769059739 Chr.17: 47952170-47952170 on Build GRCh38
rs1047856590 Chr.17: 47951994-47951994 on Build GRCh38
rs950460061 Chr.17: 47951789-47951789 on Build GRCh38
rs568565201 Chr.17: 47951897-47951897 on Build GRCh38
rs572287394 Chr.17: 47952092-47952092 on Build GRCh38
rs982225478 Chr.17: 47952176-47952176 on Build GRCh38
rs756768799 Chr.17: 47951825-47951825 on Build GRCh38
rs749654140 Chr.17: 47952078-47952078 on Build GRCh38
rs1005939699 Chr.17: 47951793-47951793 on Build GRCh38
rs1043789206 Chr.17: 47951879-47951879 on Build GRCh38
rs946600554 Chr.17: 47951896-47951896 on Build GRCh38
rs1035502864 Chr.17: 47952102-47952102 on Build GRCh38
rs35611198 Chr.17: 47952197-47952198 on Build GRCh38
rs750956178 Chr.17: 47951942-47951942 on Build GRCh38
rs933559532 Chr.17: 47951952-47951952 on Build GRCh38
rs537572684 Chr.17: 47951987-47951987 on Build GRCh38
rs905720889 Chr.17: 47952076-47952076 on Build GRCh38
rs754079049 Chr.17: 47952067-47952067 on Build GRCh38
rs535027254 Chr.17: 47951802-47951802 on Build GRCh38
rs893703431 Chr.17: 47951878-47951878 on Build GRCh38
rs543128024 Chr.17: 47952202-47952202 on Build GRCh38
rs935338738 Chr.17: 47951819-47951819 on Build GRCh38
rs541824916 Chr.17: 47952003-47952003 on Build GRCh38
rs565888686 Chr.17: 47951766-47951766 on Build GRCh38
rs1006184150 Chr.17: 47952077-47952077 on Build GRCh38
rs540618129 Chr.17: 47952146-47952146 on Build GRCh38
rs945467848 Chr.17: 47951871-47951871 on Build GRCh38
rs992584015 Chr.17: 47951856-47951856 on Build GRCh38
rs1027710320 Chr.17: 47952063-47952063 on Build GRCh38
rs3096 Chr.17: 47952066-47952066 on Build GRCh38
rs779099995 Chr.17: 47952209-47952209 on Build GRCh38
rs755296345 Chr.17: 47952084-47952084 on Build GRCh38
rs913872275 Chr.17: 47951859-47951859 on Build GRCh38
rs928020245 Chr.17: 47952200-47952200 on Build GRCh38
rs1008642922 Chr.17: 47951973-47951973 on Build GRCh38
rs56273045 Chr.17: 47951849-47951849 on Build GRCh38
rs187880934 Chr.17: 47951949-47951949 on Build GRCh38
rs1016106429 Chr.17: 47951809-47951809 on Build GRCh38
rs902111462 Chr.17: 47951939-47951939 on Build GRCh38
rs560576757 Chr.17: 47952193-47952193 on Build GRCh38
rs969875214 Chr.17: 47951815-47951815 on Build GRCh38
rs943528050 Chr.17: 47951925-47951925 on Build GRCh38

View Full Product Details